Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58788136-58788676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_011540191.2 XM_017016677.1 XM_017016678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
41469 c.547-3T>C p.? Chr.10: 58788367-58788367 on Build GRCh38
6066270 c.571G>C p.G191R Chr.10: 58788394-58788394 on Build GRCh38
4626670 c.558G>A p.A186A Chr.10: 58788381-58788381 on Build GRCh38
919157 c.557C>T p.A186V Chr.10: 58788380-58788380 on Build GRCh38
919158 c.598C>T p.R200W Chr.10: 58788421-58788421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752904680 Chr.10: 58788390-58788390 on Build GRCh38
rs375917847 Chr.10: 58788376-58788376 on Build GRCh38
rs1006471918 Chr.10: 58788348-58788348 on Build GRCh38
rs1011896130 Chr.10: 58788168-58788168 on Build GRCh38
rs1035741792 Chr.10: 58788421-58788421 on Build GRCh38
rs78539120 Chr.10: 58788164-58788164 on Build GRCh38
rs537252238 Chr.10: 58788643-58788643 on Build GRCh38
rs976833386 Chr.10: 58788558-58788558 on Build GRCh38
rs552699593 Chr.10: 58788650-58788650 on Build GRCh38
rs776206792 Chr.10: 58788450-58788450 on Build GRCh38
rs547875114 Chr.10: 58788520-58788520 on Build GRCh38
rs547096611 Chr.10: 58788438-58788438 on Build GRCh38
rs922338184 Chr.10: 58788600-58788600 on Build GRCh38
rs751710629 Chr.10: 58788377-58788377 on Build GRCh38
rs200873651 Chr.10: 58788460-58788460 on Build GRCh38
rs542327714 Chr.10: 58788275-58788275 on Build GRCh38
rs778650709 Chr.10: 58788410-58788410 on Build GRCh38
rs191601281 Chr.10: 58788178-58788178 on Build GRCh38
rs779930565 Chr.10: 58788437-58788437 on Build GRCh38
rs935138790 Chr.10: 58788502-58788502 on Build GRCh38
rs374301012 Chr.10: 58788339-58788339 on Build GRCh38
rs757842688 Chr.10: 58788493-58788493 on Build GRCh38
rs913109504 Chr.10: 58788175-58788175 on Build GRCh38
rs968550609 Chr.10: 58788504-58788504 on Build GRCh38
rs910875531 Chr.10: 58788251-58788251 on Build GRCh38
rs34367678 Chr.10: 58788606-58788606 on Build GRCh38
rs566408991 Chr.10: 58788258-58788258 on Build GRCh38
rs931427597 Chr.10: 58788637-58788637 on Build GRCh38
rs373594414 Chr.10: 58788344-58788344 on Build GRCh38
rs1040776183 Chr.10: 58788262-58788262 on Build GRCh38
rs760188085 Chr.10: 58788320-58788320 on Build GRCh38
rs981581616 Chr.10: 58788179-58788179 on Build GRCh38
rs906292680 Chr.10: 58788636-58788636 on Build GRCh38
rs763520535 Chr.10: 58788375-58788375 on Build GRCh38
rs766349799 Chr.10: 58788429-58788432 on Build GRCh38
rs575007987 Chr.10: 58788271-58788271 on Build GRCh38
rs376626098 Chr.10: 58788381-58788381 on Build GRCh38
rs978629661 Chr.10: 58788505-58788505 on Build GRCh38
rs893700047 Chr.10: 58788539-58788539 on Build GRCh38
rs35224302 Chr.10: 58788311-58788312 on Build GRCh38
rs770320369 Chr.10: 58788475-58788475 on Build GRCh38
rs774962089 Chr.10: 58788319-58788319 on Build GRCh38
rs911147857 Chr.10: 58788515-58788515 on Build GRCh38
rs141141471 Chr.10: 58788372-58788372 on Build GRCh38
rs745857191 Chr.10: 58788423-58788423 on Build GRCh38
rs768228261 Chr.10: 58788325-58788325 on Build GRCh38
rs911323619 Chr.10: 58788406-58788406 on Build GRCh38
rs781271366 Chr.10: 58788386-58788386 on Build GRCh38
rs985456029 Chr.10: 58788391-58788391 on Build GRCh38
rs757054959 Chr.10: 58788397-58788397 on Build GRCh38
rs929710372 Chr.10: 58788610-58788610 on Build GRCh38
rs532399999 Chr.10: 58788430-58788430 on Build GRCh38
rs1037990030 Chr.10: 58788360-58788360 on Build GRCh38
rs7071051 Chr.10: 58788290-58788290 on Build GRCh38
rs569964693 Chr.10: 58788627-58788627 on Build GRCh38
rs576773850 Chr.10: 58788568-58788568 on Build GRCh38
rs747218993 Chr.10: 58788226-58788226 on Build GRCh38
rs576299352 Chr.10: 58788331-58788331 on Build GRCh38
rs11812567 Chr.10: 58788516-58788516 on Build GRCh38
rs1012337381 Chr.10: 58788551-58788551 on Build GRCh38

View Full Product Details