Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68230922-68231431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH7
Entrez Gene ID
220202
Gene Name
atonal bHLH transcription factor 7
Gene Aliases
Math5, NCRNA, PHPVAR, RNANC, bHLHa13
UniGene
Hs.175396
Gene Chromosome Location
Chr.10: 68230595-68232113 on Build GRCh38
Transcripts
NM_145178.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5738914 c.247C>T p.Q83* Chr.10: 68231431-68231431 on Build GRCh38
1183977 c.438G>T p.E146D Chr.10: 68231240-68231240 on Build GRCh38
5676723 c.297_301delCGAGC p.E100fs*13 Chr.10: 68231377-68231381 on Build GRCh38
5505302 c.258G>T p.L86L Chr.10: 68231420-68231420 on Build GRCh38
5505301 c.259A>T p.S87C Chr.10: 68231419-68231419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913837209 Chr.10: 68231049-68231049 on Build GRCh38
rs539429596 Chr.10: 68231098-68231098 on Build GRCh38
rs566619619 Chr.10: 68231173-68231173 on Build GRCh38
rs201955526 Chr.10: 68231342-68231342 on Build GRCh38
rs774680912 Chr.10: 68231188-68231188 on Build GRCh38
rs371394364 Chr.10: 68231205-68231205 on Build GRCh38
rs979283464 Chr.10: 68231229-68231229 on Build GRCh38
rs754075380 Chr.10: 68231209-68231209 on Build GRCh38
rs529045217 Chr.10: 68230962-68230962 on Build GRCh38
rs780545737 Chr.10: 68231331-68231331 on Build GRCh38
rs932331777 Chr.10: 68231145-68231145 on Build GRCh38
rs751407372 Chr.10: 68231217-68231217 on Build GRCh38
rs776797595 Chr.10: 68231274-68231274 on Build GRCh38
rs767426217 Chr.10: 68231303-68231303 on Build GRCh38
rs762047633 Chr.10: 68231276-68231276 on Build GRCh38
rs146898613 Chr.10: 68231285-68231285 on Build GRCh38
rs372403899 Chr.10: 68231110-68231110 on Build GRCh38
rs764447991 Chr.10: 68231273-68231273 on Build GRCh38
rs760740816 Chr.10: 68231404-68231404 on Build GRCh38
rs368934719 Chr.10: 68231282-68231282 on Build GRCh38
rs575314578 Chr.10: 68231310-68231310 on Build GRCh38
rs750560313 Chr.10: 68231284-68231284 on Build GRCh38
rs912285318 Chr.10: 68231219-68231219 on Build GRCh38
rs548790876 Chr.10: 68231005-68231005 on Build GRCh38
rs993462335 Chr.10: 68230950-68230950 on Build GRCh38
rs368807165 Chr.10: 68231369-68231369 on Build GRCh38
rs991961653 Chr.10: 68231031-68231031 on Build GRCh38
rs747549064 Chr.10: 68231332-68231332 on Build GRCh38
rs760110611 Chr.10: 68231195-68231195 on Build GRCh38
rs1027580992 Chr.10: 68230961-68230961 on Build GRCh38
rs906154183 Chr.10: 68231024-68231024 on Build GRCh38
rs994771090 Chr.10: 68231391-68231391 on Build GRCh38
rs760882760 Chr.10: 68231199-68231199 on Build GRCh38
rs568628212 Chr.10: 68231015-68231015 on Build GRCh38
rs555446400 Chr.10: 68231223-68231223 on Build GRCh38
rs773961010 Chr.10: 68231252-68231252 on Build GRCh38
rs201162238 Chr.10: 68231322-68231322 on Build GRCh38
rs952074196 Chr.10: 68230986-68230986 on Build GRCh38
rs1005821943 Chr.10: 68231326-68231326 on Build GRCh38
rs772704355 Chr.10: 68231255-68231255 on Build GRCh38
rs544124160 Chr.10: 68231156-68231156 on Build GRCh38
rs139387078 Chr.10: 68231406-68231406 on Build GRCh38
rs550959111 Chr.10: 68231054-68231054 on Build GRCh38
rs769635297 Chr.10: 68231175-68231175 on Build GRCh38
rs748745867 Chr.10: 68231183-68231183 on Build GRCh38
rs772832904 Chr.10: 68231179-68231179 on Build GRCh38
rs895923196 Chr.10: 68231272-68231272 on Build GRCh38
rs531157640 Chr.10: 68231050-68231050 on Build GRCh38
rs535518735 Chr.10: 68231215-68231215 on Build GRCh38
rs955136135 Chr.10: 68231159-68231159 on Build GRCh38
rs755983779 Chr.10: 68231237-68231237 on Build GRCh38
rs748029058 Chr.10: 68231234-68231234 on Build GRCh38
rs763021073 Chr.10: 68231372-68231372 on Build GRCh38
rs771498531 Chr.10: 68231329-68231329 on Build GRCh38
rs1053582980 Chr.10: 68230978-68230978 on Build GRCh38
rs767751252 Chr.10: 68231198-68231198 on Build GRCh38
rs961816948 Chr.10: 68231091-68231091 on Build GRCh38
rs571158822 Chr.10: 68231097-68231097 on Build GRCh38
rs746329264 Chr.10: 68231253-68231253 on Build GRCh38
rs770514524 Chr.10: 68231245-68231245 on Build GRCh38
rs895884083 Chr.10: 68230993-68230993 on Build GRCh38
rs552998481 Chr.10: 68231141-68231141 on Build GRCh38
rs960851028 Chr.10: 68231022-68231022 on Build GRCh38
rs777498271 Chr.10: 68231242-68231242 on Build GRCh38
rs1017112604 Chr.10: 68231068-68231068 on Build GRCh38
rs757263460 Chr.10: 68231212-68231212 on Build GRCh38
rs777134503 Chr.10: 68230957-68230957 on Build GRCh38
rs1044421755 Chr.10: 68231289-68231289 on Build GRCh38
rs768962996 Chr.10: 68231334-68231334 on Build GRCh38
rs1005769635 Chr.10: 68231033-68231033 on Build GRCh38
rs766397067 Chr.10: 68231380-68231380 on Build GRCh38
rs760974406 Chr.10: 68231271-68231271 on Build GRCh38
rs544409421 Chr.10: 68231398-68231398 on Build GRCh38
rs979099181 Chr.10: 68231117-68231117 on Build GRCh38
rs1056287623 Chr.10: 68231249-68231249 on Build GRCh38
rs773316578 Chr.10: 68231370-68231370 on Build GRCh38
rs985123898 Chr.10: 68231164-68231164 on Build GRCh38
rs778941278 Chr.10: 68231203-68231214 on Build GRCh38
rs972525353 Chr.10: 68231115-68231115 on Build GRCh38
rs748402895 Chr.10: 68231320-68231325 on Build GRCh38
rs778283214 Chr.10: 68231314-68231314 on Build GRCh38
rs764360015 Chr.10: 68231203-68231203 on Build GRCh38
rs747737101 Chr.10: 68231170-68231170 on Build GRCh38
rs755930408 Chr.10: 68231309-68231309 on Build GRCh38
rs761994815 Chr.10: 68231349-68231349 on Build GRCh38
rs775705457 Chr.10: 68231270-68231270 on Build GRCh38
rs748978805 Chr.10: 68231243-68231243 on Build GRCh38
rs944894782 Chr.10: 68231231-68231231 on Build GRCh38
rs757008955 Chr.10: 68231312-68231312 on Build GRCh38

View Full Product Details