Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68231662-68232168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH7
Entrez Gene ID
220202
Gene Name
atonal bHLH transcription factor 7
Gene Aliases
Math5, NCRNA, PHPVAR, RNANC, bHLHa13
UniGene
Hs.175396
Gene Chromosome Location
Chr.10: 68230595-68232113 on Build GRCh38
Transcripts
NM_145178.3

SNP Detail

SNP ID Chromosome Location
rs918324235 Chr.10: 68231727-68231727 on Build GRCh38
rs533488076 Chr.10: 68231847-68231847 on Build GRCh38
rs979335645 Chr.10: 68231784-68231784 on Build GRCh38
rs952424093 Chr.10: 68232097-68232098 on Build GRCh38
rs1000974469 Chr.10: 68232128-68232128 on Build GRCh38
rs534038078 Chr.10: 68232101-68232101 on Build GRCh38
rs1051703216 Chr.10: 68232033-68232033 on Build GRCh38
rs7916697 Chr.10: 68232096-68232096 on Build GRCh38
rs972481753 Chr.10: 68231725-68231725 on Build GRCh38
rs551184653 Chr.10: 68231717-68231717 on Build GRCh38
rs187364801 Chr.10: 68232067-68232067 on Build GRCh38
rs775885623 Chr.10: 68231730-68231730 on Build GRCh38
rs1019933180 Chr.10: 68232004-68232004 on Build GRCh38
rs967758791 Chr.10: 68232108-68232108 on Build GRCh38
rs754779263 Chr.10: 68231977-68231977 on Build GRCh38
rs984952198 Chr.10: 68231792-68231795 on Build GRCh38
rs549436173 Chr.10: 68231718-68231718 on Build GRCh38
rs991109927 Chr.10: 68232143-68232143 on Build GRCh38
rs1001110908 Chr.10: 68231922-68231922 on Build GRCh38
rs909413782 Chr.10: 68232126-68232126 on Build GRCh38
rs891260380 Chr.10: 68232062-68232062 on Build GRCh38
rs938241112 Chr.10: 68231827-68231827 on Build GRCh38
rs897431729 Chr.10: 68232018-68232018 on Build GRCh38
rs994424349 Chr.10: 68232028-68232028 on Build GRCh38
rs992441367 Chr.10: 68231875-68231875 on Build GRCh38
rs1030664126 Chr.10: 68232129-68232129 on Build GRCh38
rs1019313422 Chr.10: 68231703-68231703 on Build GRCh38
rs764050323 Chr.10: 68231860-68231860 on Build GRCh38
rs1053591796 Chr.10: 68231953-68231953 on Build GRCh38
rs560458990 Chr.10: 68232100-68232100 on Build GRCh38
rs917720197 Chr.10: 68231880-68231880 on Build GRCh38
rs923884847 Chr.10: 68231807-68231807 on Build GRCh38
rs546900907 Chr.10: 68231851-68231851 on Build GRCh38
rs965045597 Chr.10: 68232019-68232019 on Build GRCh38
rs958261099 Chr.10: 68231995-68231995 on Build GRCh38
rs61854782 Chr.10: 68231992-68231992 on Build GRCh38
rs569365534 Chr.10: 68231985-68231985 on Build GRCh38
rs555405231 Chr.10: 68231921-68231921 on Build GRCh38
rs772358498 Chr.10: 68232005-68232005 on Build GRCh38
rs6480319 Chr.10: 68231862-68231862 on Build GRCh38
rs985369737 Chr.10: 68232115-68232115 on Build GRCh38
rs763049857 Chr.10: 68231792-68231792 on Build GRCh38
rs371492363 Chr.10: 68231904-68231904 on Build GRCh38
rs950123823 Chr.10: 68231881-68231881 on Build GRCh38
rs967658701 Chr.10: 68231722-68231722 on Build GRCh38
rs929488075 Chr.10: 68231733-68231733 on Build GRCh38
rs6480320 Chr.10: 68231911-68231911 on Build GRCh38
rs896527591 Chr.10: 68231877-68231877 on Build GRCh38

View Full Product Details