Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44874671-44875105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001166588.2 NM_001261418.1 NM_020365.4 XM_017002745.1 XM_017002746.1 XM_017002747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
681192 c.1195G>C p.E399Q Chr.1: 44874685-44874685 on Build GRCh38
6063435 c.1190C>G p.T397S Chr.1: 44874690-44874690 on Build GRCh38
1180213 c.1058G>C p.G353A Chr.1: 44874822-44874822 on Build GRCh38
6479159 c.1147G>A p.D383N Chr.1: 44874733-44874733 on Build GRCh38
4448657 c.1157delC p.T386fs*8 Chr.1: 44874723-44874723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767371278 Chr.1: 44874764-44874764 on Build GRCh38
rs773530595 Chr.1: 44874739-44874739 on Build GRCh38
rs369175038 Chr.1: 44874784-44874784 on Build GRCh38
rs756556008 Chr.1: 44874861-44874861 on Build GRCh38
rs77257900 Chr.1: 44874976-44874976 on Build GRCh38
rs755472775 Chr.1: 44874720-44874720 on Build GRCh38
rs778133362 Chr.1: 44874870-44874870 on Build GRCh38
rs985975888 Chr.1: 44874702-44874702 on Build GRCh38
rs940924882 Chr.1: 44875045-44875045 on Build GRCh38
rs770256354 Chr.1: 44874737-44874737 on Build GRCh38
rs774443617 Chr.1: 44874749-44874749 on Build GRCh38
rs978851230 Chr.1: 44875051-44875051 on Build GRCh38
rs139247652 Chr.1: 44874766-44874766 on Build GRCh38
rs997248160 Chr.1: 44874814-44874814 on Build GRCh38
rs112491079 Chr.1: 44874862-44874862 on Build GRCh38
rs142651157 Chr.1: 44874744-44874744 on Build GRCh38
rs925118611 Chr.1: 44874722-44874722 on Build GRCh38
rs763994002 Chr.1: 44874772-44874772 on Build GRCh38
rs561461416 Chr.1: 44875011-44875011 on Build GRCh38
rs143688644 Chr.1: 44874729-44874729 on Build GRCh38
rs956569013 Chr.1: 44874926-44874926 on Build GRCh38
rs767253511 Chr.1: 44874900-44874900 on Build GRCh38
rs757337355 Chr.1: 44874863-44874863 on Build GRCh38
rs775556092 Chr.1: 44874765-44874765 on Build GRCh38
rs887783636 Chr.1: 44875037-44875037 on Build GRCh38
rs917729994 Chr.1: 44874946-44874946 on Build GRCh38
rs111737503 Chr.1: 44874991-44874991 on Build GRCh38
rs774696817 Chr.1: 44874750-44874750 on Build GRCh38
rs372697557 Chr.1: 44874836-44874836 on Build GRCh38
rs572682440 Chr.1: 44874910-44874910 on Build GRCh38
rs980654249 Chr.1: 44874980-44874980 on Build GRCh38
rs1032017233 Chr.1: 44874903-44874903 on Build GRCh38
rs778619398 Chr.1: 44874712-44874712 on Build GRCh38
rs953193336 Chr.1: 44874889-44874889 on Build GRCh38
rs763196787 Chr.1: 44874717-44874717 on Build GRCh38
rs937461335 Chr.1: 44875033-44875033 on Build GRCh38
rs749560754 Chr.1: 44874742-44874742 on Build GRCh38
rs749964150 Chr.1: 44874837-44874837 on Build GRCh38
rs765355307 Chr.1: 44874916-44874916 on Build GRCh38
rs748587746 Chr.1: 44874731-44874731 on Build GRCh38
rs749611682 Chr.1: 44874871-44874871 on Build GRCh38
rs779422482 Chr.1: 44874857-44874857 on Build GRCh38
rs201613944 Chr.1: 44874736-44874736 on Build GRCh38
rs565412618 Chr.1: 44874902-44874902 on Build GRCh38
rs988073465 Chr.1: 44874944-44874944 on Build GRCh38
rs888602115 Chr.1: 44874887-44874887 on Build GRCh38
rs781679802 Chr.1: 44874727-44874727 on Build GRCh38
rs1049019121 Chr.1: 44875036-44875036 on Build GRCh38
rs377388021 Chr.1: 44874856-44874856 on Build GRCh38
rs957887477 Chr.1: 44875072-44875072 on Build GRCh38
rs949052759 Chr.1: 44874789-44874789 on Build GRCh38
rs149557497 Chr.1: 44874822-44874822 on Build GRCh38
rs913591184 Chr.1: 44874773-44874773 on Build GRCh38
rs150426784 Chr.1: 44874782-44874782 on Build GRCh38
rs745323934 Chr.1: 44874714-44874714 on Build GRCh38
rs1004907358 Chr.1: 44874896-44874896 on Build GRCh38
rs263955 Chr.1: 44875031-44875031 on Build GRCh38
rs748726031 Chr.1: 44874860-44874860 on Build GRCh38
rs1037037521 Chr.1: 44875067-44875067 on Build GRCh38

View Full Product Details