Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3094130-3094686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982502 c.26G>T p.C9F Chr.19: 3094677-3094677 on Build GRCh38
6003458 c.10G>A p.E4K Chr.19: 3094661-3094661 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892721605 Chr.19: 3094218-3094218 on Build GRCh38
rs944202826 Chr.19: 3094234-3094234 on Build GRCh38
rs900243126 Chr.19: 3094246-3094246 on Build GRCh38
rs755279234 Chr.19: 3094629-3094629 on Build GRCh38
rs758302558 Chr.19: 3094657-3094657 on Build GRCh38
rs1036073139 Chr.19: 3094541-3094541 on Build GRCh38
rs1036020328 Chr.19: 3094374-3094374 on Build GRCh38
rs766876309 Chr.19: 3094611-3094611 on Build GRCh38
rs960935695 Chr.19: 3094289-3094289 on Build GRCh38
rs960791044 Chr.19: 3094408-3094408 on Build GRCh38
rs113917554 Chr.19: 3094339-3094339 on Build GRCh38
rs1013412088 Chr.19: 3094293-3094293 on Build GRCh38
rs555668234 Chr.19: 3094414-3094414 on Build GRCh38
rs996370360 Chr.19: 3094268-3094268 on Build GRCh38
rs113108560 Chr.19: 3094225-3094225 on Build GRCh38
rs190424317 Chr.19: 3094217-3094217 on Build GRCh38
rs1013881321 Chr.19: 3094390-3094390 on Build GRCh38
rs1014775220 Chr.19: 3094287-3094287 on Build GRCh38
rs868435029 Chr.19: 3094304-3094304 on Build GRCh38
rs745355696 Chr.19: 3094650-3094650 on Build GRCh38
rs887413112 Chr.19: 3094274-3094274 on Build GRCh38
rs1002739335 Chr.19: 3094239-3094239 on Build GRCh38
rs146862811 Chr.19: 3094373-3094373 on Build GRCh38
rs969427467 Chr.19: 3094229-3094229 on Build GRCh38
rs756826638 Chr.19: 3094606-3094606 on Build GRCh38
rs988165025 Chr.19: 3094310-3094310 on Build GRCh38
rs921395437 Chr.19: 3094357-3094357 on Build GRCh38
rs1019706994 Chr.19: 3094651-3094651 on Build GRCh38
rs976039899 Chr.19: 3094468-3094468 on Build GRCh38
rs1048802447 Chr.19: 3094269-3094269 on Build GRCh38
rs1019667781 Chr.19: 3094430-3094430 on Build GRCh38
rs905133680 Chr.19: 3094608-3094608 on Build GRCh38
rs965412250 Chr.19: 3094452-3094452 on Build GRCh38
rs763741959 Chr.19: 3094632-3094632 on Build GRCh38
rs866779785 Chr.19: 3094529-3094529 on Build GRCh38
rs887549381 Chr.19: 3094370-3094370 on Build GRCh38
rs956669118 Chr.19: 3094306-3094306 on Build GRCh38
rs1004521916 Chr.19: 3094275-3094275 on Build GRCh38
rs532185482 Chr.19: 3094226-3094226 on Build GRCh38
rs992423531 Chr.19: 3094172-3094172 on Build GRCh38
rs1000689229 Chr.19: 3094613-3094613 on Build GRCh38
rs969251973 Chr.19: 3094303-3094303 on Build GRCh38
rs971927269 Chr.19: 3094434-3094434 on Build GRCh38
rs1022283160 Chr.19: 3094224-3094224 on Build GRCh38
rs879191762 Chr.19: 3094600-3094600 on Build GRCh38
rs925372137 Chr.19: 3094306-3094306 on Build GRCh38
rs1048773601 Chr.19: 3094360-3094360 on Build GRCh38
rs756924546 Chr.19: 3094639-3094639 on Build GRCh38
rs1040307342 Chr.19: 3094239-3094239 on Build GRCh38
rs1000719946 Chr.19: 3094405-3094405 on Build GRCh38
rs975607392 Chr.19: 3094330-3094330 on Build GRCh38
rs779989113 Chr.19: 3094660-3094660 on Build GRCh38
rs763417133 Chr.19: 3094606-3094606 on Build GRCh38
rs912807134 Chr.19: 3094321-3094321 on Build GRCh38
rs916655799 Chr.19: 3094177-3094177 on Build GRCh38
rs984281744 Chr.19: 3094481-3094481 on Build GRCh38
rs567604735 Chr.19: 3094645-3094645 on Build GRCh38
rs751944446 Chr.19: 3094628-3094628 on Build GRCh38
rs944324580 Chr.19: 3094327-3094327 on Build GRCh38
rs979454369 Chr.19: 3094179-3094179 on Build GRCh38
rs888627445 Chr.19: 3094559-3094574 on Build GRCh38
rs528327233 Chr.19: 3094166-3094166 on Build GRCh38
rs896400791 Chr.19: 3094389-3094389 on Build GRCh38
rs546764693 Chr.19: 3094175-3094175 on Build GRCh38
rs1014806164 Chr.19: 3094156-3094156 on Build GRCh38
rs568480334 Chr.19: 3094294-3094294 on Build GRCh38
rs571847017 Chr.19: 3094600-3094639 on Build GRCh38
rs908957464 Chr.19: 3094503-3094503 on Build GRCh38
rs753555122 Chr.19: 3094633-3094633 on Build GRCh38
rs1045363151 Chr.19: 3094601-3094601 on Build GRCh38
rs1010837814 Chr.19: 3094222-3094222 on Build GRCh38
rs550314159 Chr.19: 3094230-3094230 on Build GRCh38
rs1023561198 Chr.19: 3094394-3094394 on Build GRCh38
rs904996140 Chr.19: 3094397-3094397 on Build GRCh38
rs949094363 Chr.19: 3094574-3094574 on Build GRCh38
rs921546042 Chr.19: 3094469-3094469 on Build GRCh38
rs931453339 Chr.19: 3094478-3094478 on Build GRCh38
rs865916061 Chr.19: 3094358-3094358 on Build GRCh38
rs541113525 Chr.19: 3094639-3094640 on Build GRCh38

View Full Product Details