Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39269090-39269644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNL2
Entrez Gene ID
282616
Gene Name
interferon lambda 2
Gene Aliases
IL-28A, IL28A
UniGene
Hs.567792
Gene Chromosome Location
Chr.19: 39268517-39270092 on Build GRCh38
Transcripts
NM_172138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963028 c.328G>C p.E110Q Chr.19: 39269545-39269545 on Build GRCh38
3533649 c.318G>A p.L106L Chr.19: 39269535-39269535 on Build GRCh38
5749923 c.334A>G p.T112A Chr.19: 39269551-39269551 on Build GRCh38
3223478 c.233G>A p.R78H Chr.19: 39269192-39269192 on Build GRCh38
5707016 c.337G>A p.A113T Chr.19: 39269554-39269554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545058782 Chr.19: 39269580-39269580 on Build GRCh38
rs544218398 Chr.19: 39269271-39269271 on Build GRCh38
rs759411820 Chr.19: 39269567-39269567 on Build GRCh38
rs139608275 Chr.19: 39269414-39269414 on Build GRCh38
rs769950521 Chr.19: 39269180-39269180 on Build GRCh38
rs370263782 Chr.19: 39269452-39269452 on Build GRCh38
rs11670676 Chr.19: 39269503-39269503 on Build GRCh38
rs1013723361 Chr.19: 39269294-39269294 on Build GRCh38
rs755853031 Chr.19: 39269214-39269214 on Build GRCh38
rs368771426 Chr.19: 39269457-39269457 on Build GRCh38
rs573494672 Chr.19: 39269247-39269247 on Build GRCh38
rs766091742 Chr.19: 39269220-39269220 on Build GRCh38
rs770847376 Chr.19: 39269563-39269563 on Build GRCh38
rs556038833 Chr.19: 39269459-39269459 on Build GRCh38
rs772249678 Chr.19: 39269487-39269487 on Build GRCh38
rs750166522 Chr.19: 39269113-39269113 on Build GRCh38
rs749474158 Chr.19: 39269483-39269483 on Build GRCh38
rs747541240 Chr.19: 39269242-39269242 on Build GRCh38
rs747767531 Chr.19: 39269201-39269201 on Build GRCh38
rs567230607 Chr.19: 39269345-39269345 on Build GRCh38
rs777999084 Chr.19: 39269553-39269553 on Build GRCh38
rs780295844 Chr.19: 39269532-39269532 on Build GRCh38
rs890913344 Chr.19: 39269560-39269560 on Build GRCh38
rs750994212 Chr.19: 39269525-39269525 on Build GRCh38
rs963994603 Chr.19: 39269392-39269392 on Build GRCh38
rs762495988 Chr.19: 39269517-39269517 on Build GRCh38
rs766039199 Chr.19: 39269235-39269235 on Build GRCh38
rs201517019 Chr.19: 39269191-39269191 on Build GRCh38
rs372384345 Chr.19: 39269516-39269516 on Build GRCh38
rs766256279 Chr.19: 39269448-39269448 on Build GRCh38
rs778475720 Chr.19: 39269236-39269236 on Build GRCh38
rs771261302 Chr.19: 39269161-39269161 on Build GRCh38
rs577580465 Chr.19: 39269576-39269576 on Build GRCh38
rs776317212 Chr.19: 39269565-39269565 on Build GRCh38
rs779724798 Chr.19: 39269132-39269132 on Build GRCh38
rs370646585 Chr.19: 39269259-39269259 on Build GRCh38
rs1052600833 Chr.19: 39269274-39269274 on Build GRCh38
rs759762461 Chr.19: 39269169-39269169 on Build GRCh38
rs757720374 Chr.19: 39269243-39269243 on Build GRCh38
rs755760324 Chr.19: 39269118-39269118 on Build GRCh38
rs1017052621 Chr.19: 39269328-39269328 on Build GRCh38
rs918001846 Chr.19: 39269442-39269442 on Build GRCh38
rs527504578 Chr.19: 39269320-39269320 on Build GRCh38
rs573640036 Chr.19: 39269270-39269270 on Build GRCh38
rs749510524 Chr.19: 39269267-39269267 on Build GRCh38
rs761668694 Chr.19: 39269272-39269272 on Build GRCh38
rs755421550 Chr.19: 39269594-39269594 on Build GRCh38
rs746951049 Chr.19: 39269554-39269554 on Build GRCh38
rs114998203 Chr.19: 39269301-39269301 on Build GRCh38
rs550946607 Chr.19: 39269357-39269357 on Build GRCh38
rs754270577 Chr.19: 39269593-39269593 on Build GRCh38
rs569194521 Chr.19: 39269358-39269358 on Build GRCh38
rs750186678 Chr.19: 39269212-39269212 on Build GRCh38
rs186980558 Chr.19: 39269160-39269160 on Build GRCh38
rs778185023 Chr.19: 39269159-39269159 on Build GRCh38
rs117469515 Chr.19: 39269314-39269314 on Build GRCh38
rs117789288 Chr.19: 39269334-39269334 on Build GRCh38
rs746349383 Chr.19: 39269462-39269462 on Build GRCh38
rs551222601 Chr.19: 39269307-39269307 on Build GRCh38
rs905451650 Chr.19: 39269469-39269469 on Build GRCh38
rs759127860 Chr.19: 39269502-39269502 on Build GRCh38
rs28405467 Chr.19: 39269455-39269455 on Build GRCh38
rs754499511 Chr.19: 39269226-39269226 on Build GRCh38
rs781555688 Chr.19: 39269250-39269250 on Build GRCh38
rs767066131 Chr.19: 39269153-39269153 on Build GRCh38
rs760697546 Chr.19: 39269207-39269207 on Build GRCh38
rs267605471 Chr.19: 39269587-39269587 on Build GRCh38
rs748361375 Chr.19: 39269540-39269540 on Build GRCh38
rs756525825 Chr.19: 39269463-39269463 on Build GRCh38
rs760589215 Chr.19: 39269438-39269438 on Build GRCh38
rs539578886 Chr.19: 39269199-39269199 on Build GRCh38
rs764662143 Chr.19: 39269458-39269458 on Build GRCh38
rs375319879 Chr.19: 39269497-39269497 on Build GRCh38
rs772954609 Chr.19: 39269172-39269173 on Build GRCh38
rs568083772 Chr.19: 39269163-39269163 on Build GRCh38
rs375917586 Chr.19: 39269192-39269192 on Build GRCh38
rs955169112 Chr.19: 39269362-39269362 on Build GRCh38
rs374174818 Chr.19: 39269454-39269454 on Build GRCh38
rs8103362 Chr.19: 39269551-39269551 on Build GRCh38
rs773172753 Chr.19: 39269437-39269437 on Build GRCh38
rs575681055 Chr.19: 39269198-39269198 on Build GRCh38
rs776980078 Chr.19: 39269164-39269164 on Build GRCh38
rs775735271 Chr.19: 39269263-39269263 on Build GRCh38
rs772667749 Chr.19: 39269143-39269143 on Build GRCh38
rs540902358 Chr.19: 39269257-39269257 on Build GRCh38
rs751010821 Chr.19: 39269583-39269583 on Build GRCh38
rs537786079 Chr.19: 39269346-39269346 on Build GRCh38
rs761191388 Chr.19: 39269585-39269585 on Build GRCh38
rs935358335 Chr.19: 39269434-39269434 on Build GRCh38
rs763578475 Chr.19: 39269520-39269520 on Build GRCh38
rs115134580 Chr.19: 39269186-39269186 on Build GRCh38
rs754182150 Chr.19: 39269538-39269538 on Build GRCh38
rs774318204 Chr.19: 39269485-39269485 on Build GRCh38
rs748219032 Chr.19: 39269486-39269486 on Build GRCh38
rs377643651 Chr.19: 39269177-39269177 on Build GRCh38
rs763597001 Chr.19: 39269582-39269582 on Build GRCh38
rs753499684 Chr.19: 39269223-39269223 on Build GRCh38
rs574384388 Chr.19: 39269498-39269498 on Build GRCh38
rs558318815 Chr.19: 39269204-39269204 on Build GRCh38
rs780460286 Chr.19: 39269477-39269477 on Build GRCh38
rs781690954 Chr.19: 39269460-39269460 on Build GRCh38
rs986639987 Chr.19: 39269367-39269367 on Build GRCh38
rs199706890 Chr.19: 39269623-39269623 on Build GRCh38
rs200211932 Chr.19: 39269531-39269531 on Build GRCh38
rs373525938 Chr.19: 39269581-39269581 on Build GRCh38
rs962474091 Chr.19: 39269331-39269331 on Build GRCh38
rs768863662 Chr.19: 39269484-39269484 on Build GRCh38
rs200081180 Chr.19: 39269500-39269500 on Build GRCh38
rs748758261 Chr.19: 39269133-39269133 on Build GRCh38
rs1016651857 Chr.19: 39269187-39269187 on Build GRCh38
rs779134113 Chr.19: 39269618-39269618 on Build GRCh38
rs773332707 Chr.19: 39269494-39269494 on Build GRCh38
rs150593472 Chr.19: 39269359-39269359 on Build GRCh38
rs911133881 Chr.19: 39269368-39269368 on Build GRCh38
rs200736899 Chr.19: 39269626-39269626 on Build GRCh38
rs377564412 Chr.19: 39269589-39269589 on Build GRCh38
rs30481 Chr.19: 39269429-39269429 on Build GRCh38
rs758630545 Chr.19: 39269624-39269624 on Build GRCh38
rs62120533 Chr.19: 39269278-39269278 on Build GRCh38

View Full Product Details