Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50770585-50771151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR32
Entrez Gene ID
2854
Gene Name
G protein-coupled receptor 32
Gene Aliases
RVDR1
UniGene
Hs.515555
Gene Chromosome Location
Chr.19: 50770464-50771732 on Build GRCh38
Transcripts
NM_001506.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7370042 c.293C>T p.A98V Chr.19: 50770893-50770893 on Build GRCh38
4791139 c.469C>T p.R157W Chr.19: 50771069-50771069 on Build GRCh38
7370040 c.216G>A p.M72I Chr.19: 50770816-50770816 on Build GRCh38
2755188 c.295A>G p.M99V Chr.19: 50770895-50770895 on Build GRCh38
1129745 c.61C>T p.R21C Chr.19: 50770661-50770661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756364822 Chr.19: 50770905-50770905 on Build GRCh38
rs767524621 Chr.19: 50771008-50771008 on Build GRCh38
rs777061182 Chr.19: 50770768-50770768 on Build GRCh38
rs751124301 Chr.19: 50771103-50771103 on Build GRCh38
rs769794039 Chr.19: 50770663-50770663 on Build GRCh38
rs769916488 Chr.19: 50770728-50770728 on Build GRCh38
rs562117047 Chr.19: 50770868-50770868 on Build GRCh38
rs768827782 Chr.19: 50771125-50771125 on Build GRCh38
rs772773682 Chr.19: 50771118-50771118 on Build GRCh38
rs985714787 Chr.19: 50770653-50770653 on Build GRCh38
rs60302252 Chr.19: 50771062-50771062 on Build GRCh38
rs777896923 Chr.19: 50770813-50770813 on Build GRCh38
rs767828175 Chr.19: 50770880-50770880 on Build GRCh38
rs368895414 Chr.19: 50770765-50770765 on Build GRCh38
rs754689136 Chr.19: 50771058-50771058 on Build GRCh38
rs765794369 Chr.19: 50770786-50770786 on Build GRCh38
rs147807422 Chr.19: 50770799-50770799 on Build GRCh38
rs763460659 Chr.19: 50770862-50770862 on Build GRCh38
rs757665090 Chr.19: 50770633-50770633 on Build GRCh38
rs375835721 Chr.19: 50770861-50770861 on Build GRCh38
rs773267384 Chr.19: 50770748-50770748 on Build GRCh38
rs749646537 Chr.19: 50770986-50770986 on Build GRCh38
rs772927207 Chr.19: 50770613-50770613 on Build GRCh38
rs149687485 Chr.19: 50770794-50770794 on Build GRCh38
rs541459602 Chr.19: 50770892-50770892 on Build GRCh38
rs759518640 Chr.19: 50770878-50770878 on Build GRCh38
rs558516388 Chr.19: 50770756-50770756 on Build GRCh38
rs746445364 Chr.19: 50771057-50771057 on Build GRCh38
rs1007134018 Chr.19: 50770812-50770812 on Build GRCh38
rs771826086 Chr.19: 50770834-50770834 on Build GRCh38
rs772571947 Chr.19: 50771070-50771070 on Build GRCh38
rs918700371 Chr.19: 50770677-50770677 on Build GRCh38
rs753630831 Chr.19: 50770918-50770918 on Build GRCh38
rs759700202 Chr.19: 50771000-50771000 on Build GRCh38
rs978429911 Chr.19: 50770686-50770686 on Build GRCh38
rs774735970 Chr.19: 50771066-50771066 on Build GRCh38
rs148996594 Chr.19: 50770990-50770990 on Build GRCh38
rs972727487 Chr.19: 50770883-50770883 on Build GRCh38
rs745443642 Chr.19: 50770928-50770928 on Build GRCh38
rs199544421 Chr.19: 50771089-50771089 on Build GRCh38
rs143841096 Chr.19: 50771038-50771038 on Build GRCh38
rs924290483 Chr.19: 50770690-50770690 on Build GRCh38
rs778714355 Chr.19: 50770927-50770927 on Build GRCh38
rs141246887 Chr.19: 50770825-50770825 on Build GRCh38
rs878936480 Chr.19: 50770926-50770926 on Build GRCh38
rs760992285 Chr.19: 50771015-50771015 on Build GRCh38
rs766190891 Chr.19: 50770674-50770674 on Build GRCh38
rs374490694 Chr.19: 50770647-50770647 on Build GRCh38
rs756485883 Chr.19: 50770615-50770615 on Build GRCh38
rs528025406 Chr.19: 50770895-50770895 on Build GRCh38
rs781050618 Chr.19: 50770726-50770726 on Build GRCh38
rs35300823 Chr.19: 50770705-50770705 on Build GRCh38
rs866174710 Chr.19: 50770649-50770649 on Build GRCh38
rs370107165 Chr.19: 50770924-50770924 on Build GRCh38
rs536521302 Chr.19: 50770631-50770631 on Build GRCh38
rs368089874 Chr.19: 50770689-50770689 on Build GRCh38
rs746698749 Chr.19: 50770939-50770939 on Build GRCh38
rs750187431 Chr.19: 50771096-50771096 on Build GRCh38
rs764470671 Chr.19: 50771080-50771080 on Build GRCh38
rs749267775 Chr.19: 50770819-50770819 on Build GRCh38
rs772552393 Chr.19: 50770891-50770892 on Build GRCh38
rs746382678 Chr.19: 50771069-50771069 on Build GRCh38
rs770118200 Chr.19: 50770852-50770852 on Build GRCh38
rs980378202 Chr.19: 50771087-50771087 on Build GRCh38
rs755948988 Chr.19: 50770811-50770811 on Build GRCh38
rs778498523 Chr.19: 50770824-50770824 on Build GRCh38
rs375757810 Chr.19: 50770695-50770695 on Build GRCh38
rs757223479 Chr.19: 50770921-50770921 on Build GRCh38
rs142691017 Chr.19: 50770670-50770670 on Build GRCh38
rs770943742 Chr.19: 50770820-50770820 on Build GRCh38
rs766499816 Chr.19: 50770790-50770790 on Build GRCh38
rs190167592 Chr.19: 50771124-50771124 on Build GRCh38
rs751725969 Chr.19: 50770871-50770871 on Build GRCh38
rs776245921 Chr.19: 50770658-50770658 on Build GRCh38
rs538507863 Chr.19: 50770696-50770696 on Build GRCh38
rs192426883 Chr.19: 50770629-50770629 on Build GRCh38
rs138439551 Chr.19: 50770635-50770635 on Build GRCh38
rs1029805697 Chr.19: 50770636-50770636 on Build GRCh38
rs775716480 Chr.19: 50771073-50771073 on Build GRCh38
rs893372939 Chr.19: 50770755-50770755 on Build GRCh38
rs752597023 Chr.19: 50770805-50770805 on Build GRCh38
rs779724333 Chr.19: 50770938-50770938 on Build GRCh38
rs780695906 Chr.19: 50771120-50771120 on Build GRCh38
rs773566984 Chr.19: 50770975-50770975 on Build GRCh38
rs150920233 Chr.19: 50770702-50770702 on Build GRCh38
rs747001231 Chr.19: 50770893-50770894 on Build GRCh38
rs774697335 Chr.19: 50770996-50770996 on Build GRCh38
rs574073602 Chr.19: 50770891-50770891 on Build GRCh38
rs145045571 Chr.19: 50771107-50771107 on Build GRCh38
rs768479231 Chr.19: 50770655-50770655 on Build GRCh38
rs764346155 Chr.19: 50771023-50771023 on Build GRCh38
rs747829098 Chr.19: 50770727-50770727 on Build GRCh38
rs146710967 Chr.19: 50770911-50770911 on Build GRCh38
rs754459913 Chr.19: 50771108-50771108 on Build GRCh38
rs1054752513 Chr.19: 50770754-50770754 on Build GRCh38
rs946258942 Chr.19: 50770968-50770968 on Build GRCh38
rs1006753971 Chr.19: 50771052-50771052 on Build GRCh38
rs771566336 Chr.19: 50770767-50770767 on Build GRCh38
rs138848386 Chr.19: 50771110-50771110 on Build GRCh38
rs762162666 Chr.19: 50770779-50770779 on Build GRCh38
rs1005579709 Chr.19: 50770621-50770621 on Build GRCh38
rs753754669 Chr.19: 50771033-50771033 on Build GRCh38
rs750911146 Chr.19: 50770787-50770787 on Build GRCh38
rs780861174 Chr.19: 50771109-50771109 on Build GRCh38
rs374550267 Chr.19: 50771098-50771098 on Build GRCh38
rs771593122 Chr.19: 50770777-50770777 on Build GRCh38
rs776735882 Chr.19: 50771128-50771128 on Build GRCh38
rs868565431 Chr.19: 50770803-50770803 on Build GRCh38
rs535831809 Chr.19: 50771130-50771130 on Build GRCh38
rs775663056 Chr.19: 50771011-50771011 on Build GRCh38
rs758625863 Chr.19: 50770638-50770638 on Build GRCh38
rs367982246 Chr.19: 50771065-50771065 on Build GRCh38
rs779158621 Chr.19: 50771111-50771111 on Build GRCh38
rs780806070 Chr.19: 50771060-50771060 on Build GRCh38
rs761967127 Chr.19: 50771129-50771129 on Build GRCh38
rs917062643 Chr.19: 50770708-50770708 on Build GRCh38
rs560406571 Chr.19: 50770893-50770893 on Build GRCh38
rs762217628 Chr.19: 50770835-50770835 on Build GRCh38
rs748135176 Chr.19: 50770659-50770659 on Build GRCh38
rs760834395 Chr.19: 50771076-50771076 on Build GRCh38
rs770791046 Chr.19: 50771008-50771009 on Build GRCh38
rs768236752 Chr.19: 50770954-50770954 on Build GRCh38
rs267605599 Chr.19: 50770692-50770692 on Build GRCh38
rs1015684542 Chr.19: 50770625-50770625 on Build GRCh38
rs866404485 Chr.19: 50770953-50770953 on Build GRCh38
rs371882426 Chr.19: 50770789-50770789 on Build GRCh38
rs746177227 Chr.19: 50771117-50771117 on Build GRCh38
rs779566310 Chr.19: 50771041-50771041 on Build GRCh38
rs1042079886 Chr.19: 50770972-50770972 on Build GRCh38
rs374462813 Chr.19: 50770719-50770719 on Build GRCh38
rs771464020 Chr.19: 50770930-50770930 on Build GRCh38
rs139378141 Chr.19: 50770643-50770643 on Build GRCh38
rs1022493915 Chr.19: 50770950-50770950 on Build GRCh38
rs141853995 Chr.19: 50771093-50771093 on Build GRCh38
rs1003265650 Chr.19: 50771100-50771100 on Build GRCh38
rs185022065 Chr.19: 50771083-50771083 on Build GRCh38
rs992578120 Chr.19: 50771064-50771064 on Build GRCh38
rs774033486 Chr.19: 50770684-50770684 on Build GRCh38
rs751531782 Chr.19: 50770793-50770793 on Build GRCh38
rs371454037 Chr.19: 50770715-50770715 on Build GRCh38
rs201317983 Chr.19: 50770668-50770668 on Build GRCh38
rs750300251 Chr.19: 50771039-50771039 on Build GRCh38
rs747226325 Chr.19: 50770718-50770718 on Build GRCh38

View Full Product Details