Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53914308-53914721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
UniGene
Hs.631597
Gene Chromosome Location
Chr.19: 53909425-53943944 on Build GRCh38
Transcripts
NM_031896.4 XM_005259124.3 XM_017027093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618734 c.273G>T p.E91D Chr.19: 53914576-53914576 on Build GRCh38
6085841 c.271G>T p.E91* Chr.19: 53914574-53914574 on Build GRCh38
7127260 c.202G>C p.E68Q Chr.19: 53914505-53914505 on Build GRCh38
4539682 c.271G>A p.E91K Chr.19: 53914574-53914574 on Build GRCh38
4607307 c.283+2T>A p.? Chr.19: 53914588-53914588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757598896 Chr.19: 53914504-53914504 on Build GRCh38
rs758447649 Chr.19: 53914582-53914582 on Build GRCh38
rs200682285 Chr.19: 53914604-53914604 on Build GRCh38
rs766067499 Chr.19: 53914483-53914483 on Build GRCh38
rs755850198 Chr.19: 53914591-53914591 on Build GRCh38
rs1001976641 Chr.19: 53914373-53914373 on Build GRCh38
rs1014173489 Chr.19: 53914364-53914364 on Build GRCh38
rs781304642 Chr.19: 53914511-53914511 on Build GRCh38
rs906963615 Chr.19: 53914391-53914391 on Build GRCh38
rs767073488 Chr.19: 53914474-53914474 on Build GRCh38
rs149797266 Chr.19: 53914564-53914564 on Build GRCh38
rs879010434 Chr.19: 53914600-53914600 on Build GRCh38
rs752428498 Chr.19: 53914622-53914622 on Build GRCh38
rs376431747 Chr.19: 53914625-53914625 on Build GRCh38
rs752693463 Chr.19: 53914499-53914499 on Build GRCh38
rs376990952 Chr.19: 53914501-53914501 on Build GRCh38
rs778149960 Chr.19: 53914463-53914464 on Build GRCh38
rs1011207913 Chr.19: 53914437-53914437 on Build GRCh38
rs767743454 Chr.19: 53914563-53914563 on Build GRCh38
rs1025832959 Chr.19: 53914419-53914419 on Build GRCh38
rs1046534283 Chr.19: 53914372-53914372 on Build GRCh38
rs747312506 Chr.19: 53914491-53914492 on Build GRCh38
rs907221015 Chr.19: 53914353-53914353 on Build GRCh38
rs770097054 Chr.19: 53914639-53914639 on Build GRCh38
rs754759538 Chr.19: 53914496-53914496 on Build GRCh38
rs770070018 Chr.19: 53914521-53914521 on Build GRCh38
rs368647936 Chr.19: 53914453-53914453 on Build GRCh38
rs778063503 Chr.19: 53914502-53914502 on Build GRCh38
rs996012817 Chr.19: 53914667-53914667 on Build GRCh38
rs756498242 Chr.19: 53914635-53914635 on Build GRCh38
rs770299413 Chr.19: 53914467-53914467 on Build GRCh38
rs1025777287 Chr.19: 53914691-53914691 on Build GRCh38
rs1034437705 Chr.19: 53914377-53914377 on Build GRCh38
rs746181159 Chr.19: 53914520-53914520 on Build GRCh38
rs1056229394 Chr.19: 53914657-53914657 on Build GRCh38
rs764997446 Chr.19: 53914572-53914572 on Build GRCh38
rs953194748 Chr.19: 53914549-53914549 on Build GRCh38
rs372523380 Chr.19: 53914621-53914621 on Build GRCh38
rs372749580 Chr.19: 53914497-53914497 on Build GRCh38
rs1010128292 Chr.19: 53914331-53914331 on Build GRCh38
rs746400003 Chr.19: 53914401-53914401 on Build GRCh38
rs112904688 Chr.19: 53914341-53914341 on Build GRCh38
rs764243736 Chr.19: 53914585-53914585 on Build GRCh38
rs772849989 Chr.19: 53914478-53914478 on Build GRCh38
rs974853883 Chr.19: 53914515-53914515 on Build GRCh38
rs957060536 Chr.19: 53914427-53914427 on Build GRCh38
rs772758283 Chr.19: 53914548-53914548 on Build GRCh38
rs898042736 Chr.19: 53914350-53914350 on Build GRCh38
rs139711961 Chr.19: 53914546-53914546 on Build GRCh38
rs554742023 Chr.19: 53914397-53914397 on Build GRCh38
rs371625954 Chr.19: 53914573-53914573 on Build GRCh38
rs369839062 Chr.19: 53914545-53914545 on Build GRCh38
rs898552959 Chr.19: 53914662-53914662 on Build GRCh38
rs111577643 Chr.19: 53914611-53914611 on Build GRCh38
rs760361293 Chr.19: 53914481-53914481 on Build GRCh38
rs987052635 Chr.19: 53914462-53914462 on Build GRCh38
rs760201737 Chr.19: 53914559-53914559 on Build GRCh38
rs753619342 Chr.19: 53914492-53914492 on Build GRCh38
rs551136659 Chr.19: 53914634-53914634 on Build GRCh38
rs75256458 Chr.19: 53914599-53914599 on Build GRCh38
rs7248153 Chr.19: 53914617-53914617 on Build GRCh38
rs1020625998 Chr.19: 53914441-53914441 on Build GRCh38
rs183619484 Chr.19: 53914640-53914640 on Build GRCh38
rs919336163 Chr.19: 53914436-53914436 on Build GRCh38
rs199845626 Chr.19: 53914529-53914529 on Build GRCh38
rs1054266739 Chr.19: 53914565-53914565 on Build GRCh38
rs1001310133 Chr.19: 53914399-53914399 on Build GRCh38
rs573300409 Chr.19: 53914468-53914468 on Build GRCh38
rs544536217 Chr.19: 53914692-53914692 on Build GRCh38
rs747198707 Chr.19: 53914503-53914503 on Build GRCh38

View Full Product Details