Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34669820-34670330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2 XM_017028406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4598079 c.520G>A p.G174S Chr.21: 34669908-34669908 on Build GRCh38
6240442 c.761A>G p.D254G Chr.21: 34670149-34670149 on Build GRCh38
4618100 c.666G>A p.A222A Chr.21: 34670054-34670054 on Build GRCh38
4437467 c.631C>T p.Q211* Chr.21: 34670019-34670019 on Build GRCh38
224602 c.737T>G p.V246G Chr.21: 34670125-34670125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915009137 Chr.21: 34670204-34670204 on Build GRCh38
rs563057307 Chr.21: 34670035-34670035 on Build GRCh38
rs9680380 Chr.21: 34669904-34669904 on Build GRCh38
rs755257750 Chr.21: 34669949-34669949 on Build GRCh38
rs780166380 Chr.21: 34669883-34669883 on Build GRCh38
rs13049002 Chr.21: 34670210-34670210 on Build GRCh38
rs531747205 Chr.21: 34670055-34670055 on Build GRCh38
rs987952518 Chr.21: 34670304-34670304 on Build GRCh38
rs1007864420 Chr.21: 34670200-34670200 on Build GRCh38
rs183914556 Chr.21: 34670016-34670016 on Build GRCh38
rs766236550 Chr.21: 34669910-34669910 on Build GRCh38
rs199527885 Chr.21: 34670019-34670019 on Build GRCh38
rs565184857 Chr.21: 34670154-34670154 on Build GRCh38
rs768831686 Chr.21: 34669889-34669889 on Build GRCh38
rs975129293 Chr.21: 34670219-34670219 on Build GRCh38
rs956541929 Chr.21: 34669860-34669860 on Build GRCh38
rs530124031 Chr.21: 34670253-34670253 on Build GRCh38
rs950036345 Chr.21: 34669869-34669869 on Build GRCh38
rs200426571 Chr.21: 34669890-34669890 on Build GRCh38
rs537252722 Chr.21: 34669908-34669908 on Build GRCh38
rs543609987 Chr.21: 34670147-34670147 on Build GRCh38
rs747293545 Chr.21: 34669884-34669884 on Build GRCh38
rs765751599 Chr.21: 34669920-34669920 on Build GRCh38
rs566105353 Chr.21: 34669841-34669841 on Build GRCh38
rs772993069 Chr.21: 34669980-34669980 on Build GRCh38
rs552680215 Chr.21: 34669989-34669989 on Build GRCh38
rs755465153 Chr.21: 34670180-34670209 on Build GRCh38
rs760206035 Chr.21: 34670119-34670119 on Build GRCh38
rs774931572 Chr.21: 34670108-34670108 on Build GRCh38
rs747194136 Chr.21: 34669928-34669928 on Build GRCh38
rs763696583 Chr.21: 34670123-34670123 on Build GRCh38
rs13048894 Chr.21: 34670207-34670207 on Build GRCh38
rs899656310 Chr.21: 34670191-34670191 on Build GRCh38
rs13048804 Chr.21: 34670193-34670193 on Build GRCh38
rs548662808 Chr.21: 34670262-34670262 on Build GRCh38
rs1055882965 Chr.21: 34669849-34669849 on Build GRCh38
rs1053489280 Chr.21: 34670125-34670125 on Build GRCh38
rs541359989 Chr.21: 34669960-34669960 on Build GRCh38
rs756240337 Chr.21: 34670196-34670196 on Build GRCh38
rs770619828 Chr.21: 34670047-34670047 on Build GRCh38
rs887316535 Chr.21: 34670237-34670237 on Build GRCh38
rs920915557 Chr.21: 34670220-34670220 on Build GRCh38
rs758998698 Chr.21: 34670155-34670155 on Build GRCh38
rs752208168 Chr.21: 34669919-34669919 on Build GRCh38
rs946433979 Chr.21: 34670218-34670218 on Build GRCh38
rs998770359 Chr.21: 34670038-34670038 on Build GRCh38
rs557556798 Chr.21: 34670164-34670193 on Build GRCh38
rs62213789 Chr.21: 34670163-34670163 on Build GRCh38
rs760047996 Chr.21: 34670134-34670163 on Build GRCh38
rs548102329 Chr.21: 34670192-34670192 on Build GRCh38
rs751151688 Chr.21: 34669912-34669912 on Build GRCh38
rs1038922856 Chr.21: 34670255-34670255 on Build GRCh38
rs1023000207 Chr.21: 34670266-34670266 on Build GRCh38
rs62213790 Chr.21: 34670171-34670171 on Build GRCh38
rs765880721 Chr.21: 34670142-34670171 on Build GRCh38
rs769869692 Chr.21: 34669891-34669891 on Build GRCh38
rs13049239 Chr.21: 34670285-34670285 on Build GRCh38
rs574405121 Chr.21: 34669990-34669990 on Build GRCh38
rs554541361 Chr.21: 34670051-34670051 on Build GRCh38
rs749178925 Chr.21: 34670226-34670226 on Build GRCh38
rs879373292 Chr.21: 34670232-34670232 on Build GRCh38
rs192051474 Chr.21: 34669859-34669859 on Build GRCh38
rs956592646 Chr.21: 34670303-34670303 on Build GRCh38
rs754329010 Chr.21: 34669839-34669839 on Build GRCh38
rs898605447 Chr.21: 34670259-34670259 on Build GRCh38
rs4647768 Chr.21: 34669853-34669882 on Build GRCh38
rs558842821 Chr.21: 34669924-34669924 on Build GRCh38
rs766589415 Chr.21: 34670178-34670178 on Build GRCh38
rs760367294 Chr.21: 34670288-34670288 on Build GRCh38
rs750756040 Chr.21: 34669921-34669921 on Build GRCh38
rs763496806 Chr.21: 34670300-34670300 on Build GRCh38
rs772942828 Chr.21: 34670066-34670095 on Build GRCh38
rs947429515 Chr.21: 34670161-34670161 on Build GRCh38
rs75303394 Chr.21: 34670204-34670206 on Build GRCh38
rs767169617 Chr.21: 34670100-34670100 on Build GRCh38
rs13049028 Chr.21: 34670280-34670280 on Build GRCh38
rs770904205 Chr.21: 34670235-34670235 on Build GRCh38
rs779182457 Chr.21: 34669879-34669879 on Build GRCh38
rs773958725 Chr.21: 34670239-34670239 on Build GRCh38
rs369616665 Chr.21: 34670183-34670183 on Build GRCh38
rs1033283785 Chr.21: 34670064-34670064 on Build GRCh38
rs759120623 Chr.21: 34670070-34670070 on Build GRCh38
rs996585362 Chr.21: 34670107-34670107 on Build GRCh38
rs781366744 Chr.21: 34669952-34669952 on Build GRCh38
rs914584759 Chr.21: 34670149-34670149 on Build GRCh38
rs746231272 Chr.21: 34669880-34669880 on Build GRCh38
rs767040108 Chr.21: 34670036-34670065 on Build GRCh38
rs13049745 Chr.21: 34670179-34670179 on Build GRCh38
rs201318042 Chr.21: 34669926-34669926 on Build GRCh38
rs767008014 Chr.21: 34669915-34669915 on Build GRCh38
rs558777604 Chr.21: 34670298-34670298 on Build GRCh38
rs1045738840 Chr.21: 34669887-34669887 on Build GRCh38
rs878940978 Chr.21: 34670176-34670176 on Build GRCh38
rs981987625 Chr.21: 34669844-34669844 on Build GRCh38
rs769814618 Chr.21: 34669963-34669963 on Build GRCh38

View Full Product Details