Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18911558-18912108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR6
Entrez Gene ID
8214
Gene Name
DiGeorge syndrome critical region gene 6
Gene Aliases
-
UniGene
Hs.474185
Gene Chromosome Location
Chr.22: 18906223-18912088 on Build GRCh38
Transcripts
NM_005675.4 XM_011530394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849608 c.535C>T p.L179L Chr.22: 18911561-18911561 on Build GRCh38
3552086 c.607C>T p.P203S Chr.22: 18911633-18911633 on Build GRCh38
6422689 c.598C>A p.L200I Chr.22: 18911624-18911624 on Build GRCh38
6905344 c.547A>G p.I183V Chr.22: 18911573-18911573 on Build GRCh38
5748232 c.581G>T p.G194V Chr.22: 18911607-18911607 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs947654438 Chr.22: 18911747-18911747 on Build GRCh38
rs775919729 Chr.22: 18911641-18911641 on Build GRCh38
rs755763670 Chr.22: 18911690-18911690 on Build GRCh38
rs367589842 Chr.22: 18911620-18911620 on Build GRCh38
rs3208948 Chr.22: 18911969-18911969 on Build GRCh38
rs770114783 Chr.22: 18911628-18911628 on Build GRCh38
rs752640458 Chr.22: 18911608-18911608 on Build GRCh38
rs1056779 Chr.22: 18911884-18911884 on Build GRCh38
rs992040909 Chr.22: 18911916-18911916 on Build GRCh38
rs960169367 Chr.22: 18911911-18911911 on Build GRCh38
rs142204172 Chr.22: 18911655-18911655 on Build GRCh38
rs780757127 Chr.22: 18911623-18911623 on Build GRCh38
rs750664858 Chr.22: 18911660-18911660 on Build GRCh38
rs550314841 Chr.22: 18912044-18912044 on Build GRCh38
rs200501734 Chr.22: 18911901-18911902 on Build GRCh38
rs8443 Chr.22: 18911813-18911813 on Build GRCh38
rs5993411 Chr.22: 18911851-18911851 on Build GRCh38
rs1008435373 Chr.22: 18911786-18911786 on Build GRCh38
rs749619008 Chr.22: 18911700-18911700 on Build GRCh38
rs1036145110 Chr.22: 18911908-18911908 on Build GRCh38
rs1043361984 Chr.22: 18911755-18911755 on Build GRCh38
rs374709285 Chr.22: 18911777-18911777 on Build GRCh38
rs446213 Chr.22: 18911974-18911974 on Build GRCh38
rs41289055 Chr.22: 18911611-18911611 on Build GRCh38
rs1039557088 Chr.22: 18911796-18911796 on Build GRCh38
rs576442019 Chr.22: 18911902-18911902 on Build GRCh38
rs140149935 Chr.22: 18911687-18911687 on Build GRCh38
rs10602 Chr.22: 18911971-18911971 on Build GRCh38
rs767863398 Chr.22: 18911658-18911658 on Build GRCh38
rs764980774 Chr.22: 18911670-18911670 on Build GRCh38
rs3893662 Chr.22: 18912086-18912086 on Build GRCh38
rs769267295 Chr.22: 18911651-18911651 on Build GRCh38
rs878905568 Chr.22: 18911639-18911639 on Build GRCh38
rs1056797 Chr.22: 18911643-18911643 on Build GRCh38
rs776694376 Chr.22: 18911721-18911723 on Build GRCh38
rs756701449 Chr.22: 18911682-18911682 on Build GRCh38
rs148226630 Chr.22: 18911675-18911675 on Build GRCh38
rs1039544043 Chr.22: 18912078-18912078 on Build GRCh38
rs754669235 Chr.22: 18911604-18911604 on Build GRCh38
rs377419228 Chr.22: 18911727-18911727 on Build GRCh38
rs767801626 Chr.22: 18911591-18911591 on Build GRCh38
rs951395393 Chr.22: 18911900-18911900 on Build GRCh38
rs899722561 Chr.22: 18911818-18911818 on Build GRCh38
rs943814970 Chr.22: 18912072-18912072 on Build GRCh38
rs777313227 Chr.22: 18911613-18911613 on Build GRCh38
rs759583478 Chr.22: 18911723-18911723 on Build GRCh38
rs553683578 Chr.22: 18911606-18911606 on Build GRCh38
rs201062991 Chr.22: 18911710-18911710 on Build GRCh38
rs147635706 Chr.22: 18911578-18911578 on Build GRCh38
rs751075037 Chr.22: 18911677-18911677 on Build GRCh38
rs746258984 Chr.22: 18911626-18911626 on Build GRCh38
rs145917349 Chr.22: 18911665-18911665 on Build GRCh38
rs371344846 Chr.22: 18911685-18911685 on Build GRCh38
rs374844855 Chr.22: 18911697-18911697 on Build GRCh38
rs935090798 Chr.22: 18911766-18911766 on Build GRCh38
rs541960873 Chr.22: 18911943-18911943 on Build GRCh38
rs890885463 Chr.22: 18911780-18911780 on Build GRCh38
rs751661790 Chr.22: 18911583-18911583 on Build GRCh38
rs995450940 Chr.22: 18911845-18911845 on Build GRCh38
rs1052106867 Chr.22: 18912012-18912012 on Build GRCh38
rs916110710 Chr.22: 18911949-18911949 on Build GRCh38
rs934956105 Chr.22: 18911994-18911994 on Build GRCh38
rs11541280 Chr.22: 18911600-18911600 on Build GRCh38
rs1026968240 Chr.22: 18911894-18911894 on Build GRCh38
rs533479304 Chr.22: 18912035-18912035 on Build GRCh38
rs200250377 Chr.22: 18911629-18911629 on Build GRCh38
rs770600670 Chr.22: 18911622-18911622 on Build GRCh38
rs562059184 Chr.22: 18911961-18911961 on Build GRCh38
rs762178129 Chr.22: 18911590-18911590 on Build GRCh38
rs924974529 Chr.22: 18911757-18911757 on Build GRCh38
rs912272576 Chr.22: 18912016-18912016 on Build GRCh38

View Full Product Details