Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9781497-9782036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

SNP Detail

SNP ID Chromosome Location
rs906414808 Chr.4: 9781526-9781526 on Build GRCh38
rs558226166 Chr.4: 9781903-9781903 on Build GRCh38
rs770087035 Chr.4: 9781999-9781999 on Build GRCh38
rs542146372 Chr.4: 9782009-9782009 on Build GRCh38
rs960657255 Chr.4: 9781970-9781970 on Build GRCh38
rs554715522 Chr.4: 9781797-9781797 on Build GRCh38
rs769470252 Chr.4: 9781520-9781520 on Build GRCh38
rs927974472 Chr.4: 9781660-9781660 on Build GRCh38
rs967180192 Chr.4: 9781932-9781932 on Build GRCh38
rs780448872 Chr.4: 9782000-9782000 on Build GRCh38
rs749289106 Chr.4: 9782008-9782008 on Build GRCh38
rs531790860 Chr.4: 9781624-9781624 on Build GRCh38
rs375251229 Chr.4: 9781861-9781861 on Build GRCh38
rs550305570 Chr.4: 9781635-9781635 on Build GRCh38
rs576892102 Chr.4: 9781904-9781904 on Build GRCh38
rs540716117 Chr.4: 9781521-9781521 on Build GRCh38
rs886173375 Chr.4: 9781743-9781743 on Build GRCh38
rs954180082 Chr.4: 9781631-9781631 on Build GRCh38
rs373637865 Chr.4: 9781704-9781704 on Build GRCh38
rs994680288 Chr.4: 9781583-9781583 on Build GRCh38
rs895019785 Chr.4: 9781583-9781583 on Build GRCh38
rs998429725 Chr.4: 9781616-9781616 on Build GRCh38
rs537286412 Chr.4: 9781870-9781870 on Build GRCh38
rs2076907 Chr.4: 9781804-9781804 on Build GRCh38
rs1022491049 Chr.4: 9781885-9781885 on Build GRCh38
rs954313936 Chr.4: 9781831-9781831 on Build GRCh38
rs529978867 Chr.4: 9781590-9781590 on Build GRCh38
rs978486227 Chr.4: 9781647-9781647 on Build GRCh38
rs990837435 Chr.4: 9781972-9781972 on Build GRCh38
rs533889618 Chr.4: 9781699-9781699 on Build GRCh38
rs187411304 Chr.4: 9781584-9781584 on Build GRCh38
rs886206991 Chr.4: 9781589-9781589 on Build GRCh38
rs548367019 Chr.4: 9781597-9781597 on Build GRCh38
rs1055014617 Chr.4: 9781683-9781683 on Build GRCh38
rs536560116 Chr.4: 9781788-9781788 on Build GRCh38
rs563493722 Chr.4: 9781601-9781601 on Build GRCh38
rs939279297 Chr.4: 9781554-9781554 on Build GRCh38
rs768752880 Chr.4: 9782013-9782013 on Build GRCh38
rs915290036 Chr.4: 9781636-9781636 on Build GRCh38
rs539022301 Chr.4: 9781676-9781676 on Build GRCh38
rs571812427 Chr.4: 9781644-9781644 on Build GRCh38
rs774344700 Chr.4: 9782014-9782014 on Build GRCh38
rs1049349176 Chr.4: 9782001-9782001 on Build GRCh38
rs866013074 Chr.4: 9781727-9781727 on Build GRCh38
rs372681931 Chr.4: 9781935-9781935 on Build GRCh38
rs368710755 Chr.4: 9781998-9781998 on Build GRCh38
rs1040439284 Chr.4: 9781769-9781769 on Build GRCh38
rs749309741 Chr.4: 9781830-9781830 on Build GRCh38
rs1054946571 Chr.4: 9781579-9781579 on Build GRCh38
rs1004925094 Chr.4: 9781595-9781595 on Build GRCh38
rs371572831 Chr.4: 9781818-9781818 on Build GRCh38
rs12504432 Chr.4: 9781673-9781673 on Build GRCh38
rs927943382 Chr.4: 9781947-9781947 on Build GRCh38
rs1004658201 Chr.4: 9781758-9781758 on Build GRCh38
rs952516156 Chr.4: 9781779-9781779 on Build GRCh38
rs980951078 Chr.4: 9781523-9781523 on Build GRCh38
rs547504779 Chr.4: 9781686-9781686 on Build GRCh38
rs796966952 Chr.4: 9781741-9781741 on Build GRCh38
rs965580700 Chr.4: 9781605-9781605 on Build GRCh38
rs901548920 Chr.4: 9781786-9781786 on Build GRCh38
rs939248070 Chr.4: 9781681-9781681 on Build GRCh38
rs559099689 Chr.4: 9781980-9781980 on Build GRCh38
rs1011465129 Chr.4: 9781877-9781877 on Build GRCh38

View Full Product Details