Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74387273-74387787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
UniGene
Hs.115263
Gene Chromosome Location
Chr.4: 74365143-74388760 on Build GRCh38
Transcripts
NM_001432.2

SNP Detail

SNP ID Chromosome Location
rs72859385 Chr.4: 74387606-74387606 on Build GRCh38
rs376445765 Chr.4: 74387725-74387725 on Build GRCh38
rs985558865 Chr.4: 74387740-74387740 on Build GRCh38
rs1031077225 Chr.4: 74387550-74387550 on Build GRCh38
rs76514531 Chr.4: 74387672-74387672 on Build GRCh38
rs976676034 Chr.4: 74387395-74387395 on Build GRCh38
rs879262365 Chr.4: 74387490-74387490 on Build GRCh38
rs143039086 Chr.4: 74387555-74387555 on Build GRCh38
rs929818906 Chr.4: 74387417-74387417 on Build GRCh38
rs1012419321 Chr.4: 74387475-74387478 on Build GRCh38
rs949738267 Chr.4: 74387709-74387709 on Build GRCh38
rs893985127 Chr.4: 74387760-74387760 on Build GRCh38
rs911138905 Chr.4: 74387349-74387349 on Build GRCh38
rs1046642608 Chr.4: 74387515-74387515 on Build GRCh38
rs182106804 Chr.4: 74387751-74387751 on Build GRCh38
rs1005618218 Chr.4: 74387659-74387659 on Build GRCh38
rs925944192 Chr.4: 74387531-74387531 on Build GRCh38
rs56858247 Chr.4: 74387727-74387727 on Build GRCh38
rs762893646 Chr.4: 74387452-74387452 on Build GRCh38
rs370709780 Chr.4: 74387382-74387382 on Build GRCh38
rs532269363 Chr.4: 74387634-74387634 on Build GRCh38
rs761385364 Chr.4: 74387487-74387487 on Build GRCh38
rs961929548 Chr.4: 74387700-74387700 on Build GRCh38
rs1055477296 Chr.4: 74387698-74387698 on Build GRCh38
rs977308678 Chr.4: 74387364-74387364 on Build GRCh38
rs906677682 Chr.4: 74387543-74387543 on Build GRCh38
rs976643396 Chr.4: 74387599-74387599 on Build GRCh38
rs909627861 Chr.4: 74387685-74387685 on Build GRCh38
rs954152552 Chr.4: 74387732-74387732 on Build GRCh38
rs572498771 Chr.4: 74387569-74387569 on Build GRCh38
rs776157463 Chr.4: 74387467-74387467 on Build GRCh38
rs1017734 Chr.4: 74387403-74387403 on Build GRCh38
rs550759723 Chr.4: 74387646-74387646 on Build GRCh38
rs998400727 Chr.4: 74387707-74387707 on Build GRCh38
rs115110969 Chr.4: 74387549-74387549 on Build GRCh38
rs1053721376 Chr.4: 74387554-74387554 on Build GRCh38
rs5859429 Chr.4: 74387448-74387448 on Build GRCh38
rs747001497 Chr.4: 74387434-74387434 on Build GRCh38
rs578232016 Chr.4: 74387306-74387306 on Build GRCh38
rs1054779192 Chr.4: 74387647-74387647 on Build GRCh38
rs562494608 Chr.4: 74387670-74387670 on Build GRCh38
rs1046656288 Chr.4: 74387624-74387624 on Build GRCh38
rs958403126 Chr.4: 74387552-74387552 on Build GRCh38
rs894148400 Chr.4: 74387450-74387450 on Build GRCh38
rs1017736 Chr.4: 74387426-74387426 on Build GRCh38
rs1017735 Chr.4: 74387419-74387419 on Build GRCh38
rs941068012 Chr.4: 74387691-74387691 on Build GRCh38
rs888127107 Chr.4: 74387655-74387655 on Build GRCh38
rs941027457 Chr.4: 74387432-74387432 on Build GRCh38
rs906325384 Chr.4: 74387629-74387629 on Build GRCh38
rs1046620639 Chr.4: 74387719-74387719 on Build GRCh38
rs189297131 Chr.4: 74387600-74387600 on Build GRCh38

View Full Product Details