Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83429059-83429619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
UniGene
Hs.480101
Gene Chromosome Location
Chr.4: 83407343-83455883 on Build GRCh38
Transcripts
NM_001297755.1 NM_001297756.1 NM_001297757.1 NM_133636.3 XM_005262711.1 XM_005262713.2 XM_006714076.2 XM_017007679.1 XM_017007680.1 XM_017007682.1 XM_017007683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545573 c.2467G>A p.E823K Chr.4: 83429575-83429575 on Build GRCh38
2955982 c.2461G>A p.E821K Chr.4: 83429581-83429581 on Build GRCh38
7370947 c.2494A>G p.K832E Chr.4: 83429548-83429548 on Build GRCh38
4126389 c.2446A>T p.T816S Chr.4: 83429596-83429596 on Build GRCh38
3606606 c.2459C>T p.S820F Chr.4: 83429583-83429583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759897275 Chr.4: 83429278-83429278 on Build GRCh38
rs550270045 Chr.4: 83429458-83429458 on Build GRCh38
rs532039377 Chr.4: 83429434-83429434 on Build GRCh38
rs141425615 Chr.4: 83429176-83429176 on Build GRCh38
rs183975345 Chr.4: 83429429-83429429 on Build GRCh38
rs927130009 Chr.4: 83429337-83429337 on Build GRCh38
rs148996864 Chr.4: 83429539-83429539 on Build GRCh38
rs1004508172 Chr.4: 83429262-83429262 on Build GRCh38
rs186198616 Chr.4: 83429217-83429217 on Build GRCh38
rs544233750 Chr.4: 83429216-83429216 on Build GRCh38
rs1057420717 Chr.4: 83429360-83429360 on Build GRCh38
rs75238520 Chr.4: 83429586-83429586 on Build GRCh38
rs1049037418 Chr.4: 83429555-83429555 on Build GRCh38
rs775881310 Chr.4: 83429389-83429389 on Build GRCh38
rs979987594 Chr.4: 83429335-83429335 on Build GRCh38
rs953823870 Chr.4: 83429314-83429314 on Build GRCh38
rs1031680651 Chr.4: 83429484-83429484 on Build GRCh38
rs1009111808 Chr.4: 83429443-83429443 on Build GRCh38
rs1044084988 Chr.4: 83429432-83429432 on Build GRCh38
rs996333879 Chr.4: 83429417-83429417 on Build GRCh38
rs1037463251 Chr.4: 83429153-83429153 on Build GRCh38
rs527919420 Chr.4: 83429303-83429303 on Build GRCh38
rs775735529 Chr.4: 83429548-83429548 on Build GRCh38
rs755735927 Chr.4: 83429589-83429589 on Build GRCh38
rs916872123 Chr.4: 83429428-83429428 on Build GRCh38
rs759691915 Chr.4: 83429538-83429538 on Build GRCh38
rs949790817 Chr.4: 83429174-83429174 on Build GRCh38
rs778421152 Chr.4: 83429488-83429488 on Build GRCh38
rs751369936 Chr.4: 83429573-83429573 on Build GRCh38
rs763940386 Chr.4: 83429569-83429569 on Build GRCh38
rs956710389 Chr.4: 83429249-83429249 on Build GRCh38
rs899323112 Chr.4: 83429170-83429170 on Build GRCh38
rs886120888 Chr.4: 83429126-83429126 on Build GRCh38
rs747408542 Chr.4: 83429491-83429491 on Build GRCh38
rs577687824 Chr.4: 83429235-83429235 on Build GRCh38
rs1056768655 Chr.4: 83429103-83429103 on Build GRCh38
rs772624401 Chr.4: 83429483-83429483 on Build GRCh38
rs1037451995 Chr.4: 83429384-83429384 on Build GRCh38
rs191001525 Chr.4: 83429292-83429292 on Build GRCh38
rs890638376 Chr.4: 83429431-83429431 on Build GRCh38
rs767524211 Chr.4: 83429585-83429585 on Build GRCh38
rs938587045 Chr.4: 83429350-83429350 on Build GRCh38
rs545126055 Chr.4: 83429286-83429286 on Build GRCh38
rs898937889 Chr.4: 83429391-83429391 on Build GRCh38
rs907501161 Chr.4: 83429361-83429361 on Build GRCh38
rs1004595266 Chr.4: 83429149-83429149 on Build GRCh38
rs916942955 Chr.4: 83429124-83429124 on Build GRCh38
rs955012726 Chr.4: 83429215-83429215 on Build GRCh38
rs200219888 Chr.4: 83429556-83429556 on Build GRCh38
rs573976271 Chr.4: 83429177-83429177 on Build GRCh38
rs776524223 Chr.4: 83429508-83429508 on Build GRCh38
rs1010903119 Chr.4: 83429505-83429505 on Build GRCh38
rs998245331 Chr.4: 83429301-83429301 on Build GRCh38
rs372124994 Chr.4: 83429191-83429191 on Build GRCh38
rs542907493 Chr.4: 83429364-83429364 on Build GRCh38
rs74836884 Chr.4: 83429501-83429501 on Build GRCh38
rs772384734 Chr.4: 83429282-83429282 on Build GRCh38
rs938332305 Chr.4: 83429099-83429099 on Build GRCh38
rs1030978277 Chr.4: 83429244-83429244 on Build GRCh38
rs762719876 Chr.4: 83429550-83429550 on Build GRCh38
rs761611311 Chr.4: 83429581-83429581 on Build GRCh38

View Full Product Details