Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142658196-142658789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JRK
Entrez Gene ID
8629
Gene Name
Jrk helix-turn-helix protein
Gene Aliases
JH8, jerky
UniGene
Hs.535903
Gene Chromosome Location
Chr.8: 142651501-142670132 on Build GRCh38
Transcripts
NM_001077527.2 NM_001279352.1 NM_003724.3 XM_006716677.3 XM_006716678.3 XM_011517354.1 XM_011517355.1

SNP Detail

SNP ID Chromosome Location
rs113894368 Chr.8: 142658342-142658342 on Build GRCh38
rs587597019 Chr.8: 142658673-142658673 on Build GRCh38
rs1033520154 Chr.8: 142658288-142658288 on Build GRCh38
rs782499095 Chr.8: 142658335-142658335 on Build GRCh38
rs80055830 Chr.8: 142658566-142658566 on Build GRCh38
rs10656550 Chr.8: 142658405-142658406 on Build GRCh38
rs962951022 Chr.8: 142658690-142658690 on Build GRCh38
rs188539963 Chr.8: 142658249-142658249 on Build GRCh38
rs147432825 Chr.8: 142658344-142658344 on Build GRCh38
rs587680185 Chr.8: 142658420-142658420 on Build GRCh38
rs587610365 Chr.8: 142658274-142658274 on Build GRCh38
rs895007807 Chr.8: 142658585-142658585 on Build GRCh38
rs757272369 Chr.8: 142658446-142658446 on Build GRCh38
rs782611103 Chr.8: 142658558-142658558 on Build GRCh38
rs587613931 Chr.8: 142658498-142658498 on Build GRCh38
rs587745634 Chr.8: 142658307-142658307 on Build GRCh38
rs1801343 Chr.8: 142658562-142658562 on Build GRCh38
rs1056012 Chr.8: 142658755-142658755 on Build GRCh38
rs587653137 Chr.8: 142658584-142658584 on Build GRCh38
rs587703800 Chr.8: 142658266-142658266 on Build GRCh38
rs587659611 Chr.8: 142658300-142658300 on Build GRCh38
rs73379607 Chr.8: 142658602-142658602 on Build GRCh38
rs76079990 Chr.8: 142658240-142658240 on Build GRCh38
rs587713594 Chr.8: 142658672-142658672 on Build GRCh38
rs974377335 Chr.8: 142658503-142658503 on Build GRCh38
rs150553786 Chr.8: 142658597-142658597 on Build GRCh38
rs782321651 Chr.8: 142658600-142658600 on Build GRCh38
rs760264086 Chr.8: 142658311-142658311 on Build GRCh38
rs782507383 Chr.8: 142658406-142658407 on Build GRCh38
rs207470045 Chr.8: 142658633-142658633 on Build GRCh38
rs4401833 Chr.8: 142658317-142658317 on Build GRCh38
rs587774813 Chr.8: 142658391-142658392 on Build GRCh38
rs150398750 Chr.8: 142658710-142658711 on Build GRCh38
rs1007004279 Chr.8: 142658662-142658662 on Build GRCh38
rs999361106 Chr.8: 142658607-142658607 on Build GRCh38
rs587694912 Chr.8: 142658747-142658747 on Build GRCh38
rs955346420 Chr.8: 142658610-142658610 on Build GRCh38
rs928937764 Chr.8: 142658231-142658231 on Build GRCh38
rs148047570 Chr.8: 142658518-142658518 on Build GRCh38
rs375924854 Chr.8: 142658239-142658239 on Build GRCh38
rs72690908 Chr.8: 142658441-142658441 on Build GRCh38
rs898860194 Chr.8: 142658225-142658225 on Build GRCh38
rs372428907 Chr.8: 142658341-142658341 on Build GRCh38
rs911407873 Chr.8: 142658484-142658484 on Build GRCh38
rs782033875 Chr.8: 142658753-142658753 on Build GRCh38
rs587644987 Chr.8: 142658552-142658552 on Build GRCh38
rs587752144 Chr.8: 142658343-142658343 on Build GRCh38
rs587644186 Chr.8: 142658609-142658609 on Build GRCh38
rs918842192 Chr.8: 142658504-142658504 on Build GRCh38
rs587604121 Chr.8: 142658711-142658711 on Build GRCh38
rs140828733 Chr.8: 142658584-142658588 on Build GRCh38
rs1010836990 Chr.8: 142658327-142658327 on Build GRCh38
rs902429545 Chr.8: 142658606-142658606 on Build GRCh38
rs587749238 Chr.8: 142658711-142658712 on Build GRCh38
rs4336593 Chr.8: 142658233-142658233 on Build GRCh38
rs985926030 Chr.8: 142658457-142658457 on Build GRCh38
rs941514770 Chr.8: 142658497-142658497 on Build GRCh38
rs141745134 Chr.8: 142658523-142658523 on Build GRCh38
rs1003258836 Chr.8: 142658258-142658258 on Build GRCh38
rs587681623 Chr.8: 142658251-142658251 on Build GRCh38
rs1029585530 Chr.8: 142658428-142658428 on Build GRCh38

View Full Product Details