Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129636725-129637241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2 XM_005251939.3 XM_017014642.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2 XM_005251939.3 XM_017014642.1

SNP Detail

SNP ID Chromosome Location
rs932341744 Chr.9: 129636864-129636864 on Build GRCh38
rs905821240 Chr.9: 129636997-129636997 on Build GRCh38
rs1045647 Chr.9: 129636921-129636921 on Build GRCh38
rs996428346 Chr.9: 129636919-129636923 on Build GRCh38
rs184441078 Chr.9: 129637203-129637203 on Build GRCh38
rs1040857605 Chr.9: 129637151-129637151 on Build GRCh38
rs922320624 Chr.9: 129637171-129637171 on Build GRCh38
rs970122273 Chr.9: 129636755-129636755 on Build GRCh38
rs992933959 Chr.9: 129636797-129636797 on Build GRCh38
rs946431940 Chr.9: 129636832-129636832 on Build GRCh38
rs531402534 Chr.9: 129636754-129636754 on Build GRCh38
rs961871148 Chr.9: 129636996-129636996 on Build GRCh38
rs980102892 Chr.9: 129636825-129636825 on Build GRCh38
rs940257990 Chr.9: 129637164-129637164 on Build GRCh38
rs768093741 Chr.9: 129636971-129636971 on Build GRCh38
rs547263904 Chr.9: 129637033-129637033 on Build GRCh38
rs979556483 Chr.9: 129637116-129637116 on Build GRCh38
rs888676681 Chr.9: 129636922-129636927 on Build GRCh38
rs957964543 Chr.9: 129637049-129637049 on Build GRCh38
rs534705532 Chr.9: 129636913-129636913 on Build GRCh38
rs927907266 Chr.9: 129636783-129636783 on Build GRCh38
rs960603017 Chr.9: 129637034-129637034 on Build GRCh38
rs1043524551 Chr.9: 129636845-129636845 on Build GRCh38
rs920918908 Chr.9: 129636862-129636862 on Build GRCh38
rs921024325 Chr.9: 129637132-129637132 on Build GRCh38
rs778152931 Chr.9: 129636746-129636746 on Build GRCh38
rs114929263 Chr.9: 129637204-129637204 on Build GRCh38
rs1035320817 Chr.9: 129637007-129637007 on Build GRCh38
rs867473890 Chr.9: 129636931-129636931 on Build GRCh38
rs10988455 Chr.9: 129636813-129636813 on Build GRCh38
rs1005657446 Chr.9: 129636868-129636868 on Build GRCh38
rs1020799077 Chr.9: 129637097-129637097 on Build GRCh38
rs760746653 Chr.9: 129636798-129636799 on Build GRCh38
rs967910281 Chr.9: 129637111-129637111 on Build GRCh38
rs945250033 Chr.9: 129637141-129637141 on Build GRCh38
rs945116069 Chr.9: 129636887-129636887 on Build GRCh38
rs555313439 Chr.9: 129637113-129637113 on Build GRCh38
rs1051743469 Chr.9: 129636865-129636865 on Build GRCh38
rs1023367969 Chr.9: 129636753-129636753 on Build GRCh38
rs566105082 Chr.9: 129636899-129636899 on Build GRCh38
rs866184342 Chr.9: 129637138-129637138 on Build GRCh38
rs961458805 Chr.9: 129636779-129636779 on Build GRCh38
rs912258193 Chr.9: 129637150-129637150 on Build GRCh38
rs1023057677 Chr.9: 129636982-129636982 on Build GRCh38
rs987986578 Chr.9: 129636812-129636812 on Build GRCh38
rs1006233235 Chr.9: 129636942-129636942 on Build GRCh38
rs879333833 Chr.9: 129637193-129637193 on Build GRCh38
rs1040807067 Chr.9: 129636910-129636910 on Build GRCh38
rs539692774 Chr.9: 129636999-129636999 on Build GRCh38
rs948741295 Chr.9: 129636824-129636824 on Build GRCh38
rs1037284402 Chr.9: 129637165-129637165 on Build GRCh38
rs1016186919 Chr.9: 129636744-129636744 on Build GRCh38
rs535818787 Chr.9: 129637103-129637103 on Build GRCh38
rs898659407 Chr.9: 129637190-129637190 on Build GRCh38
rs778289572 Chr.9: 129637005-129637005 on Build GRCh38
rs960505687 Chr.9: 129636796-129636796 on Build GRCh38
rs905960784 Chr.9: 129637205-129637205 on Build GRCh38
rs891394594 Chr.9: 129636884-129636884 on Build GRCh38
rs981063738 Chr.9: 129636776-129636776 on Build GRCh38
rs913728926 Chr.9: 129637074-129637074 on Build GRCh38
rs980071442 Chr.9: 129637026-129637026 on Build GRCh38
rs7043953 Chr.9: 129636831-129636831 on Build GRCh38
rs748779420 Chr.9: 129636932-129636932 on Build GRCh38
rs986494239 Chr.9: 129637139-129637139 on Build GRCh38
rs567086033 Chr.9: 129637055-129637055 on Build GRCh38

View Full Product Details