Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42701760-42702257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EZH1
Entrez Gene ID
2145
Gene Name
enhancer of zeste 1 polycomb repressive complex 2 subunit
Gene Aliases
KMT6B
UniGene
Hs.194669
Gene Chromosome Location
Chr.17: 42700275-42745096 on Build GRCh38
Transcripts
NM_001321079.1 NM_001321081.1 NM_001321082.1 NM_001991.4 XM_005257145.2 XM_011524517.2 XM_017024350.1 XM_017024351.1 XM_017024352.1

SNP Detail

SNP ID Chromosome Location
rs747999188 Chr.17: 42702232-42702232 on Build GRCh38
rs58857209 Chr.17: 42702236-42702236 on Build GRCh38
rs747148332 Chr.17: 42702182-42702182 on Build GRCh38
rs888395527 Chr.17: 42702178-42702178 on Build GRCh38
rs755859570 Chr.17: 42702227-42702227 on Build GRCh38
rs757735785 Chr.17: 42701994-42701994 on Build GRCh38
rs754477674 Chr.17: 42702132-42702132 on Build GRCh38
rs112608760 Chr.17: 42701923-42701923 on Build GRCh38
rs140380284 Chr.17: 42702001-42702001 on Build GRCh38
rs996772538 Chr.17: 42701810-42701810 on Build GRCh38
rs915516791 Chr.17: 42702062-42702062 on Build GRCh38
rs560426606 Chr.17: 42701862-42701862 on Build GRCh38
rs932794183 Chr.17: 42701984-42701984 on Build GRCh38
rs911876950 Chr.17: 42702081-42702081 on Build GRCh38
rs906153060 Chr.17: 42702222-42702222 on Build GRCh38
rs137861843 Chr.17: 42702177-42702177 on Build GRCh38
rs145260558 Chr.17: 42702124-42702124 on Build GRCh38
rs944809306 Chr.17: 42702115-42702115 on Build GRCh38
rs1036740566 Chr.17: 42702195-42702195 on Build GRCh38
rs577690061 Chr.17: 42701920-42701920 on Build GRCh38
rs1048315397 Chr.17: 42701840-42701840 on Build GRCh38
rs959498156 Chr.17: 42701992-42701992 on Build GRCh38
rs574017383 Chr.17: 42701838-42701838 on Build GRCh38
rs989708961 Chr.17: 42702046-42702046 on Build GRCh38
rs968507778 Chr.17: 42702186-42702186 on Build GRCh38
rs780078215 Chr.17: 42701970-42701970 on Build GRCh38
rs531343921 Chr.17: 42702230-42702230 on Build GRCh38
rs527859545 Chr.17: 42702094-42702094 on Build GRCh38
rs1041295992 Chr.17: 42701948-42701948 on Build GRCh38
rs947081766 Chr.17: 42701877-42701877 on Build GRCh38
rs73983746 Chr.17: 42701899-42701899 on Build GRCh38
rs1003702136 Chr.17: 42701951-42701951 on Build GRCh38
rs556244308 Chr.17: 42701900-42701900 on Build GRCh38
rs1004530491 Chr.17: 42702192-42702192 on Build GRCh38
rs932452207 Chr.17: 42702169-42702169 on Build GRCh38
rs560775330 Chr.17: 42702009-42702009 on Build GRCh38
rs1012702739 Chr.17: 42702148-42702148 on Build GRCh38
rs372485243 Chr.17: 42701971-42701971 on Build GRCh38
rs921113596 Chr.17: 42702143-42702143 on Build GRCh38
rs145656198 Chr.17: 42701795-42701795 on Build GRCh38
rs746224632 Chr.17: 42701877-42701878 on Build GRCh38
rs1048995104 Chr.17: 42702038-42702038 on Build GRCh38
rs779845192 Chr.17: 42702008-42702008 on Build GRCh38
rs888541387 Chr.17: 42702095-42702095 on Build GRCh38
rs549773642 Chr.17: 42702233-42702233 on Build GRCh38
rs1003908781 Chr.17: 42701879-42701879 on Build GRCh38
rs906517221 Chr.17: 42702136-42702136 on Build GRCh38
rs1035556599 Chr.17: 42702144-42702144 on Build GRCh38
rs966919504 Chr.17: 42702066-42702066 on Build GRCh38
rs944285036 Chr.17: 42701929-42701929 on Build GRCh38
rs978150333 Chr.17: 42702068-42702068 on Build GRCh38

View Full Product Details