Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42953634-42954057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARSD1
Entrez Gene ID
80755
Gene Name
alanyl-tRNA synthetase domain containing 1
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42964498 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Gene Symbol
PTGES3L-AARSD1
Entrez Gene ID
100885850
Gene Name
PTGES3L-AARSD1 readthrough
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42980528 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134688 c.994G>C p.E332Q Chr.17: 42953738-42953738 on Build GRCh38
3189802 c.1333G>C p.E445Q Chr.17: 42953738-42953738 on Build GRCh38
3189801 c.1516G>C p.E506Q Chr.17: 42953738-42953738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775226969 Chr.17: 42953658-42953658 on Build GRCh38
rs759396729 Chr.17: 42953696-42953696 on Build GRCh38
rs375776812 Chr.17: 42953704-42953704 on Build GRCh38
rs150835196 Chr.17: 42953770-42953770 on Build GRCh38
rs772085294 Chr.17: 42953683-42953683 on Build GRCh38
rs373153048 Chr.17: 42953693-42953693 on Build GRCh38
rs554621479 Chr.17: 42953790-42953792 on Build GRCh38
rs910133125 Chr.17: 42953661-42953661 on Build GRCh38
rs940300239 Chr.17: 42953673-42953673 on Build GRCh38
rs777993509 Chr.17: 42953775-42953776 on Build GRCh38
rs755544751 Chr.17: 42953675-42953675 on Build GRCh38
rs769512083 Chr.17: 42953825-42953825 on Build GRCh38
rs753992752 Chr.17: 42953776-42953777 on Build GRCh38
rs764305008 Chr.17: 42953757-42953757 on Build GRCh38
rs781455295 Chr.17: 42953820-42953820 on Build GRCh38
rs199553345 Chr.17: 42953724-42953724 on Build GRCh38
rs980256083 Chr.17: 42953804-42953804 on Build GRCh38
rs753539451 Chr.17: 42953801-42953801 on Build GRCh38
rs571493075 Chr.17: 42953873-42953873 on Build GRCh38
rs758557782 Chr.17: 42953740-42953740 on Build GRCh38
rs759342582 Chr.17: 42953677-42953677 on Build GRCh38
rs983522950 Chr.17: 42953767-42953767 on Build GRCh38
rs973371954 Chr.17: 42954017-42954017 on Build GRCh38
rs143299815 Chr.17: 42953878-42953878 on Build GRCh38
rs374124347 Chr.17: 42953744-42953744 on Build GRCh38
rs771140509 Chr.17: 42953792-42953792 on Build GRCh38
rs761568961 Chr.17: 42953738-42953738 on Build GRCh38
rs775453978 Chr.17: 42953826-42953826 on Build GRCh38
rs560098074 Chr.17: 42953657-42953657 on Build GRCh38
rs780540729 Chr.17: 42953709-42953709 on Build GRCh38
rs961633000 Chr.17: 42953988-42953988 on Build GRCh38
rs866963244 Chr.17: 42953924-42953924 on Build GRCh38
rs538067394 Chr.17: 42953850-42953850 on Build GRCh38
rs1054710523 Chr.17: 42953899-42953899 on Build GRCh38
rs777730319 Chr.17: 42953773-42953773 on Build GRCh38
rs755451666 Chr.17: 42953749-42953749 on Build GRCh38
rs371040940 Chr.17: 42953991-42953991 on Build GRCh38
rs751205444 Chr.17: 42953726-42953726 on Build GRCh38
rs762491072 Chr.17: 42953720-42953720 on Build GRCh38
rs567342625 Chr.17: 42953798-42953798 on Build GRCh38
rs747256107 Chr.17: 42953692-42953692 on Build GRCh38
rs745644636 Chr.17: 42953823-42953823 on Build GRCh38
rs370114932 Chr.17: 42953736-42953736 on Build GRCh38
rs950276440 Chr.17: 42953776-42953776 on Build GRCh38
rs891492620 Chr.17: 42953918-42953918 on Build GRCh38
rs1055179786 Chr.17: 42953705-42953705 on Build GRCh38
rs372416896 Chr.17: 42953935-42953935 on Build GRCh38
rs775335493 Chr.17: 42953719-42953719 on Build GRCh38
rs369935771 Chr.17: 42953682-42953682 on Build GRCh38
rs778826708 Chr.17: 42953819-42953820 on Build GRCh38
rs753176340 Chr.17: 42953760-42953760 on Build GRCh38
rs147677113 Chr.17: 42953688-42953688 on Build GRCh38
rs549934434 Chr.17: 42953864-42953864 on Build GRCh38
rs148312040 Chr.17: 42953947-42953947 on Build GRCh38
rs112418201 Chr.17: 42953695-42953695 on Build GRCh38
rs747170345 Chr.17: 42953785-42953785 on Build GRCh38
rs953838390 Chr.17: 42953884-42953884 on Build GRCh38

View Full Product Details