Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31721548-31722044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf25
Entrez Gene ID
80739
Gene Name
chromosome 6 open reading frame 25
Gene Aliases
G6b, NG31
UniGene
Hs.247879
Gene Chromosome Location
Chr.6: 31720557-31726710 on Build GRCh38
Transcripts
XM_011514920.2 XM_011514921.2 XM_017011330.1 XM_017011332.1 XM_017011333.1 NM_025261.2

Gene Symbol
LY6G6C
Entrez Gene ID
80740
Gene Name
lymphocyte antigen 6 complex, locus G6C
Gene Aliases
C6orf24, G6c, NG24
UniGene
Hs.732272
Gene Chromosome Location
Chr.6: 31718648-31721734 on Build GRCh38
Transcripts
XM_011514920.2 XM_011514921.2 XM_017011330.1 XM_017011332.1 XM_017011333.1 NM_025261.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3346481 c.19C>A p.L7I Chr.6: 31721661-31721661 on Build GRCh38
5127439 c.1A>G p.M1V Chr.6: 31721679-31721679 on Build GRCh38
4586605 c.51A>G p.S17S Chr.6: 31721629-31721629 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142694990 Chr.6: 31721945-31721945 on Build GRCh38
rs932605750 Chr.6: 31721825-31721825 on Build GRCh38
rs976412043 Chr.6: 31721789-31721789 on Build GRCh38
rs1043326954 Chr.6: 31721628-31721628 on Build GRCh38
rs774953902 Chr.6: 31721616-31721616 on Build GRCh38
rs938760238 Chr.6: 31721570-31721570 on Build GRCh38
rs576421188 Chr.6: 31722000-31722000 on Build GRCh38
rs1036828171 Chr.6: 31721995-31721995 on Build GRCh38
rs370881879 Chr.6: 31721620-31721620 on Build GRCh38
rs549761735 Chr.6: 31721773-31721773 on Build GRCh38
rs749375292 Chr.6: 31721697-31721697 on Build GRCh38
rs943690546 Chr.6: 31721786-31721786 on Build GRCh38
rs939890041 Chr.6: 31721964-31721964 on Build GRCh38
rs374302350 Chr.6: 31721656-31721656 on Build GRCh38
rs755137977 Chr.6: 31721613-31721613 on Build GRCh38
rs897797730 Chr.6: 31721612-31721612 on Build GRCh38
rs1006687221 Chr.6: 31721595-31721595 on Build GRCh38
rs776426052 Chr.6: 31721582-31721582 on Build GRCh38
rs367797282 Chr.6: 31721720-31721720 on Build GRCh38
rs373625540 Chr.6: 31721719-31721719 on Build GRCh38
rs759142986 Chr.6: 31721601-31721601 on Build GRCh38
rs566022036 Chr.6: 31721897-31721897 on Build GRCh38
rs538663485 Chr.6: 31721834-31721834 on Build GRCh38
rs769313341 Chr.6: 31721780-31721780 on Build GRCh38
rs183618568 Chr.6: 31721682-31721682 on Build GRCh38
rs1030439741 Chr.6: 31721696-31721696 on Build GRCh38
rs200403508 Chr.6: 31721704-31721704 on Build GRCh38
rs780463007 Chr.6: 31721683-31721683 on Build GRCh38
rs750838682 Chr.6: 31721679-31721679 on Build GRCh38
rs752965796 Chr.6: 31721833-31721833 on Build GRCh38
rs867778260 Chr.6: 31721839-31721839 on Build GRCh38
rs547697236 Chr.6: 31721852-31721852 on Build GRCh38
rs535474301 Chr.6: 31721952-31721952 on Build GRCh38
rs571284450 Chr.6: 31721811-31721811 on Build GRCh38
rs867247314 Chr.6: 31721635-31721635 on Build GRCh38
rs376207722 Chr.6: 31721611-31721611 on Build GRCh38
rs757783978 Chr.6: 31721669-31721669 on Build GRCh38
rs921178386 Chr.6: 31721808-31721808 on Build GRCh38
rs531444868 Chr.6: 31721751-31721751 on Build GRCh38
rs536057526 Chr.6: 31721917-31721917 on Build GRCh38
rs909133301 Chr.6: 31721754-31721754 on Build GRCh38
rs747568109 Chr.6: 31721735-31721735 on Build GRCh38
rs897936366 Chr.6: 31722020-31722020 on Build GRCh38
rs764645100 Chr.6: 31721646-31721646 on Build GRCh38
rs755079434 Chr.6: 31721698-31721698 on Build GRCh38
rs769418639 Chr.6: 31721731-31721731 on Build GRCh38
rs904735427 Chr.6: 31721629-31721629 on Build GRCh38
rs145333192 Chr.6: 31721672-31721672 on Build GRCh38
rs745707757 Chr.6: 31721727-31721727 on Build GRCh38

View Full Product Details