Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31724022-31724515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf25
Entrez Gene ID
80739
Gene Name
chromosome 6 open reading frame 25
Gene Aliases
G6b, NG31
UniGene
Hs.247879
Gene Chromosome Location
Chr.6: 31720557-31726710 on Build GRCh38
Transcripts
NM_025260.3 NM_138272.2 NM_138273.2 NM_138274.2 NM_138275.2 NM_138277.2 XM_011514920.2 XM_011514921.2 XM_017011330.1 XM_017011331.1 XM_017011332.1 XM_017011333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667916 c.448G>A p.A150T Chr.6: 31724180-31724180 on Build GRCh38
4188329 c.452G>T p.G151V Chr.6: 31724184-31724184 on Build GRCh38
7315276 c.476T>C p.L159S Chr.6: 31724208-31724208 on Build GRCh38
6409243 c.476T>C p.L159S Chr.6: 31724208-31724208 on Build GRCh38
6409105 c.437C>T p.P146L Chr.6: 31724169-31724169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371384499 Chr.6: 31724094-31724094 on Build GRCh38
rs755446480 Chr.6: 31724144-31724144 on Build GRCh38
rs995477288 Chr.6: 31724427-31724427 on Build GRCh38
rs541503374 Chr.6: 31724152-31724152 on Build GRCh38
rs576980709 Chr.6: 31724049-31724049 on Build GRCh38
rs765711901 Chr.6: 31724145-31724145 on Build GRCh38
rs764312482 Chr.6: 31724280-31724280 on Build GRCh38
rs989775817 Chr.6: 31724256-31724256 on Build GRCh38
rs56404328 Chr.6: 31724099-31724099 on Build GRCh38
rs776095281 Chr.6: 31724249-31724249 on Build GRCh38
rs559558055 Chr.6: 31724185-31724185 on Build GRCh38
rs747672830 Chr.6: 31724234-31724235 on Build GRCh38
rs891232832 Chr.6: 31724464-31724464 on Build GRCh38
rs1029718199 Chr.6: 31724137-31724137 on Build GRCh38
rs551918822 Chr.6: 31724275-31724275 on Build GRCh38
rs753090851 Chr.6: 31724146-31724146 on Build GRCh38
rs768748167 Chr.6: 31724110-31724110 on Build GRCh38
rs776860500 Chr.6: 31724099-31724099 on Build GRCh38
rs773004644 Chr.6: 31724212-31724212 on Build GRCh38
rs563961447 Chr.6: 31724367-31724367 on Build GRCh38
rs756586386 Chr.6: 31724451-31724451 on Build GRCh38
rs528295848 Chr.6: 31724381-31724381 on Build GRCh38
rs372623741 Chr.6: 31724429-31724429 on Build GRCh38
rs753075972 Chr.6: 31724262-31724262 on Build GRCh38
rs1033898790 Chr.6: 31724059-31724059 on Build GRCh38
rs1029688491 Chr.6: 31724458-31724458 on Build GRCh38
rs774681472 Chr.6: 31724326-31724326 on Build GRCh38
rs777564633 Chr.6: 31724268-31724277 on Build GRCh38
rs188943328 Chr.6: 31724074-31724074 on Build GRCh38
rs985453732 Chr.6: 31724180-31724180 on Build GRCh38
rs747971800 Chr.6: 31724195-31724195 on Build GRCh38
rs767186994 Chr.6: 31724124-31724124 on Build GRCh38
rs766058752 Chr.6: 31724243-31724243 on Build GRCh38
rs17201018 Chr.6: 31724073-31724073 on Build GRCh38
rs936549120 Chr.6: 31724449-31724449 on Build GRCh38
rs777912728 Chr.6: 31724093-31724093 on Build GRCh38
rs778498927 Chr.6: 31724148-31724162 on Build GRCh38
rs765817037 Chr.6: 31724135-31724135 on Build GRCh38
rs753377838 Chr.6: 31724141-31724141 on Build GRCh38
rs550703840 Chr.6: 31724453-31724453 on Build GRCh38
rs767953348 Chr.6: 31724194-31724194 on Build GRCh38
rs765766974 Chr.6: 31724250-31724250 on Build GRCh38
rs760279890 Chr.6: 31724216-31724216 on Build GRCh38
rs1051395328 Chr.6: 31724304-31724304 on Build GRCh38
rs569532604 Chr.6: 31724484-31724484 on Build GRCh38
rs918712064 Chr.6: 31724255-31724255 on Build GRCh38
rs805306 Chr.6: 31724386-31724386 on Build GRCh38
rs768618279 Chr.6: 31724193-31724193 on Build GRCh38
rs997315195 Chr.6: 31724457-31724457 on Build GRCh38
rs762938609 Chr.6: 31724100-31724100 on Build GRCh38
rs778201725 Chr.6: 31724153-31724153 on Build GRCh38
rs947685569 Chr.6: 31724396-31724396 on Build GRCh38
rs377294775 Chr.6: 31724128-31724128 on Build GRCh38
rs563551173 Chr.6: 31724168-31724168 on Build GRCh38
rs758953666 Chr.6: 31724265-31724265 on Build GRCh38
rs764607406 Chr.6: 31724492-31724492 on Build GRCh38
rs375016856 Chr.6: 31724096-31724096 on Build GRCh38
rs760324850 Chr.6: 31724132-31724132 on Build GRCh38
rs1056765007 Chr.6: 31724119-31724119 on Build GRCh38
rs939259039 Chr.6: 31724115-31724115 on Build GRCh38
rs766664307 Chr.6: 31724205-31724205 on Build GRCh38
rs757982433 Chr.6: 31724262-31724263 on Build GRCh38
rs939821822 Chr.6: 31724382-31724382 on Build GRCh38
rs935669552 Chr.6: 31724221-31724221 on Build GRCh38
rs376188647 Chr.6: 31724120-31724120 on Build GRCh38
rs568499852 Chr.6: 31724415-31724415 on Build GRCh38
rs772314539 Chr.6: 31724091-31724091 on Build GRCh38
rs140027534 Chr.6: 31724170-31724170 on Build GRCh38
rs753008874 Chr.6: 31724428-31724428 on Build GRCh38
rs761564341 Chr.6: 31724123-31724123 on Build GRCh38
rs141994295 Chr.6: 31724173-31724173 on Build GRCh38

View Full Product Details