Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31726618-31727113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf25
Entrez Gene ID
80739
Gene Name
chromosome 6 open reading frame 25
Gene Aliases
G6b, NG31
UniGene
Hs.247879
Gene Chromosome Location
Chr.6: 31720557-31726710 on Build GRCh38
Transcripts
NM_025260.3 NM_138272.2 NM_138273.2 NM_138274.2 NM_138275.2 NM_138277.2 NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_025260.3 NM_138272.2 NM_138273.2 NM_138274.2 NM_138275.2 NM_138277.2 NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

SNP Detail

SNP ID Chromosome Location
rs555664979 Chr.6: 31727036-31727036 on Build GRCh38
rs1027995536 Chr.6: 31726923-31726923 on Build GRCh38
rs964736718 Chr.6: 31726677-31726677 on Build GRCh38
rs184076303 Chr.6: 31726732-31726732 on Build GRCh38
rs539518750 Chr.6: 31726828-31726828 on Build GRCh38
rs575331988 Chr.6: 31726746-31726746 on Build GRCh38
rs557920597 Chr.6: 31726968-31726968 on Build GRCh38
rs950564948 Chr.6: 31726986-31726986 on Build GRCh38
rs45565837 Chr.6: 31727082-31727082 on Build GRCh38
rs986803660 Chr.6: 31726735-31726735 on Build GRCh38
rs538999035 Chr.6: 31726673-31726673 on Build GRCh38
rs949481542 Chr.6: 31727075-31727075 on Build GRCh38
rs1017191971 Chr.6: 31726652-31726652 on Build GRCh38
rs568655179 Chr.6: 31726672-31726672 on Build GRCh38
rs983305553 Chr.6: 31727009-31727009 on Build GRCh38
rs918364409 Chr.6: 31726927-31726927 on Build GRCh38
rs764486664 Chr.6: 31726651-31726651 on Build GRCh38
rs953641082 Chr.6: 31726722-31726722 on Build GRCh38
rs908624094 Chr.6: 31727074-31727074 on Build GRCh38
rs944871107 Chr.6: 31726858-31726858 on Build GRCh38
rs915230214 Chr.6: 31726662-31726662 on Build GRCh38
rs145184926 Chr.6: 31726763-31726763 on Build GRCh38
rs888099289 Chr.6: 31727032-31727032 on Build GRCh38
rs1008510802 Chr.6: 31726646-31726646 on Build GRCh38
rs557338491 Chr.6: 31726687-31726687 on Build GRCh38
rs1036495296 Chr.6: 31726886-31726886 on Build GRCh38
rs573742405 Chr.6: 31727054-31727054 on Build GRCh38
rs1048071707 Chr.6: 31726984-31726984 on Build GRCh38
rs754324785 Chr.6: 31727006-31727006 on Build GRCh38
rs556626959 Chr.6: 31726650-31726650 on Build GRCh38

View Full Product Details