Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31728554-31729057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624718 c.275G>A p.W92* Chr.6: 31728887-31728887 on Build GRCh38
4676653 c.287G>A p.R96H Chr.6: 31728875-31728875 on Build GRCh38
5959800 c.203C>T p.S68L Chr.6: 31728959-31728959 on Build GRCh38
3624717 c.309C>T p.V103V Chr.6: 31728734-31728734 on Build GRCh38
366844 c.389T>C p.L130P Chr.6: 31728654-31728654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751453578 Chr.6: 31729020-31729020 on Build GRCh38
rs1055732783 Chr.6: 31728768-31728768 on Build GRCh38
rs371435542 Chr.6: 31728957-31728957 on Build GRCh38
rs756283879 Chr.6: 31728691-31728691 on Build GRCh38
rs766636105 Chr.6: 31728968-31728968 on Build GRCh38
rs866230738 Chr.6: 31728732-31728732 on Build GRCh38
rs979355881 Chr.6: 31728681-31728681 on Build GRCh38
rs780005525 Chr.6: 31728692-31728692 on Build GRCh38
rs756843527 Chr.6: 31728714-31728714 on Build GRCh38
rs376853159 Chr.6: 31728927-31728927 on Build GRCh38
rs758360957 Chr.6: 31728701-31728701 on Build GRCh38
rs767833091 Chr.6: 31728674-31728674 on Build GRCh38
rs751346909 Chr.6: 31728870-31728870 on Build GRCh38
rs773419342 Chr.6: 31728819-31728819 on Build GRCh38
rs372370927 Chr.6: 31728694-31728694 on Build GRCh38
rs533551218 Chr.6: 31728576-31728576 on Build GRCh38
rs765301432 Chr.6: 31729018-31729018 on Build GRCh38
rs776070291 Chr.6: 31728640-31728640 on Build GRCh38
rs771788581 Chr.6: 31728608-31728608 on Build GRCh38
rs775065727 Chr.6: 31728945-31728945 on Build GRCh38
rs746854218 Chr.6: 31728710-31728710 on Build GRCh38
rs11752493 Chr.6: 31728636-31728636 on Build GRCh38
rs182426985 Chr.6: 31728706-31728706 on Build GRCh38
rs778968206 Chr.6: 31728599-31728599 on Build GRCh38
rs375404521 Chr.6: 31728646-31728646 on Build GRCh38
rs752006519 Chr.6: 31728658-31728658 on Build GRCh38
rs766404608 Chr.6: 31728849-31728849 on Build GRCh38
rs745473395 Chr.6: 31728893-31728893 on Build GRCh38
rs755755817 Chr.6: 31728901-31728901 on Build GRCh38
rs1023146369 Chr.6: 31728584-31728584 on Build GRCh38
rs762297044 Chr.6: 31728770-31728770 on Build GRCh38
rs772425050 Chr.6: 31728955-31728955 on Build GRCh38
rs561348708 Chr.6: 31728836-31728836 on Build GRCh38
rs753958371 Chr.6: 31728851-31728851 on Build GRCh38
rs767628982 Chr.6: 31728782-31728782 on Build GRCh38
rs767307185 Chr.6: 31729027-31729027 on Build GRCh38
rs769286637 Chr.6: 31728725-31728725 on Build GRCh38
rs200500263 Chr.6: 31728716-31728716 on Build GRCh38
rs922056221 Chr.6: 31728610-31728610 on Build GRCh38
rs757006320 Chr.6: 31728876-31728876 on Build GRCh38
rs188974620 Chr.6: 31728637-31728637 on Build GRCh38
rs113769425 Chr.6: 31728617-31728617 on Build GRCh38
rs769638520 Chr.6: 31728688-31728690 on Build GRCh38
rs576302601 Chr.6: 31728868-31728868 on Build GRCh38
rs780252044 Chr.6: 31728737-31728777 on Build GRCh38
rs769330680 Chr.6: 31728757-31728757 on Build GRCh38
rs750457637 Chr.6: 31728686-31728686 on Build GRCh38
rs1009304954 Chr.6: 31728993-31728993 on Build GRCh38
rs190717045 Chr.6: 31728753-31728753 on Build GRCh38
rs776679194 Chr.6: 31728977-31728977 on Build GRCh38
rs774837244 Chr.6: 31728764-31728764 on Build GRCh38
rs775981641 Chr.6: 31728635-31728635 on Build GRCh38
rs970271252 Chr.6: 31728585-31728585 on Build GRCh38
rs368529782 Chr.6: 31728882-31728882 on Build GRCh38
rs371472036 Chr.6: 31728943-31728943 on Build GRCh38
rs780902159 Chr.6: 31728629-31728630 on Build GRCh38
rs747935166 Chr.6: 31728600-31728600 on Build GRCh38
rs1044656908 Chr.6: 31728856-31728856 on Build GRCh38
rs114820660 Chr.6: 31728590-31728590 on Build GRCh38
rs367611448 Chr.6: 31728627-31728627 on Build GRCh38
rs760918705 Chr.6: 31728966-31728966 on Build GRCh38
rs757162385 Chr.6: 31729026-31729026 on Build GRCh38
rs745679219 Chr.6: 31728631-31728632 on Build GRCh38
rs1006155349 Chr.6: 31728842-31728842 on Build GRCh38
rs186322662 Chr.6: 31728737-31728737 on Build GRCh38
rs542613838 Chr.6: 31728784-31728784 on Build GRCh38
rs368696409 Chr.6: 31728947-31728947 on Build GRCh38
rs745508363 Chr.6: 31728720-31728720 on Build GRCh38
rs1047750710 Chr.6: 31728781-31728781 on Build GRCh38
rs375654675 Chr.6: 31728860-31728860 on Build GRCh38
rs752574412 Chr.6: 31728699-31728699 on Build GRCh38
rs371970217 Chr.6: 31728903-31728903 on Build GRCh38
rs759674784 Chr.6: 31728995-31728995 on Build GRCh38
rs749775839 Chr.6: 31728755-31728755 on Build GRCh38
rs758309508 Chr.6: 31728852-31728852 on Build GRCh38
rs1045457396 Chr.6: 31728641-31728641 on Build GRCh38
rs762100335 Chr.6: 31728659-31728659 on Build GRCh38

View Full Product Details