Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31969424-31969926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DXO
Entrez Gene ID
1797
Gene Name
decapping exoribonuclease
Gene Aliases
DOM3L, DOM3Z, NG6, RAI1
UniGene
Hs.153299
Gene Chromosome Location
Chr.6: 31969811-31972290 on Build GRCh38
Transcripts
NM_005510.3 XM_006715005.3 XM_017010329.1 NM_006929.4

Gene Symbol
SKIV2L
Entrez Gene ID
6499
Gene Name
Ski2 like RNA helicase
Gene Aliases
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, SKIV2L1, THES2
UniGene
Hs.89864
Gene Chromosome Location
Chr.6: 31958804-31969852 on Build GRCh38
Transcripts
NM_005510.3 XM_006715005.3 XM_017010329.1 NM_006929.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
143528 c.1169_1170CC>GT p.P390R Chr.6: 31969898-31969899 on Build GRCh38
5870247 c.3706G>A p.D1236N Chr.6: 31969680-31969680 on Build GRCh38
7142260 c.3622C>A p.R1208S Chr.6: 31969596-31969596 on Build GRCh38
1621461 c.3695T>G p.L1232W Chr.6: 31969669-31969669 on Build GRCh38
4993937 c.1174A>G p.T392A Chr.6: 31969894-31969894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866655612 Chr.6: 31969774-31969774 on Build GRCh38
rs932589204 Chr.6: 31969810-31969810 on Build GRCh38
rs771278734 Chr.6: 31969605-31969606 on Build GRCh38
rs748569958 Chr.6: 31969661-31969661 on Build GRCh38
rs144418335 Chr.6: 31969596-31969596 on Build GRCh38
rs766514567 Chr.6: 31969578-31969578 on Build GRCh38
rs750120175 Chr.6: 31969605-31969605 on Build GRCh38
rs990088341 Chr.6: 31969754-31969754 on Build GRCh38
rs766101164 Chr.6: 31969899-31969899 on Build GRCh38
rs780914425 Chr.6: 31969603-31969603 on Build GRCh38
rs372370078 Chr.6: 31969862-31969862 on Build GRCh38
rs764060843 Chr.6: 31969692-31969692 on Build GRCh38
rs765938677 Chr.6: 31969724-31969724 on Build GRCh38
rs776687798 Chr.6: 31969859-31969859 on Build GRCh38
rs769349127 Chr.6: 31969646-31969646 on Build GRCh38
rs1054314263 Chr.6: 31969806-31969806 on Build GRCh38
rs747322612 Chr.6: 31969849-31969849 on Build GRCh38
rs990053058 Chr.6: 31969452-31969452 on Build GRCh38
rs760762616 Chr.6: 31969567-31969567 on Build GRCh38
rs780818507 Chr.6: 31969486-31969486 on Build GRCh38
rs41267110 Chr.6: 31969809-31969809 on Build GRCh38
rs773464401 Chr.6: 31969566-31969566 on Build GRCh38
rs767302498 Chr.6: 31969696-31969696 on Build GRCh38
rs761696355 Chr.6: 31969694-31969694 on Build GRCh38
rs755655461 Chr.6: 31969617-31969617 on Build GRCh38
rs762751779 Chr.6: 31969513-31969513 on Build GRCh38
rs749805656 Chr.6: 31969515-31969515 on Build GRCh38
rs530304291 Chr.6: 31969813-31969813 on Build GRCh38
rs758232874 Chr.6: 31969476-31969476 on Build GRCh38
rs369082949 Chr.6: 31969828-31969828 on Build GRCh38
rs142860816 Chr.6: 31969640-31969640 on Build GRCh38
rs531570604 Chr.6: 31969660-31969660 on Build GRCh38
rs771429107 Chr.6: 31969841-31969842 on Build GRCh38
rs566544171 Chr.6: 31969541-31969541 on Build GRCh38
rs41267106 Chr.6: 31969775-31969775 on Build GRCh38
rs549307349 Chr.6: 31969825-31969825 on Build GRCh38
rs754512168 Chr.6: 31969728-31969728 on Build GRCh38
rs753425980 Chr.6: 31969727-31969727 on Build GRCh38
rs924456674 Chr.6: 31969761-31969761 on Build GRCh38
rs752706408 Chr.6: 31969465-31969465 on Build GRCh38
rs773122407 Chr.6: 31969897-31969897 on Build GRCh38
rs2746400 Chr.6: 31969687-31969687 on Build GRCh38
rs553455420 Chr.6: 31969898-31969898 on Build GRCh38
rs372110283 Chr.6: 31969589-31969589 on Build GRCh38
rs769228093 Chr.6: 31969530-31969530 on Build GRCh38
rs368453711 Chr.6: 31969579-31969579 on Build GRCh38
rs3180876 Chr.6: 31969816-31969816 on Build GRCh38
rs945846577 Chr.6: 31969523-31969523 on Build GRCh38
rs755854159 Chr.6: 31969709-31969709 on Build GRCh38
rs774251531 Chr.6: 31969693-31969693 on Build GRCh38
rs769366379 Chr.6: 31969508-31969508 on Build GRCh38
rs913096735 Chr.6: 31969500-31969500 on Build GRCh38
rs1015744027 Chr.6: 31969621-31969621 on Build GRCh38
rs748698977 Chr.6: 31969826-31969826 on Build GRCh38
rs772461288 Chr.6: 31969563-31969563 on Build GRCh38
rs751297738 Chr.6: 31969480-31969480 on Build GRCh38
rs555838384 Chr.6: 31969860-31969860 on Build GRCh38
rs779628909 Chr.6: 31969635-31969635 on Build GRCh38
rs773412607 Chr.6: 31969674-31969674 on Build GRCh38
rs921956286 Chr.6: 31969895-31969895 on Build GRCh38
rs900478464 Chr.6: 31969637-31969637 on Build GRCh38
rs759479779 Chr.6: 31969733-31969735 on Build GRCh38
rs778218081 Chr.6: 31969843-31969843 on Build GRCh38
rs3178785 Chr.6: 31969814-31969814 on Build GRCh38
rs571209078 Chr.6: 31969876-31969876 on Build GRCh38
rs745412456 Chr.6: 31969490-31969490 on Build GRCh38
rs192273589 Chr.6: 31969900-31969900 on Build GRCh38
rs375183559 Chr.6: 31969597-31969597 on Build GRCh38
rs543924059 Chr.6: 31969678-31969678 on Build GRCh38
rs150954621 Chr.6: 31969879-31969879 on Build GRCh38
rs752684336 Chr.6: 31969847-31969847 on Build GRCh38
rs777553799 Chr.6: 31969478-31969478 on Build GRCh38
rs756962606 Chr.6: 31969485-31969485 on Build GRCh38
rs2075702 Chr.6: 31969734-31969734 on Build GRCh38
rs565424838 Chr.6: 31969685-31969685 on Build GRCh38
rs368151732 Chr.6: 31969451-31969451 on Build GRCh38
rs146598176 Chr.6: 31969618-31969618 on Build GRCh38
rs774910596 Chr.6: 31969538-31969538 on Build GRCh38
rs1027850789 Chr.6: 31969657-31969657 on Build GRCh38
rs890871895 Chr.6: 31969606-31969606 on Build GRCh38
rs112517904 Chr.6: 31969791-31969791 on Build GRCh38
rs368503645 Chr.6: 31969877-31969877 on Build GRCh38
rs750126726 Chr.6: 31969706-31969706 on Build GRCh38
rs772589665 Chr.6: 31969666-31969666 on Build GRCh38
rs142555775 Chr.6: 31969853-31969853 on Build GRCh38
rs777054757 Chr.6: 31969728-31969735 on Build GRCh38
rs142540780 Chr.6: 31969675-31969675 on Build GRCh38

View Full Product Details