Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34248649-34249151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
UniGene
Hs.381300
Gene Chromosome Location
Chr.6: 34246380-34249108 on Build GRCh38
Transcripts
NM_001008703.2 NM_001008704.2 NM_001287396.1 NM_001287397.1 NM_001287398.1 NM_178508.4

SNP Detail

SNP ID Chromosome Location
rs1042782813 Chr.6: 34248704-34248704 on Build GRCh38
rs1011123764 Chr.6: 34248786-34248786 on Build GRCh38
rs916279455 Chr.6: 34248960-34248960 on Build GRCh38
rs972101755 Chr.6: 34249129-34249129 on Build GRCh38
rs553300701 Chr.6: 34248790-34248790 on Build GRCh38
rs938255067 Chr.6: 34248928-34248928 on Build GRCh38
rs893953986 Chr.6: 34249024-34249024 on Build GRCh38
rs945458841 Chr.6: 34248972-34248972 on Build GRCh38
rs370916132 Chr.6: 34248789-34248789 on Build GRCh38
rs931250638 Chr.6: 34248911-34248911 on Build GRCh38
rs55835960 Chr.6: 34248730-34248730 on Build GRCh38
rs1029644128 Chr.6: 34249030-34249030 on Build GRCh38
rs1052608042 Chr.6: 34249013-34249013 on Build GRCh38
rs758186877 Chr.6: 34248682-34248682 on Build GRCh38
rs534684747 Chr.6: 34248738-34248738 on Build GRCh38
rs1048762661 Chr.6: 34248954-34248954 on Build GRCh38
rs983183632 Chr.6: 34248827-34248827 on Build GRCh38
rs950547464 Chr.6: 34249091-34249091 on Build GRCh38
rs1005117454 Chr.6: 34249094-34249094 on Build GRCh38
rs953473280 Chr.6: 34248833-34248833 on Build GRCh38
rs375825870 Chr.6: 34249037-34249037 on Build GRCh38
rs773570666 Chr.6: 34248774-34248774 on Build GRCh38
rs1011174572 Chr.6: 34249025-34249025 on Build GRCh38
rs11544122 Chr.6: 34248815-34248815 on Build GRCh38
rs947695075 Chr.6: 34248966-34248966 on Build GRCh38
rs906275327 Chr.6: 34248971-34248971 on Build GRCh38
rs191427530 Chr.6: 34248893-34248893 on Build GRCh38
rs967244854 Chr.6: 34248977-34248977 on Build GRCh38
rs935034703 Chr.6: 34249010-34249010 on Build GRCh38
rs535902557 Chr.6: 34248819-34248819 on Build GRCh38
rs1030135532 Chr.6: 34248813-34248813 on Build GRCh38
rs1031293047 Chr.6: 34248708-34248708 on Build GRCh38
rs1019061076 Chr.6: 34249095-34249095 on Build GRCh38
rs964604846 Chr.6: 34248751-34248751 on Build GRCh38
rs903816617 Chr.6: 34248709-34248709 on Build GRCh38
rs575428956 Chr.6: 34248991-34248991 on Build GRCh38
rs889144860 Chr.6: 34249031-34249031 on Build GRCh38
rs748383080 Chr.6: 34248859-34248859 on Build GRCh38
rs1046186539 Chr.6: 34248970-34248970 on Build GRCh38
rs1056542161 Chr.6: 34248677-34248677 on Build GRCh38
rs911944152 Chr.6: 34248865-34248865 on Build GRCh38
rs918383035 Chr.6: 34248897-34248897 on Build GRCh38
rs759194834 Chr.6: 34248804-34248804 on Build GRCh38
rs972530737 Chr.6: 34248894-34248894 on Build GRCh38
rs962108229 Chr.6: 34249111-34249111 on Build GRCh38
rs796579553 Chr.6: 34248696-34248696 on Build GRCh38
rs1030581749 Chr.6: 34249064-34249064 on Build GRCh38
rs570891590 Chr.6: 34248726-34248726 on Build GRCh38
rs997409255 Chr.6: 34249047-34249047 on Build GRCh38
rs1017570412 Chr.6: 34248737-34248737 on Build GRCh38
rs903931010 Chr.6: 34248997-34248997 on Build GRCh38
rs1009626915 Chr.6: 34248697-34248697 on Build GRCh38

View Full Product Details