Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150331291-150331794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC61
Entrez Gene ID
65999
Gene Name
leucine rich repeat containing 61
Gene Aliases
HSPC295
UniGene
Hs.647119
Gene Chromosome Location
Chr.7: 150322639-150338156 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Gene Symbol
ZBED6CL
Entrez Gene ID
113763
Gene Name
ZBED6 C-terminal like
Gene Aliases
C7orf29
UniGene
Hs.655915
Gene Chromosome Location
Chr.7: 150329849-150332721 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

SNP Detail

SNP ID Chromosome Location
rs943798224 Chr.7: 150331553-150331553 on Build GRCh38
rs114973325 Chr.7: 150331575-150331575 on Build GRCh38
rs960821217 Chr.7: 150331737-150331737 on Build GRCh38
rs35957016 Chr.7: 150331410-150331411 on Build GRCh38
rs992501702 Chr.7: 150331451-150331451 on Build GRCh38
rs531150061 Chr.7: 150331733-150331733 on Build GRCh38
rs1014945380 Chr.7: 150331687-150331687 on Build GRCh38
rs1045488704 Chr.7: 150331663-150331663 on Build GRCh38
rs935483109 Chr.7: 150331633-150331633 on Build GRCh38
rs746095840 Chr.7: 150331537-150331537 on Build GRCh38
rs550928657 Chr.7: 150331771-150331771 on Build GRCh38
rs1026857165 Chr.7: 150331481-150331481 on Build GRCh38
rs890261998 Chr.7: 150331521-150331521 on Build GRCh38
rs748213500 Chr.7: 150331585-150331585 on Build GRCh38
rs972650549 Chr.7: 150331479-150331479 on Build GRCh38
rs960894113 Chr.7: 150331423-150331423 on Build GRCh38
rs543002012 Chr.7: 150331625-150331625 on Build GRCh38
rs1035192770 Chr.7: 150331356-150331357 on Build GRCh38
rs948288925 Chr.7: 150331656-150331656 on Build GRCh38
rs1038475686 Chr.7: 150331332-150331332 on Build GRCh38
rs940335651 Chr.7: 150331486-150331487 on Build GRCh38
rs772154244 Chr.7: 150331601-150331601 on Build GRCh38
rs74334311 Chr.7: 150331639-150331639 on Build GRCh38
rs985538358 Chr.7: 150331503-150331503 on Build GRCh38
rs924128841 Chr.7: 150331613-150331613 on Build GRCh38
rs185586313 Chr.7: 150331676-150331676 on Build GRCh38
rs982929079 Chr.7: 150331744-150331744 on Build GRCh38
rs981454354 Chr.7: 150331345-150331345 on Build GRCh38
rs57013052 Chr.7: 150331574-150331574 on Build GRCh38
rs952447054 Chr.7: 150331486-150331486 on Build GRCh38
rs931917631 Chr.7: 150331505-150331505 on Build GRCh38
rs568579185 Chr.7: 150331584-150331585 on Build GRCh38
rs3800783 Chr.7: 150331453-150331453 on Build GRCh38
rs181268374 Chr.7: 150331641-150331641 on Build GRCh38
rs897035194 Chr.7: 150331731-150331731 on Build GRCh38
rs952742158 Chr.7: 150331764-150331764 on Build GRCh38
rs1003557659 Chr.7: 150331412-150331412 on Build GRCh38
rs1050808653 Chr.7: 150331507-150331507 on Build GRCh38
rs944527041 Chr.7: 150331535-150331535 on Build GRCh38
rs565107272 Chr.7: 150331677-150331677 on Build GRCh38
rs1054021339 Chr.7: 150331339-150331339 on Build GRCh38
rs1002451949 Chr.7: 150331730-150331730 on Build GRCh38
rs544276016 Chr.7: 150331396-150331396 on Build GRCh38
rs993954738 Chr.7: 150331741-150331741 on Build GRCh38
rs969654481 Chr.7: 150331330-150331330 on Build GRCh38
rs1022941086 Chr.7: 150331329-150331329 on Build GRCh38
rs1026787178 Chr.7: 150331752-150331752 on Build GRCh38
rs201645766 Chr.7: 150331326-150331326 on Build GRCh38
rs1012652154 Chr.7: 150331361-150331361 on Build GRCh38
rs1052497559 Chr.7: 150331554-150331554 on Build GRCh38
rs1023600636 Chr.7: 150331383-150331383 on Build GRCh38
rs893845736 Chr.7: 150331340-150331340 on Build GRCh38
rs1035503999 Chr.7: 150331439-150331439 on Build GRCh38
rs11978222 Chr.7: 150331311-150331311 on Build GRCh38

View Full Product Details