Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150519850-150520347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP7
Entrez Gene ID
168537
Gene Name
GTPase, IMAP family member 7
Gene Aliases
IAN7, hIAN7
UniGene
Hs.647074
Gene Chromosome Location
Chr.7: 150514857-150521073 on Build GRCh38
Transcripts
NM_153236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3635971 c.94G>A p.E32K Chr.7: 150520068-150520068 on Build GRCh38
5578629 c.276C>T p.P92P Chr.7: 150520250-150520250 on Build GRCh38
4386623 c.166G>T p.G56W Chr.7: 150520140-150520140 on Build GRCh38
6671018 c.213G>T p.K71N Chr.7: 150520187-150520187 on Build GRCh38
1208182 c.268C>A p.P90T Chr.7: 150520242-150520242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374792037 Chr.7: 150520293-150520293 on Build GRCh38
rs959673940 Chr.7: 150520162-150520162 on Build GRCh38
rs7800829 Chr.7: 150520001-150520001 on Build GRCh38
rs781236062 Chr.7: 150520007-150520008 on Build GRCh38
rs757278229 Chr.7: 150519974-150519975 on Build GRCh38
rs201064674 Chr.7: 150520048-150520048 on Build GRCh38
rs766704147 Chr.7: 150520027-150520027 on Build GRCh38
rs3735081 Chr.7: 150520160-150520160 on Build GRCh38
rs768173111 Chr.7: 150520149-150520151 on Build GRCh38
rs914295886 Chr.7: 150519919-150519919 on Build GRCh38
rs115726102 Chr.7: 150520281-150520281 on Build GRCh38
rs749566768 Chr.7: 150520003-150520003 on Build GRCh38
rs145339506 Chr.7: 150519994-150519994 on Build GRCh38
rs774971719 Chr.7: 150519936-150519936 on Build GRCh38
rs200412468 Chr.7: 150520250-150520250 on Build GRCh38
rs755914150 Chr.7: 150520034-150520035 on Build GRCh38
rs114367962 Chr.7: 150520062-150520062 on Build GRCh38
rs748916089 Chr.7: 150520313-150520313 on Build GRCh38
rs779265360 Chr.7: 150520190-150520190 on Build GRCh38
rs543994152 Chr.7: 150519911-150519911 on Build GRCh38
rs772643551 Chr.7: 150520124-150520124 on Build GRCh38
rs1010189661 Chr.7: 150520319-150520319 on Build GRCh38
rs956011145 Chr.7: 150520088-150520088 on Build GRCh38
rs758475358 Chr.7: 150520193-150520193 on Build GRCh38
rs749012072 Chr.7: 150520132-150520133 on Build GRCh38
rs574187935 Chr.7: 150519930-150519930 on Build GRCh38
rs368679926 Chr.7: 150520230-150520230 on Build GRCh38
rs114459899 Chr.7: 150520049-150520049 on Build GRCh38
rs142322540 Chr.7: 150520024-150520024 on Build GRCh38
rs148683928 Chr.7: 150519943-150519943 on Build GRCh38
rs772641323 Chr.7: 150520012-150520012 on Build GRCh38
rs776236952 Chr.7: 150520029-150520029 on Build GRCh38
rs267601406 Chr.7: 150520234-150520234 on Build GRCh38
rs147029703 Chr.7: 150520111-150520111 on Build GRCh38
rs138301747 Chr.7: 150520127-150520127 on Build GRCh38
rs142236871 Chr.7: 150520007-150520007 on Build GRCh38
rs549466095 Chr.7: 150520128-150520128 on Build GRCh38
rs772616708 Chr.7: 150520280-150520280 on Build GRCh38
rs781469566 Chr.7: 150520220-150520220 on Build GRCh38
rs536471579 Chr.7: 150520083-150520083 on Build GRCh38
rs867255139 Chr.7: 150519979-150519979 on Build GRCh38
rs762549629 Chr.7: 150520242-150520242 on Build GRCh38
rs768201444 Chr.7: 150520316-150520316 on Build GRCh38
rs752588717 Chr.7: 150519980-150519980 on Build GRCh38
rs138568397 Chr.7: 150520308-150520308 on Build GRCh38
rs770236213 Chr.7: 150519970-150519970 on Build GRCh38
rs900606701 Chr.7: 150520319-150520326 on Build GRCh38
rs750252314 Chr.7: 150520173-150520173 on Build GRCh38
rs529928390 Chr.7: 150519992-150519992 on Build GRCh38
rs570426719 Chr.7: 150520169-150520170 on Build GRCh38
rs1030270223 Chr.7: 150520133-150520133 on Build GRCh38
rs773934835 Chr.7: 150520232-150520234 on Build GRCh38
rs767729101 Chr.7: 150519942-150519942 on Build GRCh38
rs1030224963 Chr.7: 150520064-150520064 on Build GRCh38
rs781166918 Chr.7: 150520057-150520057 on Build GRCh38
rs762443766 Chr.7: 150519940-150519940 on Build GRCh38
rs374256990 Chr.7: 150519968-150519968 on Build GRCh38
rs942866533 Chr.7: 150520255-150520255 on Build GRCh38
rs760524047 Chr.7: 150520260-150520260 on Build GRCh38
rs199907558 Chr.7: 150520016-150520016 on Build GRCh38
rs759130851 Chr.7: 150520139-150520139 on Build GRCh38
rs930266071 Chr.7: 150520240-150520240 on Build GRCh38
rs559144497 Chr.7: 150519985-150519985 on Build GRCh38
rs770198027 Chr.7: 150520222-150520222 on Build GRCh38
rs530752120 Chr.7: 150520037-150520037 on Build GRCh38
rs778322751 Chr.7: 150520123-150520123 on Build GRCh38
rs761747672 Chr.7: 150520253-150520253 on Build GRCh38
rs768911658 Chr.7: 150519933-150519933 on Build GRCh38
rs764413246 Chr.7: 150520035-150520035 on Build GRCh38
rs367811003 Chr.7: 150520129-150520129 on Build GRCh38
rs149518773 Chr.7: 150520263-150520263 on Build GRCh38
rs764766169 Chr.7: 150519978-150519978 on Build GRCh38
rs781652739 Chr.7: 150520295-150520295 on Build GRCh38
rs775183470 Chr.7: 150520235-150520235 on Build GRCh38
rs751595609 Chr.7: 150520284-150520284 on Build GRCh38
rs559037929 Chr.7: 150519959-150519959 on Build GRCh38
rs773341058 Chr.7: 150520257-150520257 on Build GRCh38
rs144054418 Chr.7: 150520307-150520307 on Build GRCh38
rs752912337 Chr.7: 150520192-150520192 on Build GRCh38
rs746202631 Chr.7: 150519969-150519969 on Build GRCh38
rs748255904 Chr.7: 150520091-150520091 on Build GRCh38
rs3735079 Chr.7: 150520247-150520247 on Build GRCh38
rs751507271 Chr.7: 150520053-150520053 on Build GRCh38
rs372010180 Chr.7: 150520039-150520039 on Build GRCh38
rs867606711 Chr.7: 150520125-150520125 on Build GRCh38
rs887628601 Chr.7: 150520274-150520274 on Build GRCh38
rs191315835 Chr.7: 150519915-150519915 on Build GRCh38
rs758570629 Chr.7: 150520279-150520279 on Build GRCh38
rs976379674 Chr.7: 150519905-150519905 on Build GRCh38
rs761682617 Chr.7: 150520167-150520167 on Build GRCh38
rs747499661 Chr.7: 150520217-150520217 on Build GRCh38
rs754463949 Chr.7: 150519977-150519977 on Build GRCh38
rs751480260 Chr.7: 150520166-150520166 on Build GRCh38
rs748612836 Chr.7: 150520008-150520008 on Build GRCh38
rs768819463 Chr.7: 150520004-150520004 on Build GRCh38
rs755269572 Chr.7: 150520067-150520067 on Build GRCh38
rs759216663 Chr.7: 150520032-150520032 on Build GRCh38
rs938356459 Chr.7: 150519957-150519957 on Build GRCh38
rs3735080 Chr.7: 150520221-150520221 on Build GRCh38
rs769439572 Chr.7: 150520141-150520141 on Build GRCh38
rs116726339 Chr.7: 150520155-150520155 on Build GRCh38
rs1052462119 Chr.7: 150520296-150520296 on Build GRCh38
rs1026187186 Chr.7: 150520047-150520047 on Build GRCh38
rs756043172 Chr.7: 150520185-150520185 on Build GRCh38
rs762507656 Chr.7: 150520156-150520156 on Build GRCh38
rs757051269 Chr.7: 150520054-150520054 on Build GRCh38
rs368210582 Chr.7: 150519993-150519993 on Build GRCh38
rs745662426 Chr.7: 150520010-150520011 on Build GRCh38
rs545570989 Chr.7: 150520214-150520214 on Build GRCh38

View Full Product Details