Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151053720-151054224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164190 c.803A>G p.K268R Chr.7: 151054085-151054085 on Build GRCh38
6279358 c.792+10C>A p.? Chr.7: 151054202-151054202 on Build GRCh38
7270429 c.821G>A p.R274H Chr.7: 151054067-151054067 on Build GRCh38
4511404 c.867C>T p.F289F Chr.7: 151054021-151054021 on Build GRCh38
5997316 c.861C>T p.S287S Chr.7: 151054027-151054027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925205487 Chr.7: 151053999-151053999 on Build GRCh38
rs889621925 Chr.7: 151053802-151053802 on Build GRCh38
rs565598389 Chr.7: 151054129-151054129 on Build GRCh38
rs371735422 Chr.7: 151054003-151054003 on Build GRCh38
rs1018272311 Chr.7: 151053941-151053941 on Build GRCh38
rs368178302 Chr.7: 151053995-151053995 on Build GRCh38
rs201424831 Chr.7: 151054192-151054192 on Build GRCh38
rs773329997 Chr.7: 151054015-151054015 on Build GRCh38
rs913272774 Chr.7: 151053772-151053772 on Build GRCh38
rs893139065 Chr.7: 151054197-151054197 on Build GRCh38
rs1054854907 Chr.7: 151053885-151053885 on Build GRCh38
rs894906606 Chr.7: 151053890-151053890 on Build GRCh38
rs766831959 Chr.7: 151054016-151054016 on Build GRCh38
rs529902552 Chr.7: 151053828-151053828 on Build GRCh38
rs2069461 Chr.7: 151053894-151053894 on Build GRCh38
rs755859799 Chr.7: 151054117-151054117 on Build GRCh38
rs777405337 Chr.7: 151053962-151053962 on Build GRCh38
rs902453827 Chr.7: 151054090-151054090 on Build GRCh38
rs773454607 Chr.7: 151054172-151054172 on Build GRCh38
rs945304658 Chr.7: 151053781-151053781 on Build GRCh38
rs879845113 Chr.7: 151053893-151053894 on Build GRCh38
rs758106701 Chr.7: 151054049-151054049 on Build GRCh38
rs775198732 Chr.7: 151053959-151053959 on Build GRCh38
rs746908389 Chr.7: 151053980-151053980 on Build GRCh38
rs767784515 Chr.7: 151053788-151053788 on Build GRCh38
rs79627316 Chr.7: 151053994-151053994 on Build GRCh38
rs35051446 Chr.7: 151053923-151053923 on Build GRCh38
rs961049678 Chr.7: 151053960-151053960 on Build GRCh38
rs747025084 Chr.7: 151054177-151054177 on Build GRCh38
rs748297614 Chr.7: 151054131-151054131 on Build GRCh38
rs772015673 Chr.7: 151054012-151054012 on Build GRCh38
rs953022516 Chr.7: 151053739-151053739 on Build GRCh38
rs777125495 Chr.7: 151054026-151054026 on Build GRCh38
rs982108549 Chr.7: 151053924-151053924 on Build GRCh38
rs867496203 Chr.7: 151054073-151054073 on Build GRCh38
rs553172337 Chr.7: 151054037-151054037 on Build GRCh38
rs751078477 Chr.7: 151054061-151054061 on Build GRCh38
rs1027674304 Chr.7: 151053922-151053922 on Build GRCh38
rs2069460 Chr.7: 151054004-151054004 on Build GRCh38
rs1048605499 Chr.7: 151054160-151054160 on Build GRCh38
rs202114425 Chr.7: 151054054-151054054 on Build GRCh38
rs750832633 Chr.7: 151054185-151054185 on Build GRCh38
rs772289977 Chr.7: 151054142-151054142 on Build GRCh38
rs200333664 Chr.7: 151054046-151054046 on Build GRCh38
rs1041000556 Chr.7: 151054065-151054065 on Build GRCh38
rs768071664 Chr.7: 151054165-151054165 on Build GRCh38
rs753001382 Chr.7: 151053859-151053859 on Build GRCh38
rs9278 Chr.7: 151053893-151053893 on Build GRCh38
rs976721125 Chr.7: 151053757-151053757 on Build GRCh38
rs985546087 Chr.7: 151053742-151053742 on Build GRCh38
rs986038278 Chr.7: 151053939-151053939 on Build GRCh38
rs751476293 Chr.7: 151054170-151054170 on Build GRCh38
rs994259674 Chr.7: 151054153-151054153 on Build GRCh38
rs781221793 Chr.7: 151054068-151054068 on Build GRCh38
rs1019491833 Chr.7: 151053812-151053812 on Build GRCh38
rs1023595233 Chr.7: 151053903-151053903 on Build GRCh38
rs765484613 Chr.7: 151054199-151054199 on Build GRCh38
rs1008563319 Chr.7: 151053810-151053810 on Build GRCh38
rs1042726304 Chr.7: 151053799-151053799 on Build GRCh38
rs548208867 Chr.7: 151054067-151054067 on Build GRCh38
rs116863165 Chr.7: 151053891-151053891 on Build GRCh38
rs745860943 Chr.7: 151054082-151054082 on Build GRCh38
rs575163799 Chr.7: 151053916-151053916 on Build GRCh38
rs1050054876 Chr.7: 151053743-151053743 on Build GRCh38
rs144982250 Chr.7: 151053874-151053874 on Build GRCh38
rs554235263 Chr.7: 151053933-151053933 on Build GRCh38
rs755148813 Chr.7: 151053784-151053784 on Build GRCh38
rs777107002 Chr.7: 151054194-151054194 on Build GRCh38
rs779941691 Chr.7: 151054118-151054118 on Build GRCh38
rs11541602 Chr.7: 151054045-151054045 on Build GRCh38
rs763349691 Chr.7: 151054123-151054123 on Build GRCh38
rs763946310 Chr.7: 151054027-151054027 on Build GRCh38
rs936519525 Chr.7: 151053863-151053863 on Build GRCh38
rs566748363 Chr.7: 151053888-151053888 on Build GRCh38

View Full Product Details