Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151054300-151054792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625522 c.699G>A p.L233L Chr.7: 151054417-151054417 on Build GRCh38
3745624 c.711+9G>T p.? Chr.7: 151054396-151054396 on Build GRCh38
4635486 c.680G>T p.W227L Chr.7: 151054436-151054436 on Build GRCh38
3879362 c.662C>T p.T221M Chr.7: 151054454-151054454 on Build GRCh38
3381860 c.669C>T p.T223T Chr.7: 151054447-151054447 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554147188 Chr.7: 151054599-151054599 on Build GRCh38
rs749880275 Chr.7: 151054428-151054428 on Build GRCh38
rs1000324881 Chr.7: 151054558-151054558 on Build GRCh38
rs868305943 Chr.7: 151054346-151054346 on Build GRCh38
rs546786609 Chr.7: 151054394-151054394 on Build GRCh38
rs369291948 Chr.7: 151054447-151054447 on Build GRCh38
rs374928837 Chr.7: 151054433-151054433 on Build GRCh38
rs753731719 Chr.7: 151054361-151054361 on Build GRCh38
rs757027718 Chr.7: 151054405-151054405 on Build GRCh38
rs1039903067 Chr.7: 151054689-151054689 on Build GRCh38
rs943765356 Chr.7: 151054472-151054472 on Build GRCh38
rs764992346 Chr.7: 151054475-151054475 on Build GRCh38
rs76486938 Chr.7: 151054474-151054474 on Build GRCh38
rs190949721 Chr.7: 151054345-151054345 on Build GRCh38
rs780601142 Chr.7: 151054420-151054420 on Build GRCh38
rs903385123 Chr.7: 151054752-151054752 on Build GRCh38
rs1002901213 Chr.7: 151054596-151054596 on Build GRCh38
rs981942419 Chr.7: 151054635-151054635 on Build GRCh38
rs372757682 Chr.7: 151054492-151054492 on Build GRCh38
rs1011549480 Chr.7: 151054546-151054546 on Build GRCh38
rs2069459 Chr.7: 151054503-151054503 on Build GRCh38
rs111391436 Chr.7: 151054652-151054652 on Build GRCh38
rs758217657 Chr.7: 151054498-151054498 on Build GRCh38
rs533656607 Chr.7: 151054664-151054664 on Build GRCh38
rs537402504 Chr.7: 151054552-151054552 on Build GRCh38
rs566658552 Chr.7: 151054397-151054397 on Build GRCh38
rs760134185 Chr.7: 151054324-151054324 on Build GRCh38
rs965190297 Chr.7: 151054714-151054714 on Build GRCh38
rs905972434 Chr.7: 151054570-151054570 on Build GRCh38
rs369566121 Chr.7: 151054668-151054668 on Build GRCh38
rs766272981 Chr.7: 151054357-151054357 on Build GRCh38
rs753809198 Chr.7: 151054473-151054473 on Build GRCh38
rs756567765 Chr.7: 151054364-151054365 on Build GRCh38
rs935117781 Chr.7: 151054722-151054722 on Build GRCh38
rs970521882 Chr.7: 151054631-151054631 on Build GRCh38
rs985164624 Chr.7: 151054667-151054667 on Build GRCh38
rs967870619 Chr.7: 151054549-151054549 on Build GRCh38
rs983913041 Chr.7: 151054659-151054659 on Build GRCh38
rs1020292924 Chr.7: 151054539-151054539 on Build GRCh38
rs75745731 Chr.7: 151054699-151054699 on Build GRCh38
rs879368917 Chr.7: 151054538-151054538 on Build GRCh38
rs750673037 Chr.7: 151054511-151054511 on Build GRCh38
rs561324088 Chr.7: 151054344-151054344 on Build GRCh38
rs746590054 Chr.7: 151054569-151054569 on Build GRCh38
rs35186917 Chr.7: 151054441-151054441 on Build GRCh38
rs537002354 Chr.7: 151054598-151054598 on Build GRCh38
rs376602868 Chr.7: 151054525-151054525 on Build GRCh38
rs781288951 Chr.7: 151054390-151054390 on Build GRCh38
rs923760772 Chr.7: 151054484-151054484 on Build GRCh38
rs778744723 Chr.7: 151054362-151054362 on Build GRCh38
rs2069458 Chr.7: 151054637-151054637 on Build GRCh38
rs760173955 Chr.7: 151054453-151054453 on Build GRCh38
rs976495967 Chr.7: 151054715-151054715 on Build GRCh38
rs749383012 Chr.7: 151054638-151054638 on Build GRCh38
rs369223059 Chr.7: 151054454-151054454 on Build GRCh38
rs866868472 Chr.7: 151054686-151054686 on Build GRCh38
rs751747789 Chr.7: 151054368-151054368 on Build GRCh38
rs958764499 Chr.7: 151054589-151054589 on Build GRCh38
rs916521409 Chr.7: 151054630-151054630 on Build GRCh38
rs752437126 Chr.7: 151054477-151054477 on Build GRCh38
rs35414105 Chr.7: 151054541-151054542 on Build GRCh38
rs556423508 Chr.7: 151054651-151054651 on Build GRCh38
rs923772177 Chr.7: 151054718-151054718 on Build GRCh38
rs942387718 Chr.7: 151054666-151054666 on Build GRCh38
rs762292489 Chr.7: 151054446-151054446 on Build GRCh38
rs372091017 Chr.7: 151054681-151054681 on Build GRCh38
rs1033419234 Chr.7: 151054573-151054573 on Build GRCh38
rs553484800 Chr.7: 151054741-151054741 on Build GRCh38
rs972954427 Chr.7: 151054393-151054393 on Build GRCh38
rs757516342 Chr.7: 151054382-151054382 on Build GRCh38

View Full Product Details