Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151054942-151055449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5942109 c.484-3C>T p.? Chr.7: 151055376-151055376 on Build GRCh38
5919250 c.651+4C>T p.? Chr.7: 151055022-151055022 on Build GRCh38
5997722 c.503G>A p.R168H Chr.7: 151055354-151055354 on Build GRCh38
363441 c.500A>G p.Y167C Chr.7: 151055357-151055357 on Build GRCh38
452630 c.484G>T p.V162L Chr.7: 151055373-151055373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754137385 Chr.7: 151055106-151055109 on Build GRCh38
rs1024301055 Chr.7: 151055255-151055255 on Build GRCh38
rs772428305 Chr.7: 151055050-151055050 on Build GRCh38
rs745307324 Chr.7: 151055240-151055240 on Build GRCh38
rs753931050 Chr.7: 151055124-151055124 on Build GRCh38
rs769355512 Chr.7: 151055287-151055287 on Build GRCh38
rs200162750 Chr.7: 151055195-151055195 on Build GRCh38
rs765479272 Chr.7: 151055350-151055350 on Build GRCh38
rs775406194 Chr.7: 151055290-151055290 on Build GRCh38
rs891507 Chr.7: 151054979-151054979 on Build GRCh38
rs989337807 Chr.7: 151055047-151055047 on Build GRCh38
rs769267567 Chr.7: 151055411-151055411 on Build GRCh38
rs762042930 Chr.7: 151055080-151055080 on Build GRCh38
rs755114226 Chr.7: 151055127-151055127 on Build GRCh38
rs746649408 Chr.7: 151055227-151055227 on Build GRCh38
rs375581902 Chr.7: 151055233-151055233 on Build GRCh38
rs1030728605 Chr.7: 151054993-151054993 on Build GRCh38
rs759067345 Chr.7: 151055053-151055053 on Build GRCh38
rs376759171 Chr.7: 151055397-151055397 on Build GRCh38
rs888609039 Chr.7: 151055215-151055215 on Build GRCh38
rs927321505 Chr.7: 151055106-151055106 on Build GRCh38
rs754127997 Chr.7: 151055347-151055347 on Build GRCh38
rs143158445 Chr.7: 151055122-151055122 on Build GRCh38
rs1018908554 Chr.7: 151055230-151055230 on Build GRCh38
rs997827730 Chr.7: 151055267-151055267 on Build GRCh38
rs529906634 Chr.7: 151055384-151055384 on Build GRCh38
rs776051072 Chr.7: 151055029-151055029 on Build GRCh38
rs769584607 Chr.7: 151055219-151055219 on Build GRCh38
rs372172153 Chr.7: 151055151-151055151 on Build GRCh38
rs772336586 Chr.7: 151054994-151054994 on Build GRCh38
rs779590936 Chr.7: 151055419-151055419 on Build GRCh38
rs773970698 Chr.7: 151055308-151055308 on Build GRCh38
rs543967033 Chr.7: 151055384-151055385 on Build GRCh38
rs1057161459 Chr.7: 151055125-151055125 on Build GRCh38
rs748154347 Chr.7: 151055383-151055384 on Build GRCh38
rs786205164 Chr.7: 151055276-151055276 on Build GRCh38
rs749792320 Chr.7: 151054984-151054984 on Build GRCh38
rs778037739 Chr.7: 151055144-151055144 on Build GRCh38
rs1007145014 Chr.7: 151055223-151055223 on Build GRCh38
rs930047508 Chr.7: 151055180-151055180 on Build GRCh38
rs560617148 Chr.7: 151055152-151055152 on Build GRCh38
rs758784706 Chr.7: 151055138-151055138 on Build GRCh38
rs775219441 Chr.7: 151055313-151055313 on Build GRCh38
rs780040704 Chr.7: 151054980-151054980 on Build GRCh38
rs372279072 Chr.7: 151055384-151055384 on Build GRCh38
rs762727632 Chr.7: 151055067-151055067 on Build GRCh38
rs757641095 Chr.7: 151055389-151055389 on Build GRCh38
rs182527190 Chr.7: 151055013-151055013 on Build GRCh38
rs572198432 Chr.7: 151055303-151055303 on Build GRCh38
rs766622044 Chr.7: 151054985-151054986 on Build GRCh38
rs370428571 Chr.7: 151055101-151055101 on Build GRCh38
rs3214588 Chr.7: 151055110-151055111 on Build GRCh38
rs779505525 Chr.7: 151055128-151055128 on Build GRCh38
rs1013042813 Chr.7: 151055234-151055234 on Build GRCh38
rs753222804 Chr.7: 151055378-151055378 on Build GRCh38
rs984252869 Chr.7: 151054965-151054965 on Build GRCh38
rs759675039 Chr.7: 151055348-151055348 on Build GRCh38
rs894573544 Chr.7: 151055225-151055225 on Build GRCh38
rs779595379 Chr.7: 151054988-151054988 on Build GRCh38
rs368677466 Chr.7: 151055314-151055314 on Build GRCh38
rs3214589 Chr.7: 151055385-151055386 on Build GRCh38
rs376291427 Chr.7: 151055003-151055003 on Build GRCh38
rs763930713 Chr.7: 151055307-151055307 on Build GRCh38
rs773782162 Chr.7: 151055150-151055150 on Build GRCh38
rs780761259 Chr.7: 151055391-151055391 on Build GRCh38
rs778992179 Chr.7: 151055110-151055110 on Build GRCh38
rs907230295 Chr.7: 151055372-151055372 on Build GRCh38
rs373172008 Chr.7: 151054995-151054995 on Build GRCh38
rs553668929 Chr.7: 151055198-151055198 on Build GRCh38
rs776292430 Chr.7: 151055238-151055238 on Build GRCh38
rs376139702 Chr.7: 151055065-151055065 on Build GRCh38
rs368933497 Chr.7: 151055391-151055392 on Build GRCh38
rs778363443 Chr.7: 151055386-151055386 on Build GRCh38
rs745805861 Chr.7: 151055420-151055422 on Build GRCh38
rs375689583 Chr.7: 151055062-151055062 on Build GRCh38
rs763866365 Chr.7: 151055083-151055083 on Build GRCh38
rs762841652 Chr.7: 151055293-151055293 on Build GRCh38
rs748457769 Chr.7: 151054990-151054990 on Build GRCh38
rs756343351 Chr.7: 151055115-151055115 on Build GRCh38
rs751828643 Chr.7: 151055388-151055388 on Build GRCh38
rs368044523 Chr.7: 151055136-151055136 on Build GRCh38
rs770442459 Chr.7: 151055027-151055027 on Build GRCh38

View Full Product Details