Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151056657-151057163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7118544 c.94C>G p.L32V Chr.7: 151057104-151057104 on Build GRCh38
39941 c.46G>A p.G16R Chr.7: 151057152-151057152 on Build GRCh38
7100477 c.105G>A p.V35V Chr.7: 151057093-151057093 on Build GRCh38
3261437 c.168G>T p.K56N Chr.7: 151056934-151056934 on Build GRCh38
421538 c.91G>A p.A31T Chr.7: 151057107-151057107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762731440 Chr.7: 151056786-151056786 on Build GRCh38
rs760261892 Chr.7: 151057078-151057078 on Build GRCh38
rs375655625 Chr.7: 151056844-151056844 on Build GRCh38
rs1022629799 Chr.7: 151056727-151056727 on Build GRCh38
rs746425237 Chr.7: 151056887-151056887 on Build GRCh38
rs759883551 Chr.7: 151056766-151056766 on Build GRCh38
rs377383594 Chr.7: 151056954-151056954 on Build GRCh38
rs777878897 Chr.7: 151056738-151056738 on Build GRCh38
rs781167283 Chr.7: 151057019-151057019 on Build GRCh38
rs755933600 Chr.7: 151056866-151056866 on Build GRCh38
rs753588894 Chr.7: 151056981-151056981 on Build GRCh38
rs781027595 Chr.7: 151056699-151056699 on Build GRCh38
rs202081553 Chr.7: 151056724-151056724 on Build GRCh38
rs776043437 Chr.7: 151056890-151056890 on Build GRCh38
rs747044107 Chr.7: 151056880-151056880 on Build GRCh38
rs760579593 Chr.7: 151056961-151056961 on Build GRCh38
rs763865593 Chr.7: 151056829-151056829 on Build GRCh38
rs780714328 Chr.7: 151057032-151057032 on Build GRCh38
rs759312847 Chr.7: 151057121-151057121 on Build GRCh38
rs751783668 Chr.7: 151056996-151056996 on Build GRCh38
rs879019629 Chr.7: 151056812-151056812 on Build GRCh38
rs371590011 Chr.7: 151056713-151056713 on Build GRCh38
rs749616563 Chr.7: 151057034-151057034 on Build GRCh38
rs765078681 Chr.7: 151057123-151057123 on Build GRCh38
rs771591100 Chr.7: 151057060-151057060 on Build GRCh38
rs369848852 Chr.7: 151056885-151056885 on Build GRCh38
rs775128740 Chr.7: 151056799-151056800 on Build GRCh38
rs766236667 Chr.7: 151057111-151057111 on Build GRCh38
rs777007468 Chr.7: 151056761-151056761 on Build GRCh38
rs974834525 Chr.7: 151056886-151056886 on Build GRCh38
rs369446117 Chr.7: 151056745-151056745 on Build GRCh38
rs934470666 Chr.7: 151057003-151057003 on Build GRCh38
rs561605555 Chr.7: 151056879-151056879 on Build GRCh38
rs768156497 Chr.7: 151056845-151056848 on Build GRCh38
rs1036372253 Chr.7: 151056834-151056834 on Build GRCh38
rs1021541772 Chr.7: 151057125-151057125 on Build GRCh38
rs752895368 Chr.7: 151056867-151056867 on Build GRCh38
rs757350446 Chr.7: 151056999-151056999 on Build GRCh38
rs551975005 Chr.7: 151056896-151056896 on Build GRCh38
rs899846034 Chr.7: 151057027-151057027 on Build GRCh38
rs754771314 Chr.7: 151056990-151056990 on Build GRCh38
rs979667306 Chr.7: 151056721-151056721 on Build GRCh38
rs748438212 Chr.7: 151056715-151056715 on Build GRCh38
rs1038919252 Chr.7: 151056994-151056994 on Build GRCh38
rs370736023 Chr.7: 151056822-151056822 on Build GRCh38
rs2069450 Chr.7: 151056682-151056682 on Build GRCh38
rs905557030 Chr.7: 151056774-151056774 on Build GRCh38
rs373690199 Chr.7: 151056837-151056837 on Build GRCh38
rs879089142 Chr.7: 151056811-151056811 on Build GRCh38
rs531146226 Chr.7: 151056775-151056775 on Build GRCh38
rs775274881 Chr.7: 151056784-151056784 on Build GRCh38
rs1003109582 Chr.7: 151056777-151056777 on Build GRCh38
rs776562781 Chr.7: 151057120-151057120 on Build GRCh38
rs374660006 Chr.7: 151056889-151056889 on Build GRCh38
rs750386255 Chr.7: 151056861-151056861 on Build GRCh38
rs757172305 Chr.7: 151056692-151056692 on Build GRCh38
rs761373694 Chr.7: 151056941-151056941 on Build GRCh38
rs879174395 Chr.7: 151056740-151056740 on Build GRCh38
rs950885523 Chr.7: 151056853-151056853 on Build GRCh38
rs746360978 Chr.7: 151057028-151057028 on Build GRCh38
rs767434391 Chr.7: 151056858-151056858 on Build GRCh38
rs34247699 Chr.7: 151057035-151057036 on Build GRCh38
rs748993694 Chr.7: 151056819-151056828 on Build GRCh38
rs200087231 Chr.7: 151057113-151057113 on Build GRCh38
rs376160817 Chr.7: 151056726-151056726 on Build GRCh38
rs774164033 Chr.7: 151056914-151056914 on Build GRCh38
rs986705037 Chr.7: 151056957-151056957 on Build GRCh38
rs755876265 Chr.7: 151056711-151056711 on Build GRCh38
rs868359935 Chr.7: 151056686-151056686 on Build GRCh38
rs767003240 Chr.7: 151056952-151056952 on Build GRCh38
rs771429450 Chr.7: 151056757-151056757 on Build GRCh38
rs751076575 Chr.7: 151056831-151056831 on Build GRCh38
rs751770862 Chr.7: 151057126-151057126 on Build GRCh38
rs35475821 Chr.7: 151057127-151057128 on Build GRCh38
rs774002632 Chr.7: 151057037-151057040 on Build GRCh38
rs750169157 Chr.7: 151056705-151056705 on Build GRCh38
rs540638569 Chr.7: 151057029-151057029 on Build GRCh38
rs757297273 Chr.7: 151056882-151056882 on Build GRCh38
rs764241616 Chr.7: 151056995-151056995 on Build GRCh38
rs766324720 Chr.7: 151056967-151056967 on Build GRCh38
rs774049952 Chr.7: 151057048-151057048 on Build GRCh38
rs1016313768 Chr.7: 151056877-151056877 on Build GRCh38
rs760492249 Chr.7: 151057040-151057040 on Build GRCh38
rs1044191954 Chr.7: 151057103-151057103 on Build GRCh38
rs749704291 Chr.7: 151056894-151056894 on Build GRCh38
rs772550156 Chr.7: 151057062-151057062 on Build GRCh38
rs1029653154 Chr.7: 151057030-151057030 on Build GRCh38
rs758565818 Chr.7: 151056869-151056869 on Build GRCh38
rs912475421 Chr.7: 151056970-151056970 on Build GRCh38
rs747796391 Chr.7: 151057054-151057054 on Build GRCh38
rs954726581 Chr.7: 151056915-151056915 on Build GRCh38

View Full Product Details