Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27775383-27775929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESCO2
Entrez Gene ID
157570
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Gene Aliases
2410004I17Rik, EFO2, RBS
UniGene
Hs.99480
Gene Chromosome Location
Chr.8: 27771949-27812404 on Build GRCh38
Transcripts
NM_001017420.2 XM_011544421.2 XM_011544422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4487526 c.31C>G p.Q11E Chr.8: 27775545-27775545 on Build GRCh38
1580209 c.20G>A p.R7K Chr.8: 27775534-27775534 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760397891 Chr.8: 27775479-27775479 on Build GRCh38
rs753857305 Chr.8: 27775493-27775493 on Build GRCh38
rs901712193 Chr.8: 27775893-27775893 on Build GRCh38
rs916070606 Chr.8: 27775738-27775738 on Build GRCh38
rs771125221 Chr.8: 27775552-27775552 on Build GRCh38
rs781698140 Chr.8: 27775532-27775532 on Build GRCh38
rs188145997 Chr.8: 27775862-27775862 on Build GRCh38
rs145248677 Chr.8: 27775540-27775540 on Build GRCh38
rs748624179 Chr.8: 27775534-27775534 on Build GRCh38
rs183741062 Chr.8: 27775476-27775476 on Build GRCh38
rs769417701 Chr.8: 27775493-27775493 on Build GRCh38
rs750009035 Chr.8: 27775506-27775506 on Build GRCh38
rs764814109 Chr.8: 27775502-27775502 on Build GRCh38
rs975973678 Chr.8: 27775643-27775643 on Build GRCh38
rs751895817 Chr.8: 27775509-27775509 on Build GRCh38
rs924058722 Chr.8: 27775742-27775742 on Build GRCh38
rs965962199 Chr.8: 27775640-27775640 on Build GRCh38
rs762551069 Chr.8: 27775522-27775551 on Build GRCh38
rs768303970 Chr.8: 27775588-27775588 on Build GRCh38
rs936099791 Chr.8: 27775730-27775731 on Build GRCh38
rs923922856 Chr.8: 27775436-27775436 on Build GRCh38
rs997380401 Chr.8: 27775905-27775905 on Build GRCh38
rs906687862 Chr.8: 27775895-27775895 on Build GRCh38
rs761877763 Chr.8: 27775498-27775498 on Build GRCh38
rs765329515 Chr.8: 27775613-27775613 on Build GRCh38
rs141275859 Chr.8: 27775670-27775670 on Build GRCh38
rs376276857 Chr.8: 27775618-27775618 on Build GRCh38
rs758061966 Chr.8: 27775508-27775508 on Build GRCh38
rs755324574 Chr.8: 27775522-27775522 on Build GRCh38
rs115774603 Chr.8: 27775485-27775485 on Build GRCh38
rs775127606 Chr.8: 27775502-27775502 on Build GRCh38
rs1017874971 Chr.8: 27775619-27775619 on Build GRCh38
rs978193872 Chr.8: 27775423-27775423 on Build GRCh38
rs978161150 Chr.8: 27775737-27775737 on Build GRCh38
rs746778204 Chr.8: 27775576-27775576 on Build GRCh38
rs12335307 Chr.8: 27775871-27775871 on Build GRCh38
rs995137387 Chr.8: 27775651-27775651 on Build GRCh38
rs965267875 Chr.8: 27775733-27775733 on Build GRCh38
rs761824562 Chr.8: 27775609-27775609 on Build GRCh38
rs989488823 Chr.8: 27775749-27775749 on Build GRCh38
rs906927109 Chr.8: 27775565-27775565 on Build GRCh38
rs1045389398 Chr.8: 27775535-27775535 on Build GRCh38
rs1029614945 Chr.8: 27775650-27775650 on Build GRCh38
rs954113350 Chr.8: 27775692-27775692 on Build GRCh38
rs1056428817 Chr.8: 27775428-27775428 on Build GRCh38
rs141013081 Chr.8: 27775564-27775564 on Build GRCh38
rs763645904 Chr.8: 27775612-27775613 on Build GRCh38
rs1027442302 Chr.8: 27775547-27775547 on Build GRCh38
rs145048467 Chr.8: 27775723-27775723 on Build GRCh38
rs563281582 Chr.8: 27775434-27775434 on Build GRCh38
rs916809126 Chr.8: 27775851-27775851 on Build GRCh38
rs75977974 Chr.8: 27775720-27775720 on Build GRCh38
rs1038833135 Chr.8: 27775889-27775889 on Build GRCh38
rs201025850 Chr.8: 27775548-27775548 on Build GRCh38
rs896824499 Chr.8: 27775639-27775639 on Build GRCh38
rs897489669 Chr.8: 27775803-27775803 on Build GRCh38
rs895390052 Chr.8: 27775515-27775515 on Build GRCh38
rs528630260 Chr.8: 27775558-27775558 on Build GRCh38
rs928986834 Chr.8: 27775813-27775813 on Build GRCh38
rs146504750 Chr.8: 27775537-27775537 on Build GRCh38
rs936616725 Chr.8: 27775748-27775748 on Build GRCh38
rs776613336 Chr.8: 27775592-27775592 on Build GRCh38
rs985289277 Chr.8: 27775694-27775694 on Build GRCh38
rs751040173 Chr.8: 27775615-27775616 on Build GRCh38

View Full Product Details