Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28527771-28528280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD3
Entrez Gene ID
7976
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3
UniGene
Hs.40735
Gene Chromosome Location
Chr.8: 28494205-28574268 on Build GRCh38
Transcripts
NM_017412.3 NM_145866.1 XM_017013841.1 XM_017013842.1 XM_017013843.1 XM_017013844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910227 c.1012A>G p.I338V Chr.8: 28527772-28527772 on Build GRCh38
4924345 c.1158G>C p.G386G Chr.8: 28527918-28527918 on Build GRCh38
6261408 c.1404+5G>T p.? Chr.8: 28528169-28528169 on Build GRCh38
4842395 c.1057G>T p.E353* Chr.8: 28527817-28527817 on Build GRCh38
77743 c.1288G>C p.V430L Chr.8: 28528048-28528048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750667980 Chr.8: 28528077-28528077 on Build GRCh38
rs62001923 Chr.8: 28527827-28527827 on Build GRCh38
rs773528605 Chr.8: 28528225-28528225 on Build GRCh38
rs767945705 Chr.8: 28528047-28528047 on Build GRCh38
rs377106083 Chr.8: 28527927-28527927 on Build GRCh38
rs752479695 Chr.8: 28528194-28528194 on Build GRCh38
rs754801930 Chr.8: 28527933-28527933 on Build GRCh38
rs761890190 Chr.8: 28528063-28528063 on Build GRCh38
rs754930200 Chr.8: 28527858-28527858 on Build GRCh38
rs753973028 Chr.8: 28528061-28528061 on Build GRCh38
rs368964858 Chr.8: 28528120-28528120 on Build GRCh38
rs769145436 Chr.8: 28528167-28528167 on Build GRCh38
rs749229314 Chr.8: 28527814-28527814 on Build GRCh38
rs758164477 Chr.8: 28527807-28527807 on Build GRCh38
rs759992378 Chr.8: 28527889-28527889 on Build GRCh38
rs376897924 Chr.8: 28528181-28528181 on Build GRCh38
rs756134564 Chr.8: 28528115-28528115 on Build GRCh38
rs761069532 Chr.8: 28528160-28528160 on Build GRCh38
rs757539061 Chr.8: 28527924-28527924 on Build GRCh38
rs965485773 Chr.8: 28527809-28527809 on Build GRCh38
rs140023424 Chr.8: 28527879-28527879 on Build GRCh38
rs775987296 Chr.8: 28528158-28528158 on Build GRCh38
rs144171566 Chr.8: 28528023-28528023 on Build GRCh38
rs754710245 Chr.8: 28528082-28528082 on Build GRCh38
rs754026318 Chr.8: 28527912-28527912 on Build GRCh38
rs916568920 Chr.8: 28527934-28527934 on Build GRCh38
rs759937454 Chr.8: 28528044-28528044 on Build GRCh38
rs764281221 Chr.8: 28527905-28527905 on Build GRCh38
rs373092388 Chr.8: 28528175-28528175 on Build GRCh38
rs772289774 Chr.8: 28528152-28528152 on Build GRCh38
rs778766174 Chr.8: 28527995-28527995 on Build GRCh38
rs925745297 Chr.8: 28528219-28528219 on Build GRCh38
rs146503796 Chr.8: 28528098-28528098 on Build GRCh38
rs752661059 Chr.8: 28527802-28527802 on Build GRCh38
rs375741315 Chr.8: 28527838-28527838 on Build GRCh38
rs756222099 Chr.8: 28527803-28527803 on Build GRCh38
rs369898333 Chr.8: 28528191-28528191 on Build GRCh38
rs752575152 Chr.8: 28528100-28528100 on Build GRCh38
rs756096158 Chr.8: 28527903-28527903 on Build GRCh38
rs755866346 Chr.8: 28528216-28528216 on Build GRCh38
rs763304022 Chr.8: 28528186-28528186 on Build GRCh38
rs748935848 Chr.8: 28527996-28527996 on Build GRCh38
rs752336388 Chr.8: 28528183-28528183 on Build GRCh38
rs780945967 Chr.8: 28527935-28527935 on Build GRCh38
rs371916844 Chr.8: 28527837-28527837 on Build GRCh38
rs766846977 Chr.8: 28527878-28527878 on Build GRCh38
rs761281939 Chr.8: 28528048-28528048 on Build GRCh38
rs767844094 Chr.8: 28527890-28527890 on Build GRCh38
rs746743884 Chr.8: 28527930-28527930 on Build GRCh38
rs747574173 Chr.8: 28527847-28527847 on Build GRCh38
rs769156364 Chr.8: 28527951-28527951 on Build GRCh38
rs770596220 Chr.8: 28528129-28528129 on Build GRCh38
rs36106848 Chr.8: 28528151-28528152 on Build GRCh38
rs201317475 Chr.8: 28528116-28528116 on Build GRCh38
rs772847603 Chr.8: 28527962-28527962 on Build GRCh38
rs367706061 Chr.8: 28527805-28527805 on Build GRCh38
rs372290611 Chr.8: 28527859-28527859 on Build GRCh38
rs764072748 Chr.8: 28528056-28528056 on Build GRCh38
rs753227868 Chr.8: 28528236-28528236 on Build GRCh38
rs748739796 Chr.8: 28528125-28528125 on Build GRCh38
rs980009789 Chr.8: 28528020-28528020 on Build GRCh38
rs778658159 Chr.8: 28528130-28528130 on Build GRCh38
rs202185186 Chr.8: 28528164-28528164 on Build GRCh38
rs977535241 Chr.8: 28528203-28528203 on Build GRCh38
rs745564151 Chr.8: 28528140-28528140 on Build GRCh38
rs761836950 Chr.8: 28528179-28528179 on Build GRCh38
rs748169720 Chr.8: 28527950-28527950 on Build GRCh38
rs369695773 Chr.8: 28528066-28528066 on Build GRCh38
rs946118749 Chr.8: 28528196-28528196 on Build GRCh38
rs769370555 Chr.8: 28527856-28527856 on Build GRCh38
rs148953574 Chr.8: 28527804-28527804 on Build GRCh38

View Full Product Details