Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75071430-75071938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs752639475 Chr.X: 75071501-75071501 on Build GRCh38
rs756266541 Chr.X: 75071555-75071555 on Build GRCh38
rs758400043 Chr.X: 75071507-75071507 on Build GRCh38
rs951418177 Chr.X: 75071789-75071789 on Build GRCh38
rs759618917 Chr.X: 75071715-75071715 on Build GRCh38
rs1054919 Chr.X: 75071639-75071639 on Build GRCh38
rs955046524 Chr.X: 75071632-75071632 on Build GRCh38
rs887974659 Chr.X: 75071491-75071491 on Build GRCh38
rs770439607 Chr.X: 75071694-75071694 on Build GRCh38
rs758906595 Chr.X: 75071706-75071706 on Build GRCh38
rs1029747021 Chr.X: 75071631-75071631 on Build GRCh38
rs960162078 Chr.X: 75071874-75071874 on Build GRCh38
rs780346552 Chr.X: 75071566-75071566 on Build GRCh38
rs778824146 Chr.X: 75071466-75071466 on Build GRCh38
rs751420864 Chr.X: 75071515-75071515 on Build GRCh38
rs763867013 Chr.X: 75071513-75071513 on Build GRCh38
rs144548601 Chr.X: 75071892-75071892 on Build GRCh38
rs72554634 Chr.X: 75071516-75071516 on Build GRCh38
rs189786647 Chr.X: 75071724-75071724 on Build GRCh38
rs749798732 Chr.X: 75071585-75071585 on Build GRCh38
rs1052386741 Chr.X: 75071517-75071517 on Build GRCh38
rs995626687 Chr.X: 75071712-75071712 on Build GRCh38
rs1004416904 Chr.X: 75071842-75071842 on Build GRCh38
rs777525606 Chr.X: 75071891-75071891 on Build GRCh38
rs139311518 Chr.X: 75071542-75071542 on Build GRCh38
rs201269113 Chr.X: 75071484-75071484 on Build GRCh38
rs1008068103 Chr.X: 75071586-75071586 on Build GRCh38
rs776356620 Chr.X: 75071702-75071702 on Build GRCh38
rs989932613 Chr.X: 75071846-75071846 on Build GRCh38
rs182454573 Chr.X: 75071766-75071766 on Build GRCh38
rs769292032 Chr.X: 75071711-75071711 on Build GRCh38
rs779385966 Chr.X: 75071687-75071687 on Build GRCh38
rs928640506 Chr.X: 75071909-75071909 on Build GRCh38
rs962282106 Chr.X: 75071614-75071614 on Build GRCh38
rs945969400 Chr.X: 75071861-75071861 on Build GRCh38
rs1035903516 Chr.X: 75071855-75071855 on Build GRCh38
rs765098755 Chr.X: 75071499-75071499 on Build GRCh38
rs769334662 Chr.X: 75071630-75071630 on Build GRCh38
rs756708420 Chr.X: 75071857-75071857 on Build GRCh38

View Full Product Details