Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 76177594-76178095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBDC1
Entrez Gene ID
51260
Gene Name
polysaccharide biosynthesis domain containing 1
Gene Aliases
CXorf26
UniGene
Hs.370100
Gene Chromosome Location
Chr.X: 76172929-76178310 on Build GRCh38
Transcripts
NM_001300888.1 NM_016500.3

SNP Detail

SNP ID Chromosome Location
rs781798798 Chr.X: 76177727-76177727 on Build GRCh38
rs781892755 Chr.X: 76177790-76177791 on Build GRCh38
rs782526685 Chr.X: 76177753-76177753 on Build GRCh38
rs782506241 Chr.X: 76177732-76177733 on Build GRCh38
rs782609609 Chr.X: 76177760-76177760 on Build GRCh38
rs781917400 Chr.X: 76177876-76177876 on Build GRCh38
rs781972823 Chr.X: 76177623-76177623 on Build GRCh38
rs782240498 Chr.X: 76177806-76177806 on Build GRCh38
rs113132484 Chr.X: 76177915-76177915 on Build GRCh38
rs371423465 Chr.X: 76177725-76177725 on Build GRCh38
rs782802063 Chr.X: 76177938-76177938 on Build GRCh38
rs782704294 Chr.X: 76177967-76177967 on Build GRCh38
rs782479623 Chr.X: 76178011-76178011 on Build GRCh38
rs782614821 Chr.X: 76178012-76178012 on Build GRCh38
rs371947297 Chr.X: 76177859-76177859 on Build GRCh38
rs782723141 Chr.X: 76177980-76177980 on Build GRCh38
rs782574776 Chr.X: 76177872-76177873 on Build GRCh38
rs201110824 Chr.X: 76177929-76177929 on Build GRCh38
rs782522144 Chr.X: 76177746-76177746 on Build GRCh38
rs1018383402 Chr.X: 76178036-76178036 on Build GRCh38
rs375986100 Chr.X: 76177657-76177657 on Build GRCh38
rs781821096 Chr.X: 76177707-76177707 on Build GRCh38
rs782093848 Chr.X: 76177713-76177713 on Build GRCh38
rs367739861 Chr.X: 76177851-76177851 on Build GRCh38
rs781806861 Chr.X: 76177972-76177972 on Build GRCh38
rs138763959 Chr.X: 76178025-76178025 on Build GRCh38
rs782257187 Chr.X: 76178005-76178005 on Build GRCh38
rs894994978 Chr.X: 76177842-76177842 on Build GRCh38
rs782020940 Chr.X: 76177665-76177665 on Build GRCh38
rs782416718 Chr.X: 76177828-76177828 on Build GRCh38
rs781904025 Chr.X: 76177776-76177776 on Build GRCh38
rs782709186 Chr.X: 76177658-76177658 on Build GRCh38
rs201962153 Chr.X: 76177874-76177874 on Build GRCh38
rs896805478 Chr.X: 76177918-76177918 on Build GRCh38
rs200499663 Chr.X: 76177671-76177671 on Build GRCh38
rs781866536 Chr.X: 76177751-76177751 on Build GRCh38
rs782578760 Chr.X: 76177818-76177818 on Build GRCh38
rs782180884 Chr.X: 76178020-76178020 on Build GRCh38
rs782629564 Chr.X: 76177698-76177698 on Build GRCh38
rs782776824 Chr.X: 76177684-76177684 on Build GRCh38
rs782591890 Chr.X: 76177966-76177966 on Build GRCh38
rs374809824 Chr.X: 76177748-76177748 on Build GRCh38
rs782362053 Chr.X: 76178065-76178065 on Build GRCh38
rs782325020 Chr.X: 76178024-76178024 on Build GRCh38
rs782190448 Chr.X: 76177988-76177988 on Build GRCh38
rs782782122 Chr.X: 76177789-76177790 on Build GRCh38
rs782786228 Chr.X: 76177959-76177959 on Build GRCh38
rs782235441 Chr.X: 76177763-76177763 on Build GRCh38
rs200305612 Chr.X: 76177941-76177941 on Build GRCh38
rs41305417 Chr.X: 76177965-76177965 on Build GRCh38
rs782276032 Chr.X: 76177819-76177819 on Build GRCh38
rs782284152 Chr.X: 76178051-76178051 on Build GRCh38
rs782003432 Chr.X: 76177831-76177831 on Build GRCh38
rs782571223 Chr.X: 76177947-76177947 on Build GRCh38

View Full Product Details