Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 96885006-96885516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH2
Entrez Gene ID
1730
Gene Name
diaphanous related formin 2
Gene Aliases
DIA, DIA2, DRF2, POF, POF2
UniGene
Hs.226483
Gene Chromosome Location
Chr.X: 96684663-97600598 on Build GRCh38
Transcripts
NM_006729.4 NM_007309.3 NM_013347.4

Gene Symbol
RPA4
Entrez Gene ID
29935
Gene Name
replication protein A4
Gene Aliases
HSU24186
UniGene
Hs.659349
Gene Chromosome Location
Chr.X: 96883908-96885467 on Build GRCh38
Transcripts
NM_006729.4 NM_007309.3 NM_013347.4

SNP Detail

SNP ID Chromosome Location
rs759307486 Chr.X: 96885051-96885051 on Build GRCh38
rs750760682 Chr.X: 96885092-96885092 on Build GRCh38
rs200339720 Chr.X: 96885091-96885091 on Build GRCh38
rs770119909 Chr.X: 96885239-96885239 on Build GRCh38
rs926179539 Chr.X: 96885180-96885180 on Build GRCh38
rs878867138 Chr.X: 96885469-96885469 on Build GRCh38
rs189894696 Chr.X: 96885397-96885397 on Build GRCh38
rs751615388 Chr.X: 96885086-96885086 on Build GRCh38
rs899024247 Chr.X: 96885472-96885472 on Build GRCh38
rs748114324 Chr.X: 96885047-96885047 on Build GRCh38
rs60706314 Chr.X: 96885472-96885474 on Build GRCh38
rs762246801 Chr.X: 96885088-96885088 on Build GRCh38
rs1034303727 Chr.X: 96885476-96885476 on Build GRCh38
rs890208479 Chr.X: 96885389-96885389 on Build GRCh38
rs752450234 Chr.X: 96885124-96885124 on Build GRCh38
rs369646035 Chr.X: 96885075-96885075 on Build GRCh38
rs931679096 Chr.X: 96885285-96885285 on Build GRCh38
rs746159525 Chr.X: 96885157-96885157 on Build GRCh38
rs770863451 Chr.X: 96885040-96885040 on Build GRCh38
rs910085722 Chr.X: 96885454-96885454 on Build GRCh38
rs1055286501 Chr.X: 96885402-96885402 on Build GRCh38
rs3138319 Chr.X: 96885358-96885358 on Build GRCh38
rs778673464 Chr.X: 96885037-96885037 on Build GRCh38
rs201295573 Chr.X: 96885050-96885050 on Build GRCh38
rs1032834162 Chr.X: 96885188-96885188 on Build GRCh38
rs1046541047 Chr.X: 96885447-96885447 on Build GRCh38
rs894475970 Chr.X: 96885478-96885478 on Build GRCh38
rs756616469 Chr.X: 96885104-96885104 on Build GRCh38
rs988727190 Chr.X: 96885272-96885272 on Build GRCh38
rs773250379 Chr.X: 96885400-96885400 on Build GRCh38
rs982189765 Chr.X: 96885336-96885336 on Build GRCh38
rs905019930 Chr.X: 96885449-96885449 on Build GRCh38
rs746960917 Chr.X: 96885038-96885038 on Build GRCh38
rs994745730 Chr.X: 96885475-96885475 on Build GRCh38
rs375123066 Chr.X: 96885066-96885066 on Build GRCh38
rs936275504 Chr.X: 96885211-96885211 on Build GRCh38
rs928057000 Chr.X: 96885344-96885344 on Build GRCh38
rs967120371 Chr.X: 96885283-96885283 on Build GRCh38
rs1006658486 Chr.X: 96885184-96885185 on Build GRCh38
rs373416639 Chr.X: 96885117-96885117 on Build GRCh38
rs913863192 Chr.X: 96885273-96885276 on Build GRCh38
rs921605855 Chr.X: 96885245-96885245 on Build GRCh38
rs937867352 Chr.X: 96885490-96885490 on Build GRCh38
rs757353111 Chr.X: 96885060-96885060 on Build GRCh38
rs34989154 Chr.X: 96885171-96885171 on Build GRCh38
rs764194530 Chr.X: 96885076-96885076 on Build GRCh38

View Full Product Details