Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15572786-15573383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMAT
Entrez Gene ID
79814
Gene Name
agmatinase
Gene Aliases
-
UniGene
Hs.461532
Gene Chromosome Location
Chr.1: 15571699-15585110 on Build GRCh38
Transcripts
NM_024758.4

SNP Detail

SNP ID Chromosome Location
rs955532765 Chr.1: 15573167-15573167 on Build GRCh38
rs538052488 Chr.1: 15573035-15573035 on Build GRCh38
rs144631679 Chr.1: 15572819-15572819 on Build GRCh38
rs912912179 Chr.1: 15573215-15573215 on Build GRCh38
rs888091511 Chr.1: 15573337-15573337 on Build GRCh38
rs183502615 Chr.1: 15573006-15573006 on Build GRCh38
rs970232350 Chr.1: 15572974-15572974 on Build GRCh38
rs71572165 Chr.1: 15573269-15573269 on Build GRCh38
rs972727254 Chr.1: 15572868-15572868 on Build GRCh38
rs758186793 Chr.1: 15573319-15573319 on Build GRCh38
rs956280624 Chr.1: 15573227-15573227 on Build GRCh38
rs987419948 Chr.1: 15572923-15572923 on Build GRCh38
rs75693268 Chr.1: 15572838-15572838 on Build GRCh38
rs1038219453 Chr.1: 15573251-15573252 on Build GRCh38
rs943863645 Chr.1: 15573120-15573120 on Build GRCh38
rs898293538 Chr.1: 15573260-15573261 on Build GRCh38
rs748226692 Chr.1: 15573335-15573335 on Build GRCh38
rs985457048 Chr.1: 15573263-15573263 on Build GRCh38
rs753910644 Chr.1: 15573191-15573191 on Build GRCh38
rs538988089 Chr.1: 15573033-15573033 on Build GRCh38
rs889585680 Chr.1: 15573073-15573073 on Build GRCh38
rs540900142 Chr.1: 15573183-15573183 on Build GRCh38
rs1018566602 Chr.1: 15573297-15573297 on Build GRCh38
rs947453399 Chr.1: 15573021-15573021 on Build GRCh38
rs987083570 Chr.1: 15573168-15573168 on Build GRCh38
rs750280711 Chr.1: 15573295-15573295 on Build GRCh38
rs34201882 Chr.1: 15573269-15573269 on Build GRCh38
rs397979324 Chr.1: 15573268-15573269 on Build GRCh38
rs990985118 Chr.1: 15572905-15572905 on Build GRCh38
rs1023100833 Chr.1: 15572965-15572965 on Build GRCh38
rs58974289 Chr.1: 15572837-15572837 on Build GRCh38
rs573637851 Chr.1: 15573241-15573241 on Build GRCh38
rs969133450 Chr.1: 15573244-15573244 on Build GRCh38
rs937193113 Chr.1: 15573245-15573245 on Build GRCh38
rs549770999 Chr.1: 15572820-15572820 on Build GRCh38
rs1025936255 Chr.1: 15572892-15572892 on Build GRCh38
rs757282233 Chr.1: 15573208-15573208 on Build GRCh38
rs542645314 Chr.1: 15573267-15573267 on Build GRCh38
rs569339147 Chr.1: 15573034-15573034 on Build GRCh38
rs1045675072 Chr.1: 15573237-15573237 on Build GRCh38
rs1040849551 Chr.1: 15573282-15573282 on Build GRCh38
rs1048025438 Chr.1: 15573169-15573169 on Build GRCh38
rs1001757774 Chr.1: 15573118-15573118 on Build GRCh38
rs111441028 Chr.1: 15573174-15573174 on Build GRCh38
rs188083413 Chr.1: 15573235-15573235 on Build GRCh38
rs911412829 Chr.1: 15572955-15572955 on Build GRCh38
rs994527614 Chr.1: 15573160-15573160 on Build GRCh38
rs117761427 Chr.1: 15572975-15572975 on Build GRCh38
rs1035893265 Chr.1: 15573214-15573214 on Build GRCh38
rs918425158 Chr.1: 15573286-15573286 on Build GRCh38
rs1040882231 Chr.1: 15573126-15573126 on Build GRCh38
rs775760521 Chr.1: 15573116-15573116 on Build GRCh38
rs1027107088 Chr.1: 15572824-15572824 on Build GRCh38
rs925737625 Chr.1: 15573200-15573200 on Build GRCh38
rs71572164 Chr.1: 15573239-15573239 on Build GRCh38
rs11587541 Chr.1: 15572983-15572983 on Build GRCh38
rs1023376768 Chr.1: 15573190-15573190 on Build GRCh38
rs111820574 Chr.1: 15573159-15573159 on Build GRCh38
rs891420076 Chr.1: 15572996-15572996 on Build GRCh38
rs964267012 Chr.1: 15573176-15573176 on Build GRCh38
rs36051155 Chr.1: 15573258-15573259 on Build GRCh38
rs928852685 Chr.1: 15572976-15572976 on Build GRCh38
rs952488870 Chr.1: 15572901-15572901 on Build GRCh38
rs181507186 Chr.1: 15573325-15573325 on Build GRCh38
rs898575088 Chr.1: 15573151-15573151 on Build GRCh38
rs571122175 Chr.1: 15573257-15573258 on Build GRCh38
rs141398135 Chr.1: 15572985-15572985 on Build GRCh38
rs1011901837 Chr.1: 15573349-15573349 on Build GRCh38

View Full Product Details