Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20548132-20548676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2B
Entrez Gene ID
348158
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Gene Aliases
ACSM2, HXMA, HYST1046
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20536261-20576379 on Build GRCh38
Transcripts
NM_001105069.1 NM_182617.3 XM_005255304.3 XM_011545824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
968092 c.1012G>A p.G338R Chr.16: 20548148-20548148 on Build GRCh38
968093 c.996G>T p.Q332H Chr.16: 20548164-20548164 on Build GRCh38
138072 c.947G>A p.R316Q Chr.16: 20548421-20548421 on Build GRCh38
358988 c.927G>T p.M309I Chr.16: 20548441-20548441 on Build GRCh38
4994401 c.932C>T p.A311V Chr.16: 20548436-20548436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368782954 Chr.16: 20548503-20548503 on Build GRCh38
rs753389371 Chr.16: 20548160-20548160 on Build GRCh38
rs748079187 Chr.16: 20548368-20548368 on Build GRCh38
rs781221104 Chr.16: 20548360-20548360 on Build GRCh38
rs377750115 Chr.16: 20548218-20548218 on Build GRCh38
rs747642495 Chr.16: 20548424-20548424 on Build GRCh38
rs751238118 Chr.16: 20548518-20548518 on Build GRCh38
rs745608503 Chr.16: 20548171-20548171 on Build GRCh38
rs759981521 Chr.16: 20548153-20548153 on Build GRCh38
rs143170043 Chr.16: 20548179-20548179 on Build GRCh38
rs749408371 Chr.16: 20548194-20548194 on Build GRCh38
rs552030555 Chr.16: 20548479-20548479 on Build GRCh38
rs141915122 Chr.16: 20548432-20548432 on Build GRCh38
rs769199008 Chr.16: 20548475-20548476 on Build GRCh38
rs936281651 Chr.16: 20548295-20548295 on Build GRCh38
rs971114531 Chr.16: 20548650-20548650 on Build GRCh38
rs150487730 Chr.16: 20548183-20548183 on Build GRCh38
rs527652870 Chr.16: 20548508-20548508 on Build GRCh38
rs1055010179 Chr.16: 20548296-20548296 on Build GRCh38
rs780735542 Chr.16: 20548347-20548347 on Build GRCh38
rs774605190 Chr.16: 20548203-20548203 on Build GRCh38
rs771022459 Chr.16: 20548196-20548196 on Build GRCh38
rs549091203 Chr.16: 20548520-20548520 on Build GRCh38
rs373238431 Chr.16: 20548307-20548307 on Build GRCh38
rs761101298 Chr.16: 20548220-20548220 on Build GRCh38
rs80135299 Chr.16: 20548455-20548455 on Build GRCh38
rs865944833 Chr.16: 20548174-20548174 on Build GRCh38
rs1054724397 Chr.16: 20548461-20548461 on Build GRCh38
rs545276915 Chr.16: 20548305-20548305 on Build GRCh38
rs750066612 Chr.16: 20548232-20548232 on Build GRCh38
rs549256374 Chr.16: 20548154-20548154 on Build GRCh38
rs761046293 Chr.16: 20548389-20548389 on Build GRCh38
rs769110427 Chr.16: 20548390-20548390 on Build GRCh38
rs756822230 Chr.16: 20548227-20548227 on Build GRCh38
rs762376567 Chr.16: 20548492-20548492 on Build GRCh38
rs771024973 Chr.16: 20548370-20548370 on Build GRCh38
rs1006941651 Chr.16: 20548535-20548535 on Build GRCh38
rs183078161 Chr.16: 20548175-20548175 on Build GRCh38
rs1044651606 Chr.16: 20548270-20548270 on Build GRCh38
rs762639289 Chr.16: 20548422-20548422 on Build GRCh38
rs563063734 Chr.16: 20548435-20548435 on Build GRCh38
rs372027953 Chr.16: 20548468-20548468 on Build GRCh38
rs756798470 Chr.16: 20548163-20548163 on Build GRCh38
rs772731162 Chr.16: 20548499-20548499 on Build GRCh38
rs775306499 Chr.16: 20548613-20548613 on Build GRCh38
rs754792708 Chr.16: 20548434-20548434 on Build GRCh38
rs759323773 Chr.16: 20548519-20548519 on Build GRCh38
rs773369239 Chr.16: 20548193-20548193 on Build GRCh38
rs571414902 Chr.16: 20548645-20548645 on Build GRCh38
rs762273713 Chr.16: 20548397-20548397 on Build GRCh38
rs866561627 Chr.16: 20548466-20548466 on Build GRCh38
rs577817277 Chr.16: 20548292-20548292 on Build GRCh38
rs534063904 Chr.16: 20548178-20548178 on Build GRCh38
rs778982594 Chr.16: 20548449-20548449 on Build GRCh38
rs369876624 Chr.16: 20548548-20548548 on Build GRCh38
rs537623821 Chr.16: 20548244-20548244 on Build GRCh38
rs144594944 Chr.16: 20548401-20548401 on Build GRCh38
rs138313532 Chr.16: 20548404-20548404 on Build GRCh38
rs1024158614 Chr.16: 20548649-20548649 on Build GRCh38
rs184398923 Chr.16: 20548644-20548644 on Build GRCh38
rs983198225 Chr.16: 20548420-20548420 on Build GRCh38
rs767858638 Chr.16: 20548209-20548209 on Build GRCh38
rs560930442 Chr.16: 20548391-20548391 on Build GRCh38
rs758181398 Chr.16: 20548237-20548237 on Build GRCh38
rs771975876 Chr.16: 20548252-20548252 on Build GRCh38
rs556040483 Chr.16: 20548259-20548259 on Build GRCh38
rs778649584 Chr.16: 20548167-20548167 on Build GRCh38
rs551525300 Chr.16: 20548159-20548159 on Build GRCh38
rs746101181 Chr.16: 20548377-20548377 on Build GRCh38
rs373891337 Chr.16: 20548442-20548442 on Build GRCh38
rs764594793 Chr.16: 20548221-20548221 on Build GRCh38
rs746816181 Chr.16: 20548351-20548351 on Build GRCh38
rs368523670 Chr.16: 20548448-20548448 on Build GRCh38
rs753235347 Chr.16: 20548225-20548225 on Build GRCh38
rs772229711 Chr.16: 20548458-20548458 on Build GRCh38
rs776915411 Chr.16: 20548491-20548491 on Build GRCh38
rs780282645 Chr.16: 20548459-20548459 on Build GRCh38
rs906143020 Chr.16: 20548294-20548294 on Build GRCh38
rs1018205319 Chr.16: 20548545-20548545 on Build GRCh38
rs759728707 Chr.16: 20548205-20548205 on Build GRCh38
rs1010705562 Chr.16: 20548317-20548317 on Build GRCh38
rs149635560 Chr.16: 20548421-20548421 on Build GRCh38
rs754914965 Chr.16: 20548352-20548352 on Build GRCh38
rs756077029 Chr.16: 20548446-20548446 on Build GRCh38
rs1027612759 Chr.16: 20548601-20548601 on Build GRCh38
rs950588269 Chr.16: 20548408-20548408 on Build GRCh38
rs367576858 Chr.16: 20548384-20548384 on Build GRCh38
rs192049191 Chr.16: 20548243-20548243 on Build GRCh38
rs1029734257 Chr.16: 20548333-20548333 on Build GRCh38
rs775896795 Chr.16: 20548210-20548210 on Build GRCh38
rs778920770 Chr.16: 20548372-20548372 on Build GRCh38
rs891121493 Chr.16: 20548341-20548341 on Build GRCh38
rs764694017 Chr.16: 20548228-20548228 on Build GRCh38
rs766152891 Chr.16: 20548425-20548425 on Build GRCh38
rs542777805 Chr.16: 20548386-20548386 on Build GRCh38
rs192778610 Chr.16: 20548581-20548581 on Build GRCh38
rs184866258 Chr.16: 20548349-20548349 on Build GRCh38
rs539762138 Chr.16: 20548155-20548155 on Build GRCh38
rs772283008 Chr.16: 20548381-20548381 on Build GRCh38
rs375114238 Chr.16: 20548498-20548498 on Build GRCh38
rs983313696 Chr.16: 20548642-20548642 on Build GRCh38

View Full Product Details