Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97531812-97532384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004591.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs537921043 Chr.2: 97531981-97531981 on Build GRCh38
rs775967509 Chr.2: 97532282-97532282 on Build GRCh38
rs540797794 Chr.2: 97532266-97532266 on Build GRCh38
rs188842496 Chr.2: 97532149-97532149 on Build GRCh38
rs574609575 Chr.2: 97532112-97532112 on Build GRCh38
rs554735540 Chr.2: 97532060-97532060 on Build GRCh38
rs759129475 Chr.2: 97532307-97532307 on Build GRCh38
rs558518987 Chr.2: 97531963-97531963 on Build GRCh38
rs201315366 Chr.2: 97531967-97531967 on Build GRCh38
rs577753741 Chr.2: 97532182-97532182 on Build GRCh38
rs769305366 Chr.2: 97532352-97532352 on Build GRCh38

View Full Product Details