Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31996521-31997073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4A
Entrez Gene ID
720
Gene Name
complement component 4A (Rodgers blood group)
Gene Aliases
C4, C4A2, C4A3, C4A4, C4A6, C4AD, C4S, CO4, CPAMD2, RG
UniGene
Hs.534847
Gene Chromosome Location
Chr.6: 31982057-32002680 on Build GRCh38
Transcripts
NM_001252204.1 NM_007293.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408889 c.3629T>G p.L1210R Chr.6: 31996553-31996553 on Build GRCh38
5130652 c.3615G>C p.A1205A Chr.6: 31996539-31996539 on Build GRCh38
6616086 c.3614C>T p.A1205V Chr.6: 31996538-31996538 on Build GRCh38
4408887 c.3630C>G p.L1210L Chr.6: 31996554-31996554 on Build GRCh38
7358564 c.3620T>C p.V1207A Chr.6: 31996544-31996544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780167237 Chr.6: 31996618-31996618 on Build GRCh38
rs547592026 Chr.6: 31996754-31996754 on Build GRCh38
rs201016130 Chr.6: 31997008-31997008 on Build GRCh38
rs769471597 Chr.6: 31996569-31996569 on Build GRCh38
rs189630921 Chr.6: 31996807-31996807 on Build GRCh38
rs28357077 Chr.6: 31996554-31996554 on Build GRCh38
rs963187351 Chr.6: 31996799-31996799 on Build GRCh38
rs368403366 Chr.6: 31996887-31996887 on Build GRCh38
rs571742632 Chr.6: 31996641-31996641 on Build GRCh38
rs28357074 Chr.6: 31996539-31996539 on Build GRCh38
rs754984776 Chr.6: 31996627-31996627 on Build GRCh38
rs2854862 Chr.6: 31996614-31996614 on Build GRCh38
rs755076640 Chr.6: 31997017-31997017 on Build GRCh38
rs564578360 Chr.6: 31996618-31996618 on Build GRCh38
rs556468429 Chr.6: 31997021-31997021 on Build GRCh38
rs766593106 Chr.6: 31996986-31996986 on Build GRCh38
rs776372955 Chr.6: 31996555-31996555 on Build GRCh38
rs201113435 Chr.6: 31996877-31996877 on Build GRCh38
rs752041550 Chr.6: 31996608-31996608 on Build GRCh38
rs538797527 Chr.6: 31996736-31996736 on Build GRCh38
rs774944419 Chr.6: 31996815-31996815 on Build GRCh38
rs1016397819 Chr.6: 31996738-31996738 on Build GRCh38
rs200485982 Chr.6: 31997010-31997010 on Build GRCh38
rs770810578 Chr.6: 31996639-31996639 on Build GRCh38
rs561358842 Chr.6: 31996576-31996576 on Build GRCh38
rs759091047 Chr.6: 31996558-31996558 on Build GRCh38
rs775131580 Chr.6: 31996585-31996585 on Build GRCh38
rs748152082 Chr.6: 31996542-31996542 on Build GRCh38
rs753927647 Chr.6: 31996623-31996623 on Build GRCh38
rs756257625 Chr.6: 31996615-31996615 on Build GRCh38
rs769553963 Chr.6: 31996646-31996646 on Build GRCh38
rs751558109 Chr.6: 31997023-31997023 on Build GRCh38
rs775221248 Chr.6: 31996647-31996647 on Build GRCh38
rs746876947 Chr.6: 31996638-31996638 on Build GRCh38
rs573614770 Chr.6: 31996969-31996969 on Build GRCh38
rs5030681 Chr.6: 31996878-31996878 on Build GRCh38
rs1002089632 Chr.6: 31996854-31996854 on Build GRCh38
rs555040887 Chr.6: 31996892-31996892 on Build GRCh38
rs370657873 Chr.6: 31996551-31996551 on Build GRCh38
rs531891216 Chr.6: 31996606-31996606 on Build GRCh38
rs762202053 Chr.6: 31996872-31996872 on Build GRCh38
rs533662784 Chr.6: 31996886-31996886 on Build GRCh38
rs28357075 Chr.6: 31996544-31996544 on Build GRCh38
rs777723410 Chr.6: 31996636-31996636 on Build GRCh38
rs780083644 Chr.6: 31996540-31996540 on Build GRCh38
rs764707844 Chr.6: 31996607-31996607 on Build GRCh38
rs71220974 Chr.6: 31996539-31996554 on Build GRCh38
rs745598448 Chr.6: 31996643-31996643 on Build GRCh38
rs28357076 Chr.6: 31996553-31996553 on Build GRCh38
rs1004522192 Chr.6: 31996675-31996675 on Build GRCh38
rs762553974 Chr.6: 31996783-31996783 on Build GRCh38
rs760602547 Chr.6: 31996844-31996845 on Build GRCh38

View Full Product Details