Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41728406-41728987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 NM_080817.4 XM_017029414.1

Gene Symbol
GPR82
Entrez Gene ID
27197
Gene Name
G protein-coupled receptor 82
Gene Aliases
-
UniGene
Hs.567457
Gene Chromosome Location
Chr.X: 41724155-41730135 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 NM_080817.4 XM_017029414.1

SNP Detail

SNP ID Chromosome Location
rs942338420 Chr.X: 41728849-41728849 on Build GRCh38
rs900888509 Chr.X: 41728942-41728942 on Build GRCh38
rs112349119 Chr.X: 41728824-41728824 on Build GRCh38
rs972595726 Chr.X: 41728753-41728753 on Build GRCh38
rs947811804 Chr.X: 41728707-41728707 on Build GRCh38
rs900940727 Chr.X: 41728570-41728570 on Build GRCh38
rs982153079 Chr.X: 41728717-41728717 on Build GRCh38
rs910808593 Chr.X: 41728826-41728826 on Build GRCh38
rs768457304 Chr.X: 41728906-41728906 on Build GRCh38
rs916308347 Chr.X: 41728499-41728499 on Build GRCh38
rs956771270 Chr.X: 41728669-41728669 on Build GRCh38
rs779995773 Chr.X: 41728778-41728778 on Build GRCh38
rs935032439 Chr.X: 41728728-41728728 on Build GRCh38
rs746034319 Chr.X: 41728508-41728508 on Build GRCh38
rs779304340 Chr.X: 41728481-41728481 on Build GRCh38
rs1010810945 Chr.X: 41728673-41728673 on Build GRCh38
rs1030834160 Chr.X: 41728652-41728652 on Build GRCh38
rs919413104 Chr.X: 41728767-41728767 on Build GRCh38
rs980769847 Chr.X: 41728697-41728697 on Build GRCh38
rs969352669 Chr.X: 41728679-41728679 on Build GRCh38
rs1052342675 Chr.X: 41728759-41728759 on Build GRCh38
rs930905181 Chr.X: 41728431-41728431 on Build GRCh38
rs5917445 Chr.X: 41728916-41728916 on Build GRCh38
rs758074536 Chr.X: 41728674-41728674 on Build GRCh38
rs1039389821 Chr.X: 41728556-41728556 on Build GRCh38
rs867487050 Chr.X: 41728689-41728689 on Build GRCh38
rs772200492 Chr.X: 41728668-41728668 on Build GRCh38
rs928000498 Chr.X: 41728721-41728721 on Build GRCh38
rs1022648001 Chr.X: 41728674-41728674 on Build GRCh38
rs960773334 Chr.X: 41728739-41728739 on Build GRCh38
rs764017816 Chr.X: 41728696-41728696 on Build GRCh38
rs1049799332 Chr.X: 41728480-41728480 on Build GRCh38
rs751623209 Chr.X: 41728823-41728823 on Build GRCh38
rs888097889 Chr.X: 41728485-41728485 on Build GRCh38
rs998379953 Chr.X: 41728637-41728637 on Build GRCh38
rs997927622 Chr.X: 41728952-41728952 on Build GRCh38

View Full Product Details