Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50042259-50042468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN5
Entrez Gene ID
1184
Gene Name
chloride voltage-gated channel 5
Gene Aliases
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2
UniGene
Hs.166486
Gene Chromosome Location
Chr.X: 49922615-50099235 on Build GRCh38
Transcripts
NM_001127898.3 NM_001127899.3 NM_001272102.1 XM_017029257.1 XM_017029258.1

SNP Detail

SNP ID Chromosome Location
rs782744227 Chr.X: 50042351-50042351 on Build GRCh38
rs376680776 Chr.X: 50042298-50042298 on Build GRCh38
rs781863873 Chr.X: 50042321-50042321 on Build GRCh38
rs781788759 Chr.X: 50042355-50042355 on Build GRCh38
rs916340219 Chr.X: 50042390-50042390 on Build GRCh38
rs960373474 Chr.X: 50042358-50042358 on Build GRCh38
rs782155673 Chr.X: 50042286-50042286 on Build GRCh38
rs991814636 Chr.X: 50042364-50042364 on Build GRCh38
rs782364195 Chr.X: 50042415-50042415 on Build GRCh38
rs375788993 Chr.X: 50042441-50042441 on Build GRCh38
rs111464924 Chr.X: 50042407-50042407 on Build GRCh38
rs782084525 Chr.X: 50042343-50042343 on Build GRCh38
rs202230774 Chr.X: 50042394-50042394 on Build GRCh38
rs782515109 Chr.X: 50042419-50042419 on Build GRCh38

View Full Product Details