Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 4988035-4988606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C2
Entrez Gene ID
1646
Gene Name
aldo-keto reductase family 1, member C2
Gene Aliases
AKR1C-pseudo, BABP, DD, DD-2, DD/BABP, DD2, DDH2, HAKRD, HBAB, MCDR2, SRXY8, TDD
UniGene
Hs.460260
Gene Chromosome Location
Chr.10: 4987775-5018033 on Build GRCh38
Transcripts
NM_001321027.1 NM_001354.5 NM_205845.2

SNP Detail

SNP ID Chromosome Location
rs568577093 Chr.10: 4988406-4988406 on Build GRCh38
rs185209880 Chr.10: 4988412-4988412 on Build GRCh38
rs576858828 Chr.10: 4988555-4988555 on Build GRCh38
rs548010547 Chr.10: 4988436-4988436 on Build GRCh38
rs782765357 Chr.10: 4988490-4988490 on Build GRCh38
rs7092939 Chr.10: 4988258-4988258 on Build GRCh38
rs371169226 Chr.10: 4988427-4988427 on Build GRCh38
rs188067373 Chr.10: 4988567-4988567 on Build GRCh38
rs537081793 Chr.10: 4988482-4988482 on Build GRCh38
rs778865201 Chr.10: 4988541-4988541 on Build GRCh38
rs563785896 Chr.10: 4988228-4988228 on Build GRCh38
rs781795014 Chr.10: 4988553-4988553 on Build GRCh38
rs782698003 Chr.10: 4988200-4988201 on Build GRCh38
rs45460998 Chr.10: 4988287-4988287 on Build GRCh38
rs45615836 Chr.10: 4988163-4988163 on Build GRCh38
rs541986258 Chr.10: 4988580-4988580 on Build GRCh38
rs781990273 Chr.10: 4988532-4988532 on Build GRCh38
rs180720395 Chr.10: 4988579-4988579 on Build GRCh38
rs559285155 Chr.10: 4988173-4988173 on Build GRCh38
rs569766206 Chr.10: 4988448-4988448 on Build GRCh38
rs558941983 Chr.10: 4988076-4988076 on Build GRCh38
rs373788438 Chr.10: 4988432-4988432 on Build GRCh38
rs4584482 Chr.10: 4988341-4988341 on Build GRCh38
rs764746619 Chr.10: 4988102-4988102 on Build GRCh38
rs4526684 Chr.10: 4988334-4988334 on Build GRCh38
rs143633992 Chr.10: 4988491-4988491 on Build GRCh38
rs577197610 Chr.10: 4988135-4988135 on Build GRCh38
rs562996110 Chr.10: 4988158-4988158 on Build GRCh38
rs567718455 Chr.10: 4988390-4988390 on Build GRCh38
rs11252867 Chr.10: 4988176-4988176 on Build GRCh38
rs11252866 Chr.10: 4988075-4988075 on Build GRCh38
rs373635725 Chr.10: 4988561-4988561 on Build GRCh38
rs547245513 Chr.10: 4988350-4988350 on Build GRCh38
rs541973210 Chr.10: 4988197-4988197 on Build GRCh38
rs71391984 Chr.10: 4988427-4988427 on Build GRCh38
rs534633045 Chr.10: 4988556-4988556 on Build GRCh38

View Full Product Details