Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37136122-37136689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1646283 c.644C>A p.T215N Chr.10: 37136663-37136663 on Build GRCh38
1603433 c.616C>T p.R206* Chr.10: 37136635-37136635 on Build GRCh38
3382844 c.605T>G p.M202R Chr.10: 37136624-37136624 on Build GRCh38
3382843 c.605T>G p.M202R Chr.10: 37136624-37136624 on Build GRCh38
1603432 c.616C>T p.R206* Chr.10: 37136635-37136635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541729693 Chr.10: 37136641-37136641 on Build GRCh38
rs753448468 Chr.10: 37136623-37136623 on Build GRCh38
rs944174971 Chr.10: 37136603-37136603 on Build GRCh38
rs751014359 Chr.10: 37136651-37136651 on Build GRCh38
rs766017589 Chr.10: 37136534-37136534 on Build GRCh38
rs888322338 Chr.10: 37136460-37136460 on Build GRCh38
rs1004066049 Chr.10: 37136480-37136480 on Build GRCh38
rs879076424 Chr.10: 37136618-37136618 on Build GRCh38
rs372977683 Chr.10: 37136566-37136566 on Build GRCh38
rs1015344796 Chr.10: 37136212-37136212 on Build GRCh38
rs377107925 Chr.10: 37136599-37136599 on Build GRCh38
rs924739164 Chr.10: 37136354-37136354 on Build GRCh38
rs970858546 Chr.10: 37136225-37136225 on Build GRCh38
rs778843251 Chr.10: 37136609-37136609 on Build GRCh38
rs556234146 Chr.10: 37136567-37136567 on Build GRCh38
rs1004417598 Chr.10: 37136211-37136211 on Build GRCh38
rs989112557 Chr.10: 37136544-37136544 on Build GRCh38
rs534636239 Chr.10: 37136353-37136353 on Build GRCh38
rs944106878 Chr.10: 37136365-37136365 on Build GRCh38
rs762692703 Chr.10: 37136640-37136640 on Build GRCh38
rs573679735 Chr.10: 37136522-37136522 on Build GRCh38
rs1200907 Chr.10: 37136310-37136310 on Build GRCh38
rs868405570 Chr.10: 37136191-37136191 on Build GRCh38
rs111949755 Chr.10: 37136192-37136192 on Build GRCh38
rs771207963 Chr.10: 37136602-37136602 on Build GRCh38
rs888269897 Chr.10: 37136210-37136210 on Build GRCh38
rs761284654 Chr.10: 37136625-37136625 on Build GRCh38
rs958969257 Chr.10: 37136299-37136299 on Build GRCh38
rs571576105 Chr.10: 37136256-37136256 on Build GRCh38
rs906313081 Chr.10: 37136513-37136513 on Build GRCh38
rs932604385 Chr.10: 37136425-37136425 on Build GRCh38
rs1048304983 Chr.10: 37136441-37136441 on Build GRCh38
rs554468201 Chr.10: 37136461-37136461 on Build GRCh38
rs760257197 Chr.10: 37136474-37136474 on Build GRCh38
rs565353332 Chr.10: 37136614-37136614 on Build GRCh38
rs755863563 Chr.10: 37136578-37136578 on Build GRCh38
rs761274769 Chr.10: 37136156-37136156 on Build GRCh38
rs1003687391 Chr.10: 37136520-37136520 on Build GRCh38
rs772426152 Chr.10: 37136619-37136619 on Build GRCh38
rs959019163 Chr.10: 37136530-37136530 on Build GRCh38
rs1041074369 Chr.10: 37136374-37136374 on Build GRCh38
rs754424267 Chr.10: 37136662-37136662 on Build GRCh38
rs373922256 Chr.10: 37136264-37136264 on Build GRCh38
rs1015813257 Chr.10: 37136511-37136511 on Build GRCh38
rs1026814543 Chr.10: 37136197-37136197 on Build GRCh38
rs914778313 Chr.10: 37136309-37136309 on Build GRCh38
rs538781472 Chr.10: 37136282-37136282 on Build GRCh38
rs182505178 Chr.10: 37136300-37136300 on Build GRCh38
rs776021846 Chr.10: 37136622-37136622 on Build GRCh38
rs777188054 Chr.10: 37136308-37136309 on Build GRCh38
rs977645143 Chr.10: 37136570-37136570 on Build GRCh38
rs769081186 Chr.10: 37136635-37136635 on Build GRCh38
rs542649996 Chr.10: 37136554-37136554 on Build GRCh38
rs576461920 Chr.10: 37136570-37136570 on Build GRCh38
rs201363452 Chr.10: 37136636-37136636 on Build GRCh38
rs899841722 Chr.10: 37136397-37136397 on Build GRCh38
rs777568423 Chr.10: 37136594-37136594 on Build GRCh38
rs752403702 Chr.10: 37136572-37136572 on Build GRCh38
rs1033334767 Chr.10: 37136292-37136292 on Build GRCh38
rs117816508 Chr.10: 37136158-37136158 on Build GRCh38
rs561609736 Chr.10: 37136654-37136654 on Build GRCh38

View Full Product Details