Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50009193-50009752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP6
Entrez Gene ID
414189
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 6
Gene Aliases
AGAP-6, CTGLF3, bA324H6.1
UniGene
Hs.536158
Gene Chromosome Location
Chr.10: 49981196-50010507 on Build GRCh38
Transcripts
NM_001077665.2 XM_005269841.3 XM_011539814.2 XM_011539815.2 XM_017016268.1 XM_017016269.1 XM_017016270.1 XM_017016271.1 XM_017016272.1 XM_017016273.1 XM_017016274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
427649 c.1579G>A p.E527K Chr.10: 50009704-50009704 on Build GRCh38
3438524 c.1178C>T p.P393L Chr.10: 50009303-50009303 on Build GRCh38
7354732 c.1252G>T p.G418C Chr.10: 50009377-50009377 on Build GRCh38
6246603 c.1179C>A p.P393P Chr.10: 50009304-50009304 on Build GRCh38
3438525 c.1574C>T p.P525L Chr.10: 50009699-50009699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781906789 Chr.10: 50009229-50009229 on Build GRCh38
rs782229361 Chr.10: 50009541-50009541 on Build GRCh38
rs782635075 Chr.10: 50009695-50009695 on Build GRCh38
rs556366729 Chr.10: 50009538-50009538 on Build GRCh38
rs782696877 Chr.10: 50009512-50009512 on Build GRCh38
rs782798272 Chr.10: 50009331-50009331 on Build GRCh38
rs782399202 Chr.10: 50009553-50009553 on Build GRCh38
rs576744772 Chr.10: 50009697-50009697 on Build GRCh38
rs547724641 Chr.10: 50009511-50009511 on Build GRCh38
rs782314825 Chr.10: 50009606-50009607 on Build GRCh38
rs781904809 Chr.10: 50009604-50009604 on Build GRCh38
rs781878415 Chr.10: 50009334-50009334 on Build GRCh38
rs192536832 Chr.10: 50009669-50009669 on Build GRCh38
rs781818014 Chr.10: 50009566-50009566 on Build GRCh38
rs569844118 Chr.10: 50009555-50009555 on Build GRCh38
rs202194597 Chr.10: 50009617-50009617 on Build GRCh38
rs201137183 Chr.10: 50009597-50009597 on Build GRCh38
rs782129777 Chr.10: 50009446-50009446 on Build GRCh38
rs782183685 Chr.10: 50009417-50009417 on Build GRCh38
rs781848590 Chr.10: 50009508-50009508 on Build GRCh38
rs17153398 Chr.10: 50009406-50009406 on Build GRCh38
rs782546200 Chr.10: 50009298-50009299 on Build GRCh38
rs782762320 Chr.10: 50009506-50009506 on Build GRCh38
rs782590376 Chr.10: 50009416-50009416 on Build GRCh38
rs782281904 Chr.10: 50009531-50009531 on Build GRCh38
rs559895140 Chr.10: 50009302-50009302 on Build GRCh38
rs782591361 Chr.10: 50009720-50009720 on Build GRCh38
rs782697799 Chr.10: 50009472-50009472 on Build GRCh38
rs535652391 Chr.10: 50009577-50009577 on Build GRCh38
rs782521552 Chr.10: 50009384-50009384 on Build GRCh38
rs782511800 Chr.10: 50009689-50009689 on Build GRCh38
rs782422992 Chr.10: 50009423-50009423 on Build GRCh38
rs527595643 Chr.10: 50009465-50009465 on Build GRCh38
rs782613590 Chr.10: 50009618-50009618 on Build GRCh38
rs782175600 Chr.10: 50009275-50009275 on Build GRCh38
rs782690352 Chr.10: 50009359-50009359 on Build GRCh38
rs532204635 Chr.10: 50009304-50009304 on Build GRCh38
rs782660528 Chr.10: 50009631-50009631 on Build GRCh38
rs545810161 Chr.10: 50009701-50009701 on Build GRCh38
rs782734305 Chr.10: 50009312-50009312 on Build GRCh38
rs782670419 Chr.10: 50009303-50009304 on Build GRCh38
rs551999415 Chr.10: 50009354-50009354 on Build GRCh38
rs782627730 Chr.10: 50009389-50009389 on Build GRCh38
rs782018914 Chr.10: 50009440-50009440 on Build GRCh38
rs782654601 Chr.10: 50009246-50009246 on Build GRCh38
rs531472394 Chr.10: 50009732-50009732 on Build GRCh38
rs782477493 Chr.10: 50009271-50009271 on Build GRCh38
rs572445484 Chr.10: 50009290-50009290 on Build GRCh38
rs782160378 Chr.10: 50009546-50009546 on Build GRCh38
rs562634946 Chr.10: 50009372-50009372 on Build GRCh38
rs782002512 Chr.10: 50009554-50009554 on Build GRCh38
rs781862580 Chr.10: 50009613-50009613 on Build GRCh38
rs187547602 Chr.10: 50009644-50009644 on Build GRCh38
rs781900073 Chr.10: 50009473-50009473 on Build GRCh38
rs782484069 Chr.10: 50009509-50009509 on Build GRCh38
rs145420670 Chr.10: 50009297-50009297 on Build GRCh38
rs782173673 Chr.10: 50009305-50009305 on Build GRCh38
rs782249705 Chr.10: 50009256-50009256 on Build GRCh38
rs782574361 Chr.10: 50009274-50009274 on Build GRCh38
rs782170231 Chr.10: 50009661-50009661 on Build GRCh38
rs782323010 Chr.10: 50009542-50009542 on Build GRCh38
rs568354305 Chr.10: 50009454-50009454 on Build GRCh38
rs200123722 Chr.10: 50009298-50009298 on Build GRCh38
rs782758890 Chr.10: 50009565-50009565 on Build GRCh38
rs199841016 Chr.10: 50009645-50009645 on Build GRCh38
rs781968202 Chr.10: 50009301-50009301 on Build GRCh38
rs781939409 Chr.10: 50009654-50009655 on Build GRCh38
rs548450491 Chr.10: 50009452-50009452 on Build GRCh38
rs782227612 Chr.10: 50009390-50009390 on Build GRCh38
rs782206845 Chr.10: 50009646-50009646 on Build GRCh38
rs782507972 Chr.10: 50009343-50009343 on Build GRCh38
rs782169172 Chr.10: 50009322-50009322 on Build GRCh38
rs200482356 Chr.10: 50009648-50009648 on Build GRCh38
rs539887710 Chr.10: 50009532-50009532 on Build GRCh38
rs782416513 Chr.10: 50009288-50009288 on Build GRCh38
rs781826206 Chr.10: 50009675-50009675 on Build GRCh38
rs782394225 Chr.10: 50009437-50009438 on Build GRCh38
rs781917709 Chr.10: 50009545-50009545 on Build GRCh38
rs781884810 Chr.10: 50009682-50009682 on Build GRCh38
rs782298492 Chr.10: 50009548-50009548 on Build GRCh38
rs782248215 Chr.10: 50009350-50009350 on Build GRCh38
rs781922840 Chr.10: 50009658-50009658 on Build GRCh38
rs781800962 Chr.10: 50009347-50009347 on Build GRCh38
rs782145081 Chr.10: 50009493-50009493 on Build GRCh38
rs570782198 Chr.10: 50009528-50009528 on Build GRCh38
rs782797026 Chr.10: 50009679-50009679 on Build GRCh38
rs782701537 Chr.10: 50009595-50009595 on Build GRCh38
rs782649939 Chr.10: 50009605-50009605 on Build GRCh38
rs782596634 Chr.10: 50009510-50009510 on Build GRCh38
rs782445748 Chr.10: 50009678-50009678 on Build GRCh38
rs782005152 Chr.10: 50009668-50009668 on Build GRCh38
rs781970063 Chr.10: 50009463-50009463 on Build GRCh38
rs782177959 Chr.10: 50009623-50009623 on Build GRCh38
rs782726996 Chr.10: 50009346-50009346 on Build GRCh38
rs782767193 Chr.10: 50009674-50009674 on Build GRCh38
rs782547018 Chr.10: 50009628-50009628 on Build GRCh38
rs781927015 Chr.10: 50009318-50009318 on Build GRCh38
rs562438643 Chr.10: 50009728-50009728 on Build GRCh38
rs782180358 Chr.10: 50009726-50009726 on Build GRCh38
rs562024691 Chr.10: 50009233-50009233 on Build GRCh38

View Full Product Details