Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50129771-50130335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
UniGene
Hs.365286
Gene Chromosome Location
Chr.10: 50067888-50133509 on Build GRCh38
Transcripts
NM_001005751.2 NM_001291398.1 XM_005269806.3 XM_005269807.3 XM_005269808.3 XM_005269810.3 XM_006717831.3 XM_006717832.3 XM_011539786.2 XM_017016227.1 XM_017016228.1 XM_017016229.1 XM_017016230.1 XM_017016231.1 XM_017016232.1 XM_017016233.1 XM_017016234.1 XM_017016235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4816749 c.3607G>T p.E1203* Chr.10: 50129938-50129938 on Build GRCh38
918790 c.3612T>C p.D1204D Chr.10: 50129943-50129943 on Build GRCh38
918789 c.3582G>T p.K1194N Chr.10: 50129913-50129913 on Build GRCh38
6254790 c.3527G>T p.G1176V Chr.10: 50129858-50129858 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928610438 Chr.10: 50129842-50129842 on Build GRCh38
rs915804221 Chr.10: 50129986-50129986 on Build GRCh38
rs886898146 Chr.10: 50130163-50130163 on Build GRCh38
rs529269673 Chr.10: 50130144-50130144 on Build GRCh38
rs996443251 Chr.10: 50130190-50130190 on Build GRCh38
rs779770152 Chr.10: 50130031-50130031 on Build GRCh38
rs2805247 Chr.10: 50130270-50130270 on Build GRCh38
rs1029213287 Chr.10: 50130260-50130260 on Build GRCh38
rs2490906 Chr.10: 50130235-50130235 on Build GRCh38
rs563559800 Chr.10: 50130057-50130057 on Build GRCh38
rs757236634 Chr.10: 50129866-50129866 on Build GRCh38
rs931170602 Chr.10: 50130091-50130091 on Build GRCh38
rs977861971 Chr.10: 50129837-50129837 on Build GRCh38
rs778773421 Chr.10: 50129867-50129867 on Build GRCh38
rs1048210028 Chr.10: 50130104-50130104 on Build GRCh38
rs1044597 Chr.10: 50129859-50129859 on Build GRCh38
rs577899274 Chr.10: 50129854-50129854 on Build GRCh38
rs900849034 Chr.10: 50130179-50130179 on Build GRCh38
rs761911701 Chr.10: 50129998-50130001 on Build GRCh38
rs967467880 Chr.10: 50129821-50129821 on Build GRCh38
rs1055739999 Chr.10: 50129974-50129976 on Build GRCh38
rs745414360 Chr.10: 50130003-50130003 on Build GRCh38
rs2669761 Chr.10: 50129923-50129923 on Build GRCh38
rs2261404 Chr.10: 50130191-50130191 on Build GRCh38
rs543782780 Chr.10: 50130055-50130055 on Build GRCh38
rs889352580 Chr.10: 50130269-50130269 on Build GRCh38
rs1041160104 Chr.10: 50130176-50130176 on Build GRCh38
rs549029968 Chr.10: 50130166-50130166 on Build GRCh38
rs771416807 Chr.10: 50130024-50130024 on Build GRCh38

View Full Product Details