Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50742507-50743100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2B
Entrez Gene ID
653308
Gene Name
N-acylsphingosine amidohydrolase 2B
Gene Aliases
ASAH2C, ASAH2L, bA449O16.3, bA98I6.3
UniGene
Hs.659820
Gene Chromosome Location
Chr.10: 50739688-50757188 on Build GRCh38
Transcripts
NM_001079516.3 NM_001321957.1 NM_001321958.1 NM_001321959.1 NM_001321960.1 NR_135904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4791571 c.86G>A p.R29H Chr.10: 50743010-50743010 on Build GRCh38
4663586 c.36T>C p.H12H Chr.10: 50742960-50742960 on Build GRCh38
1348146 c.28C>T p.R10C Chr.10: 50742952-50742952 on Build GRCh38
1348145 c.14G>A p.R5Q Chr.10: 50742938-50742938 on Build GRCh38
5425941 c.33G>A p.T11T Chr.10: 50742957-50742957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566698822 Chr.10: 50743015-50743015 on Build GRCh38
rs141006980 Chr.10: 50743009-50743009 on Build GRCh38
rs755238658 Chr.10: 50743036-50743036 on Build GRCh38
rs375438645 Chr.10: 50742872-50742872 on Build GRCh38
rs551101710 Chr.10: 50742938-50742938 on Build GRCh38
rs113060459 Chr.10: 50743058-50743059 on Build GRCh38
rs528397055 Chr.10: 50742713-50742713 on Build GRCh38
rs761352406 Chr.10: 50742929-50742929 on Build GRCh38
rs749562795 Chr.10: 50742996-50742996 on Build GRCh38
rs1008801919 Chr.10: 50742842-50742842 on Build GRCh38
rs751131343 Chr.10: 50743044-50743047 on Build GRCh38
rs112405752 Chr.10: 50743043-50743044 on Build GRCh38
rs1047408586 Chr.10: 50742755-50742755 on Build GRCh38
rs536636894 Chr.10: 50743047-50743047 on Build GRCh38
rs201084180 Chr.10: 50743043-50743043 on Build GRCh38
rs757030559 Chr.10: 50743044-50743045 on Build GRCh38
rs1017419061 Chr.10: 50742572-50742572 on Build GRCh38
rs767908313 Chr.10: 50743040-50743040 on Build GRCh38
rs878983542 Chr.10: 50742696-50742696 on Build GRCh38
rs749328323 Chr.10: 50743044-50743044 on Build GRCh38
rs1044576361 Chr.10: 50742670-50742671 on Build GRCh38
rs564227818 Chr.10: 50742567-50742571 on Build GRCh38
rs898802939 Chr.10: 50742738-50742738 on Build GRCh38
rs1033522070 Chr.10: 50742895-50742895 on Build GRCh38
rs772210240 Chr.10: 50743028-50743035 on Build GRCh38
rs199913552 Chr.10: 50743010-50743010 on Build GRCh38
rs370992697 Chr.10: 50742921-50742921 on Build GRCh38
rs764875712 Chr.10: 50743027-50743027 on Build GRCh38
rs767910726 Chr.10: 50743039-50743039 on Build GRCh38
rs964816377 Chr.10: 50742573-50742573 on Build GRCh38
rs917489707 Chr.10: 50742653-50742653 on Build GRCh38
rs747278108 Chr.10: 50742562-50742563 on Build GRCh38
rs1035033344 Chr.10: 50743065-50743065 on Build GRCh38
rs372083437 Chr.10: 50742952-50742952 on Build GRCh38
rs761842684 Chr.10: 50743020-50743020 on Build GRCh38
rs1008728022 Chr.10: 50743021-50743021 on Build GRCh38
rs567646122 Chr.10: 50742953-50742953 on Build GRCh38
rs577535492 Chr.10: 50742544-50742544 on Build GRCh38
rs771321958 Chr.10: 50743034-50743035 on Build GRCh38
rs993588771 Chr.10: 50742751-50742751 on Build GRCh38
rs765696411 Chr.10: 50743031-50743032 on Build GRCh38
rs201056952 Chr.10: 50743032-50743032 on Build GRCh38
rs989046400 Chr.10: 50742648-50742648 on Build GRCh38
rs563213732 Chr.10: 50742635-50742635 on Build GRCh38
rs200222392 Chr.10: 50743035-50743035 on Build GRCh38
rs78203725 Chr.10: 50743028-50743028 on Build GRCh38
rs201972903 Chr.10: 50743060-50743060 on Build GRCh38
rs543540954 Chr.10: 50742622-50742622 on Build GRCh38
rs769847170 Chr.10: 50743042-50743043 on Build GRCh38
rs755538895 Chr.10: 50742958-50742958 on Build GRCh38
rs762714922 Chr.10: 50743031-50743031 on Build GRCh38
rs565297225 Chr.10: 50742833-50742833 on Build GRCh38
rs535430308 Chr.10: 50742837-50742837 on Build GRCh38
rs979292552 Chr.10: 50742625-50742625 on Build GRCh38
rs890359748 Chr.10: 50742777-50742777 on Build GRCh38
rs199824484 Chr.10: 50742922-50742922 on Build GRCh38
rs925936321 Chr.10: 50742637-50742637 on Build GRCh38
rs530796473 Chr.10: 50742852-50742852 on Build GRCh38
rs2820741 Chr.10: 50742877-50742877 on Build GRCh38
rs200791229 Chr.10: 50742940-50742940 on Build GRCh38
rs533459084 Chr.10: 50742639-50742639 on Build GRCh38
rs757333774 Chr.10: 50742566-50742611 on Build GRCh38
rs762071523 Chr.10: 50743032-50743033 on Build GRCh38
rs781484406 Chr.10: 50742965-50742965 on Build GRCh38
rs767216286 Chr.10: 50743050-50743051 on Build GRCh38
rs890158335 Chr.10: 50743012-50743012 on Build GRCh38
rs777811994 Chr.10: 50743027-50743028 on Build GRCh38
rs189336118 Chr.10: 50743069-50743069 on Build GRCh38
rs763194796 Chr.10: 50742903-50742903 on Build GRCh38
rs778947551 Chr.10: 50743053-50743053 on Build GRCh38
rs1037865217 Chr.10: 50742719-50742719 on Build GRCh38
rs75066720 Chr.10: 50742980-50742980 on Build GRCh38
rs2820740 Chr.10: 50742843-50742843 on Build GRCh38
rs748239801 Chr.10: 50742971-50742971 on Build GRCh38
rs147249784 Chr.10: 50743002-50743002 on Build GRCh38
rs771894850 Chr.10: 50742920-50742920 on Build GRCh38
rs927412246 Chr.10: 50742966-50742966 on Build GRCh38
rs759458326 Chr.10: 50743039-50743040 on Build GRCh38
rs768655023 Chr.10: 50743011-50743011 on Build GRCh38
rs370588693 Chr.10: 50742923-50742923 on Build GRCh38
rs758742603 Chr.10: 50743045-50743046 on Build GRCh38
rs2137567 Chr.10: 50742690-50742690 on Build GRCh38
rs748134354 Chr.10: 50742876-50742876 on Build GRCh38
rs2820739 Chr.10: 50742654-50742654 on Build GRCh38
rs746092924 Chr.10: 50743058-50743058 on Build GRCh38
rs373985642 Chr.10: 50742956-50742956 on Build GRCh38
rs199570537 Chr.10: 50742563-50742567 on Build GRCh38
rs2820742 Chr.10: 50742957-50742957 on Build GRCh38
rs139833523 Chr.10: 50743028-50743031 on Build GRCh38
rs573823415 Chr.10: 50742647-50742647 on Build GRCh38
rs112930164 Chr.10: 50742889-50742889 on Build GRCh38
rs551672779 Chr.10: 50742826-50742826 on Build GRCh38
rs776541658 Chr.10: 50743038-50743039 on Build GRCh38
rs987247728 Chr.10: 50742546-50742546 on Build GRCh38

View Full Product Details