Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32530002-32530508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRB5
Entrez Gene ID
3127
Gene Name
major histocompatibility complex, class II, DR beta 5
Gene Aliases
HLA-DRB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 32517343-32530316 on Build GRCh38
Transcripts
NM_002125.3 XM_011514562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594102 c.41A>T p.K14M Chr.6: 32530184-32530184 on Build GRCh38
4383354 c.32A>G p.Y11C Chr.6: 32530193-32530193 on Build GRCh38
4354461 c.23G>A p.G8E Chr.6: 32530202-32530202 on Build GRCh38
4160958 c.74C>G p.P25R Chr.6: 32530151-32530151 on Build GRCh38
3949045 c.41A>C p.K14T Chr.6: 32530184-32530184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879090079 Chr.6: 32530344-32530344 on Build GRCh38
rs376456293 Chr.6: 32530279-32530279 on Build GRCh38
rs372737047 Chr.6: 32530321-32530321 on Build GRCh38
rs892520804 Chr.6: 32530254-32530254 on Build GRCh38
rs147565130 Chr.6: 32530154-32530154 on Build GRCh38
rs375722180 Chr.6: 32530080-32530081 on Build GRCh38
rs796754225 Chr.6: 32530131-32530131 on Build GRCh38
rs950473973 Chr.6: 32530308-32530308 on Build GRCh38
rs761350323 Chr.6: 32530210-32530210 on Build GRCh38
rs757118435 Chr.6: 32530129-32530129 on Build GRCh38
rs758791100 Chr.6: 32530260-32530260 on Build GRCh38
rs115877620 Chr.6: 32530026-32530026 on Build GRCh38
rs372766916 Chr.6: 32530317-32530320 on Build GRCh38
rs536256191 Chr.6: 32530338-32530348 on Build GRCh38
rs116331107 Chr.6: 32530397-32530397 on Build GRCh38
rs201406360 Chr.6: 32530341-32530341 on Build GRCh38
rs746445670 Chr.6: 32530263-32530263 on Build GRCh38
rs752947305 Chr.6: 32530181-32530193 on Build GRCh38
rs767848519 Chr.6: 32530081-32530081 on Build GRCh38
rs374223960 Chr.6: 32530257-32530257 on Build GRCh38
rs113487995 Chr.6: 32530249-32530249 on Build GRCh38
rs867813797 Chr.6: 32530407-32530407 on Build GRCh38
rs199711463 Chr.6: 32530107-32530107 on Build GRCh38
rs200223877 Chr.6: 32530265-32530265 on Build GRCh38
rs76918408 Chr.6: 32530070-32530070 on Build GRCh38
rs755600757 Chr.6: 32530184-32530185 on Build GRCh38
rs796472812 Chr.6: 32530277-32530277 on Build GRCh38
rs776720378 Chr.6: 32530192-32530192 on Build GRCh38
rs375527972 Chr.6: 32530083-32530084 on Build GRCh38
rs878961506 Chr.6: 32530455-32530455 on Build GRCh38
rs796691988 Chr.6: 32530289-32530289 on Build GRCh38
rs751229947 Chr.6: 32530174-32530174 on Build GRCh38
rs701884 Chr.6: 32530185-32530185 on Build GRCh38
rs879235443 Chr.6: 32530390-32530390 on Build GRCh38
rs762139450 Chr.6: 32530080-32530080 on Build GRCh38
rs796272411 Chr.6: 32530393-32530393 on Build GRCh38
rs371504447 Chr.6: 32530423-32530423 on Build GRCh38
rs775871391 Chr.6: 32530161-32530161 on Build GRCh38
rs200251101 Chr.6: 32530215-32530215 on Build GRCh38
rs146828696 Chr.6: 32530412-32530412 on Build GRCh38
rs72508462 Chr.6: 32530141-32530141 on Build GRCh38
rs150791953 Chr.6: 32530194-32530194 on Build GRCh38
rs67598789 Chr.6: 32530290-32530290 on Build GRCh38
rs774116269 Chr.6: 32530205-32530206 on Build GRCh38
rs867660709 Chr.6: 32530437-32530437 on Build GRCh38
rs879127840 Chr.6: 32530044-32530044 on Build GRCh38
rs759847860 Chr.6: 32530158-32530158 on Build GRCh38
rs749696415 Chr.6: 32530208-32530208 on Build GRCh38
rs781258565 Chr.6: 32530094-32530094 on Build GRCh38
rs79192142 Chr.6: 32530124-32530124 on Build GRCh38
rs778850959 Chr.6: 32530203-32530203 on Build GRCh38
rs796816400 Chr.6: 32530273-32530273 on Build GRCh38
rs766078306 Chr.6: 32530213-32530213 on Build GRCh38
rs748671570 Chr.6: 32530228-32530228 on Build GRCh38
rs760117345 Chr.6: 32530247-32530247 on Build GRCh38
rs569845377 Chr.6: 32530219-32530219 on Build GRCh38
rs71549218 Chr.6: 32530093-32530093 on Build GRCh38
rs118146761 Chr.6: 32530361-32530361 on Build GRCh38
rs796467893 Chr.6: 32530396-32530396 on Build GRCh38
rs143669594 Chr.6: 32530429-32530429 on Build GRCh38
rs760761586 Chr.6: 32530227-32530228 on Build GRCh38
rs148158278 Chr.6: 32530431-32530431 on Build GRCh38
rs76562035 Chr.6: 32530119-32530119 on Build GRCh38
rs772273827 Chr.6: 32530226-32530227 on Build GRCh38
rs772154187 Chr.6: 32530143-32530143 on Build GRCh38
rs773409911 Chr.6: 32530148-32530148 on Build GRCh38
rs759310728 Chr.6: 32530236-32530236 on Build GRCh38
rs75375694 Chr.6: 32530037-32530037 on Build GRCh38
rs769156845 Chr.6: 32530221-32530221 on Build GRCh38
rs201954564 Chr.6: 32530047-32530047 on Build GRCh38
rs200298244 Chr.6: 32530113-32530113 on Build GRCh38
rs71549223 Chr.6: 32530317-32530317 on Build GRCh38
rs779550694 Chr.6: 32530227-32530227 on Build GRCh38
rs745343465 Chr.6: 32530182-32530183 on Build GRCh38
rs777879171 Chr.6: 32530190-32530190 on Build GRCh38
rs113793658 Chr.6: 32530264-32530264 on Build GRCh38
rs756922265 Chr.6: 32530181-32530182 on Build GRCh38
rs71549224 Chr.6: 32530458-32530458 on Build GRCh38
rs746730708 Chr.6: 32530092-32530092 on Build GRCh38
rs201418311 Chr.6: 32530114-32530114 on Build GRCh38
rs879213035 Chr.6: 32530183-32530185 on Build GRCh38
rs78612727 Chr.6: 32530123-32530123 on Build GRCh38
rs796540798 Chr.6: 32530322-32530322 on Build GRCh38
rs200744795 Chr.6: 32530209-32530209 on Build GRCh38
rs773428210 Chr.6: 32530115-32530115 on Build GRCh38
rs72851011 Chr.6: 32530256-32530256 on Build GRCh38
rs879214621 Chr.6: 32530428-32530510 on Build GRCh38
rs757983305 Chr.6: 32530125-32530125 on Build GRCh38
rs192953520 Chr.6: 32530358-32530358 on Build GRCh38
rs114195860 Chr.6: 32530198-32530198 on Build GRCh38
rs773952098 Chr.6: 32530195-32530195 on Build GRCh38
rs62405579 Chr.6: 32530357-32530357 on Build GRCh38
rs75457801 Chr.6: 32530183-32530183 on Build GRCh38
rs6935811 Chr.6: 32530091-32530091 on Build GRCh38
rs796887252 Chr.6: 32530320-32530320 on Build GRCh38
rs755923622 Chr.6: 32530201-32530201 on Build GRCh38
rs17211043 Chr.6: 32530166-32530166 on Build GRCh38
rs796338972 Chr.6: 32530184-32530185 on Build GRCh38
rs745888965 Chr.6: 32530096-32530096 on Build GRCh38
rs753424292 Chr.6: 32530445-32530446 on Build GRCh38
rs753179812 Chr.6: 32530180-32530181 on Build GRCh38
rs1052909160 Chr.6: 32530251-32530251 on Build GRCh38
rs781460957 Chr.6: 32530205-32530205 on Build GRCh38
rs776891477 Chr.6: 32530230-32530231 on Build GRCh38
rs16868406 Chr.6: 32530428-32530428 on Build GRCh38
rs558566792 Chr.6: 32530299-32530299 on Build GRCh38
rs142225194 Chr.6: 32530191-32530191 on Build GRCh38
rs780457135 Chr.6: 32530220-32530220 on Build GRCh38
rs749189559 Chr.6: 32530185-32530186 on Build GRCh38
rs367610892 Chr.6: 32530073-32530073 on Build GRCh38
rs765668095 Chr.6: 32530128-32530128 on Build GRCh38
rs866181165 Chr.6: 32530319-32530319 on Build GRCh38
rs751570209 Chr.6: 32530204-32530204 on Build GRCh38
rs73727923 Chr.6: 32530151-32530151 on Build GRCh38
rs765082286 Chr.6: 32530237-32530237 on Build GRCh38
rs147483481 Chr.6: 32530083-32530083 on Build GRCh38
rs750059933 Chr.6: 32530199-32530199 on Build GRCh38
rs758441088 Chr.6: 32530187-32530187 on Build GRCh38
rs754442364 Chr.6: 32530087-32530087 on Build GRCh38
rs865910049 Chr.6: 32530366-32530366 on Build GRCh38
rs796196559 Chr.6: 32530465-32530465 on Build GRCh38
rs762843856 Chr.6: 32530243-32530243 on Build GRCh38
rs768644441 Chr.6: 32530190-32530191 on Build GRCh38
rs62405578 Chr.6: 32530349-32530349 on Build GRCh38
rs747906579 Chr.6: 32530207-32530208 on Build GRCh38
rs867037736 Chr.6: 32530463-32530463 on Build GRCh38
rs977632252 Chr.6: 32530089-32530089 on Build GRCh38
rs1141741 Chr.6: 32530127-32530127 on Build GRCh38
rs115699654 Chr.6: 32530477-32530477 on Build GRCh38
rs138720430 Chr.6: 32530447-32530448 on Build GRCh38
rs113810140 Chr.6: 32530086-32530086 on Build GRCh38
rs116033113 Chr.6: 32530394-32530394 on Build GRCh38
rs201200124 Chr.6: 32530079-32530079 on Build GRCh38
rs867811482 Chr.6: 32530033-32530033 on Build GRCh38
rs756226189 Chr.6: 32530255-32530255 on Build GRCh38
rs753751784 Chr.6: 32530202-32530202 on Build GRCh38
rs776278042 Chr.6: 32530235-32530235 on Build GRCh38
rs372992445 Chr.6: 32530271-32530271 on Build GRCh38
rs386699389 Chr.6: 32530264-32530265 on Build GRCh38
rs544606957 Chr.6: 32530312-32530312 on Build GRCh38
rs778524882 Chr.6: 32530088-32530088 on Build GRCh38
rs781507117 Chr.6: 32530055-32530055 on Build GRCh38
rs750434985 Chr.6: 32530250-32530250 on Build GRCh38
rs773341391 Chr.6: 32530228-32530229 on Build GRCh38
rs752268643 Chr.6: 32530090-32530090 on Build GRCh38
rs76748970 Chr.6: 32530137-32530137 on Build GRCh38
rs201644181 Chr.6: 32530193-32530193 on Build GRCh38
rs771581042 Chr.6: 32530229-32530229 on Build GRCh38
rs3179203 Chr.6: 32530234-32530234 on Build GRCh38
rs796616593 Chr.6: 32530376-32530376 on Build GRCh38
rs200358546 Chr.6: 32530110-32530110 on Build GRCh38
rs774541765 Chr.6: 32530108-32530108 on Build GRCh38
rs754268531 Chr.6: 32530120-32530120 on Build GRCh38
rs765098773 Chr.6: 32530216-32530216 on Build GRCh38
rs780335005 Chr.6: 32530207-32530207 on Build GRCh38
rs139583918 Chr.6: 32530133-32530133 on Build GRCh38
rs71549220 Chr.6: 32530140-32530140 on Build GRCh38
rs768842990 Chr.6: 32530076-32530076 on Build GRCh38
rs765902788 Chr.6: 32530273-32530274 on Build GRCh38
rs766547611 Chr.6: 32530228-32530231 on Build GRCh38
rs201497258 Chr.6: 32530374-32530374 on Build GRCh38
rs746139597 Chr.6: 32530206-32530206 on Build GRCh38
rs368120633 Chr.6: 32530157-32530157 on Build GRCh38
rs762631747 Chr.6: 32530470-32530471 on Build GRCh38
rs143038065 Chr.6: 32530321-32530322 on Build GRCh38
rs761060484 Chr.6: 32530084-32530084 on Build GRCh38
rs1064587 Chr.6: 32530184-32530184 on Build GRCh38
rs375258512 Chr.6: 32530261-32530261 on Build GRCh38
rs748566644 Chr.6: 32530258-32530258 on Build GRCh38
rs765707244 Chr.6: 32530159-32530159 on Build GRCh38
rs998190768 Chr.6: 32530103-32530103 on Build GRCh38
rs376008514 Chr.6: 32530239-32530239 on Build GRCh38
rs202110371 Chr.6: 32530100-32530100 on Build GRCh38
rs201883078 Chr.6: 32530211-32530211 on Build GRCh38
rs201863198 Chr.6: 32530294-32530294 on Build GRCh38
rs757350519 Chr.6: 32530217-32530217 on Build GRCh38
rs780712823 Chr.6: 32530183-32530184 on Build GRCh38
rs71549219 Chr.6: 32530128-32530128 on Build GRCh38
rs67598791 Chr.6: 32530290-32530290 on Build GRCh38
rs760263735 Chr.6: 32530232-32530232 on Build GRCh38
rs146966122 Chr.6: 32530136-32530136 on Build GRCh38

View Full Product Details