Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32935253-32935750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMB
Entrez Gene ID
3109
Gene Name
major histocompatibility complex, class II, DM beta
Gene Aliases
D6S221E, RING7
UniGene
Hs.351279
Gene Chromosome Location
Chr.6: 32934629-32941070 on Build GRCh38
Transcripts
NM_002118.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3625545 c.767A>T p.Y256F Chr.6: 32935350-32935350 on Build GRCh38
6424195 c.626C>T p.P209L Chr.6: 32935649-32935649 on Build GRCh38
3625544 c.769T>G p.S257A Chr.6: 32935348-32935348 on Build GRCh38
5565131 c.722G>A p.R241Q Chr.6: 32935553-32935553 on Build GRCh38
4505850 c.703C>T p.L235F Chr.6: 32935572-32935572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745508379 Chr.6: 32935417-32935417 on Build GRCh38
rs774065296 Chr.6: 32935628-32935628 on Build GRCh38
rs187493403 Chr.6: 32935504-32935504 on Build GRCh38
rs768453399 Chr.6: 32935611-32935611 on Build GRCh38
rs778237461 Chr.6: 32935553-32935553 on Build GRCh38
rs747488141 Chr.6: 32935559-32935559 on Build GRCh38
rs772997593 Chr.6: 32935632-32935632 on Build GRCh38
rs952673072 Chr.6: 32935286-32935286 on Build GRCh38
rs112252170 Chr.6: 32935309-32935309 on Build GRCh38
rs559886751 Chr.6: 32935694-32935694 on Build GRCh38
rs1020513349 Chr.6: 32935659-32935659 on Build GRCh38
rs764075421 Chr.6: 32935353-32935353 on Build GRCh38
rs752030287 Chr.6: 32935683-32935683 on Build GRCh38
rs1029891162 Chr.6: 32935714-32935714 on Build GRCh38
rs967095892 Chr.6: 32935333-32935333 on Build GRCh38
rs568672485 Chr.6: 32935436-32935436 on Build GRCh38
rs769554342 Chr.6: 32935488-32935488 on Build GRCh38
rs757550628 Chr.6: 32935686-32935686 on Build GRCh38
rs767546553 Chr.6: 32935631-32935631 on Build GRCh38
rs754021364 Chr.6: 32935369-32935369 on Build GRCh38
rs769513820 Chr.6: 32935549-32935549 on Build GRCh38
rs538995661 Chr.6: 32935461-32935461 on Build GRCh38
rs142046749 Chr.6: 32935635-32935635 on Build GRCh38
rs768306180 Chr.6: 32935703-32935704 on Build GRCh38
rs752932562 Chr.6: 32935547-32935547 on Build GRCh38
rs190433887 Chr.6: 32935525-32935525 on Build GRCh38
rs551307346 Chr.6: 32935640-32935640 on Build GRCh38
rs772546630 Chr.6: 32935389-32935389 on Build GRCh38
rs775164470 Chr.6: 32935596-32935596 on Build GRCh38
rs867964118 Chr.6: 32935352-32935353 on Build GRCh38
rs753394072 Chr.6: 32935636-32935636 on Build GRCh38
rs544034924 Chr.6: 32935578-32935578 on Build GRCh38
rs774294363 Chr.6: 32935512-32935512 on Build GRCh38
rs903426003 Chr.6: 32935595-32935595 on Build GRCh38
rs578046857 Chr.6: 32935459-32935459 on Build GRCh38
rs975161836 Chr.6: 32935703-32935703 on Build GRCh38
rs781548169 Chr.6: 32935575-32935575 on Build GRCh38
rs150265100 Chr.6: 32935696-32935696 on Build GRCh38
rs748584327 Chr.6: 32935382-32935382 on Build GRCh38
rs115136027 Chr.6: 32935403-32935403 on Build GRCh38
rs762538584 Chr.6: 32935693-32935694 on Build GRCh38
rs975214255 Chr.6: 32935452-32935452 on Build GRCh38
rs755652698 Chr.6: 32935700-32935700 on Build GRCh38
rs999784485 Chr.6: 32935681-32935681 on Build GRCh38
rs776146505 Chr.6: 32935409-32935409 on Build GRCh38
rs922494864 Chr.6: 32935465-32935465 on Build GRCh38
rs1020359287 Chr.6: 32935332-32935332 on Build GRCh38
rs762730059 Chr.6: 32935335-32935335 on Build GRCh38
rs982725792 Chr.6: 32935726-32935726 on Build GRCh38
rs1053462802 Chr.6: 32935464-32935464 on Build GRCh38
rs780133139 Chr.6: 32935368-32935368 on Build GRCh38
rs764560948 Chr.6: 32935680-32935680 on Build GRCh38
rs371915488 Chr.6: 32935524-32935524 on Build GRCh38
rs116189213 Chr.6: 32935294-32935294 on Build GRCh38
rs751475710 Chr.6: 32935354-32935354 on Build GRCh38
rs779275615 Chr.6: 32935372-32935372 on Build GRCh38
rs982742100 Chr.6: 32935513-32935513 on Build GRCh38
rs35339338 Chr.6: 32935635-32935636 on Build GRCh38
rs908372811 Chr.6: 32935296-32935296 on Build GRCh38
rs775109301 Chr.6: 32935492-32935492 on Build GRCh38
rs776921205 Chr.6: 32935333-32935334 on Build GRCh38
rs192597393 Chr.6: 32935548-32935548 on Build GRCh38
rs192338371 Chr.6: 32935517-32935517 on Build GRCh38
rs778468193 Chr.6: 32935392-32935392 on Build GRCh38
rs201379967 Chr.6: 32935329-32935329 on Build GRCh38
rs752420843 Chr.6: 32935315-32935315 on Build GRCh38
rs184737226 Chr.6: 32935519-32935519 on Build GRCh38
rs1026927736 Chr.6: 32935284-32935284 on Build GRCh38
rs759176315 Chr.6: 32935306-32935306 on Build GRCh38
rs758639050 Chr.6: 32935672-32935672 on Build GRCh38
rs148615873 Chr.6: 32935568-32935568 on Build GRCh38
rs372940202 Chr.6: 32935371-32935371 on Build GRCh38
rs770366284 Chr.6: 32935408-32935408 on Build GRCh38
rs764985732 Chr.6: 32935715-32935715 on Build GRCh38
rs374827604 Chr.6: 32935293-32935293 on Build GRCh38
rs567858022 Chr.6: 32935358-32935358 on Build GRCh38
rs1034522465 Chr.6: 32935291-32935291 on Build GRCh38
rs868386814 Chr.6: 32935351-32935352 on Build GRCh38
rs562209076 Chr.6: 32935601-32935601 on Build GRCh38
rs761224268 Chr.6: 32935531-32935531 on Build GRCh38
rs769273576 Chr.6: 32935585-32935585 on Build GRCh38
rs765601850 Chr.6: 32935544-32935544 on Build GRCh38
rs763844627 Chr.6: 32935508-32935508 on Build GRCh38
rs771325167 Chr.6: 32935331-32935333 on Build GRCh38
rs981965755 Chr.6: 32935314-32935314 on Build GRCh38

View Full Product Details