Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32938592-32939100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMB
Entrez Gene ID
3109
Gene Name
major histocompatibility complex, class II, DM beta
Gene Aliases
D6S221E, RING7
UniGene
Hs.351279
Gene Chromosome Location
Chr.6: 32934629-32941070 on Build GRCh38
Transcripts
NM_002118.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209634 c.140A>G p.N47S Chr.6: 32938881-32938881 on Build GRCh38
3374230 c.200G>A p.G67E Chr.6: 32938821-32938821 on Build GRCh38
6207680 c.162G>A p.W54* Chr.6: 32938859-32938859 on Build GRCh38
4830393 c.228C>G p.L76L Chr.6: 32938793-32938793 on Build GRCh38
3873048 c.253A>G p.T85A Chr.6: 32938768-32938768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770671320 Chr.6: 32939015-32939015 on Build GRCh38
rs1045763076 Chr.6: 32938802-32938802 on Build GRCh38
rs761448834 Chr.6: 32938854-32938854 on Build GRCh38
rs892565761 Chr.6: 32938828-32938828 on Build GRCh38
rs750367215 Chr.6: 32938718-32938719 on Build GRCh38
rs997535750 Chr.6: 32938835-32938835 on Build GRCh38
rs1048578337 Chr.6: 32938766-32938766 on Build GRCh38
rs371040681 Chr.6: 32938832-32938832 on Build GRCh38
rs780410761 Chr.6: 32938829-32938829 on Build GRCh38
rs771667092 Chr.6: 32938866-32938866 on Build GRCh38
rs774975956 Chr.6: 32938850-32938850 on Build GRCh38
rs535738804 Chr.6: 32939031-32939031 on Build GRCh38
rs569820867 Chr.6: 32938886-32938886 on Build GRCh38
rs142107957 Chr.6: 32938824-32938824 on Build GRCh38
rs150015309 Chr.6: 32938749-32938749 on Build GRCh38
rs781435121 Chr.6: 32938799-32938799 on Build GRCh38
rs944017978 Chr.6: 32938644-32938644 on Build GRCh38
rs552235689 Chr.6: 32938917-32938917 on Build GRCh38
rs762412768 Chr.6: 32938623-32938623 on Build GRCh38
rs1053054078 Chr.6: 32939062-32939062 on Build GRCh38
rs765726349 Chr.6: 32938618-32938618 on Build GRCh38
rs755915150 Chr.6: 32938661-32938661 on Build GRCh38
rs948408412 Chr.6: 32939021-32939021 on Build GRCh38
rs971919143 Chr.6: 32938669-32938669 on Build GRCh38
rs189147441 Chr.6: 32938898-32938898 on Build GRCh38
rs546277114 Chr.6: 32938619-32938619 on Build GRCh38
rs748479163 Chr.6: 32938841-32938841 on Build GRCh38
rs768294785 Chr.6: 32938747-32938747 on Build GRCh38
rs959794695 Chr.6: 32938983-32938983 on Build GRCh38
rs763469949 Chr.6: 32938745-32938745 on Build GRCh38
rs750241280 Chr.6: 32938652-32938652 on Build GRCh38
rs765211529 Chr.6: 32938893-32938893 on Build GRCh38
rs976741377 Chr.6: 32939072-32939072 on Build GRCh38
rs17583782 Chr.6: 32938939-32938939 on Build GRCh38
rs992544321 Chr.6: 32938992-32938992 on Build GRCh38
rs972335046 Chr.6: 32938949-32938949 on Build GRCh38
rs767585632 Chr.6: 32938919-32938919 on Build GRCh38
rs372802102 Chr.6: 32938868-32938868 on Build GRCh38
rs554463764 Chr.6: 32938788-32938788 on Build GRCh38
rs771718349 Chr.6: 32938957-32938957 on Build GRCh38
rs763658896 Chr.6: 32938628-32938628 on Build GRCh38
rs759944837 Chr.6: 32938613-32938613 on Build GRCh38
rs747233613 Chr.6: 32938704-32938704 on Build GRCh38
rs17617321 Chr.6: 32938809-32938809 on Build GRCh38
rs779893607 Chr.6: 32938676-32938676 on Build GRCh38
rs149133962 Chr.6: 32938984-32938984 on Build GRCh38
rs769354877 Chr.6: 32938848-32938848 on Build GRCh38
rs750626671 Chr.6: 32938930-32938930 on Build GRCh38
rs564449684 Chr.6: 32938706-32938706 on Build GRCh38
rs748615162 Chr.6: 32938948-32938948 on Build GRCh38
rs941890464 Chr.6: 32938662-32938662 on Build GRCh38
rs755243316 Chr.6: 32938839-32938839 on Build GRCh38
rs766145955 Chr.6: 32938758-32938758 on Build GRCh38
rs12176347 Chr.6: 32938803-32938803 on Build GRCh38
rs772708856 Chr.6: 32938975-32938975 on Build GRCh38
rs766972707 Chr.6: 32938702-32938703 on Build GRCh38
rs760417940 Chr.6: 32938755-32938755 on Build GRCh38
rs191286550 Chr.6: 32938934-32938934 on Build GRCh38
rs751281050 Chr.6: 32938639-32938639 on Build GRCh38
rs766090049 Chr.6: 32938880-32938880 on Build GRCh38
rs183828064 Chr.6: 32939029-32939029 on Build GRCh38
rs953215631 Chr.6: 32939024-32939024 on Build GRCh38
rs756912572 Chr.6: 32938641-32938641 on Build GRCh38
rs749764146 Chr.6: 32938834-32938834 on Build GRCh38
rs41560814 Chr.6: 32938887-32938887 on Build GRCh38
rs17617333 Chr.6: 32938875-32938875 on Build GRCh38
rs1001466428 Chr.6: 32938821-32938821 on Build GRCh38
rs547721366 Chr.6: 32938700-32938700 on Build GRCh38
rs936910735 Chr.6: 32939050-32939050 on Build GRCh38
rs779593152 Chr.6: 32938940-32938940 on Build GRCh38
rs111568424 Chr.6: 32938808-32938808 on Build GRCh38
rs758427530 Chr.6: 32938697-32938697 on Build GRCh38
rs775893880 Chr.6: 32938721-32938721 on Build GRCh38
rs763641942 Chr.6: 32938891-32938891 on Build GRCh38
rs757308409 Chr.6: 32938794-32938794 on Build GRCh38
rs112179266 Chr.6: 32938938-32938938 on Build GRCh38
rs143112264 Chr.6: 32938712-32938712 on Build GRCh38
rs966396906 Chr.6: 32939052-32939052 on Build GRCh38
rs778814079 Chr.6: 32938769-32938769 on Build GRCh38
rs767327421 Chr.6: 32938750-32938750 on Build GRCh38
rs760583941 Chr.6: 32939017-32939020 on Build GRCh38
rs751699077 Chr.6: 32938775-32938775 on Build GRCh38
rs754838716 Chr.6: 32938767-32938767 on Build GRCh38
rs767425395 Chr.6: 32938643-32938643 on Build GRCh38
rs770166673 Chr.6: 32938718-32938718 on Build GRCh38
rs116782913 Chr.6: 32939033-32939033 on Build GRCh38
rs780993274 Chr.6: 32939054-32939056 on Build GRCh38
rs780135672 Chr.6: 32938937-32938937 on Build GRCh38
rs112725763 Chr.6: 32938874-32938874 on Build GRCh38
rs148084922 Chr.6: 32938684-32938684 on Build GRCh38
rs145151976 Chr.6: 32938754-32938754 on Build GRCh38

View Full Product Details