Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33165313-33165856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A2
Entrez Gene ID
1302
Gene Name
collagen type XI alpha 2
Gene Aliases
DFNA13, DFNB53, FBCG2, HKE5, PARP, STL3
UniGene
Hs.390171
Gene Chromosome Location
Chr.6: 33162692-33193009 on Build GRCh38
Transcripts
NM_080679.2 NM_080680.2 NM_080681.2 XM_011514299.2 XM_011514300.2 XM_011514302.2 XM_017010250.1 XM_017010251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6632369 c.4652G>T p.R1551L Chr.6: 33165647-33165647 on Build GRCh38
6374992 c.4499T>G p.V1500G Chr.6: 33165800-33165800 on Build GRCh38
7214587 c.4483G>A p.G1495S Chr.6: 33165816-33165816 on Build GRCh38
5528919 c.4615G>A p.G1539R Chr.6: 33165684-33165684 on Build GRCh38
3349298 c.4579G>A p.A1527T Chr.6: 33165720-33165720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765931315 Chr.6: 33165712-33165712 on Build GRCh38
rs772312309 Chr.6: 33165783-33165783 on Build GRCh38
rs920408620 Chr.6: 33165421-33165421 on Build GRCh38
rs934714603 Chr.6: 33165741-33165741 on Build GRCh38
rs1046161149 Chr.6: 33165530-33165530 on Build GRCh38
rs751435751 Chr.6: 33165757-33165757 on Build GRCh38
rs765304181 Chr.6: 33165682-33165682 on Build GRCh38
rs757289137 Chr.6: 33165569-33165569 on Build GRCh38
rs907100612 Chr.6: 33165643-33165643 on Build GRCh38
rs2855454 Chr.6: 33165371-33165371 on Build GRCh38
rs114580597 Chr.6: 33165778-33165778 on Build GRCh38
rs772567850 Chr.6: 33165740-33165740 on Build GRCh38
rs773235349 Chr.6: 33165553-33165553 on Build GRCh38
rs758306963 Chr.6: 33165658-33165658 on Build GRCh38
rs778972205 Chr.6: 33165765-33165765 on Build GRCh38
rs370966667 Chr.6: 33165624-33165624 on Build GRCh38
rs547623894 Chr.6: 33165391-33165391 on Build GRCh38
rs745996561 Chr.6: 33165687-33165687 on Build GRCh38
rs141254777 Chr.6: 33165648-33165648 on Build GRCh38
rs137923568 Chr.6: 33165792-33165792 on Build GRCh38
rs755255647 Chr.6: 33165673-33165675 on Build GRCh38
rs762861658 Chr.6: 33165813-33165813 on Build GRCh38
rs779832829 Chr.6: 33165551-33165551 on Build GRCh38
rs754110017 Chr.6: 33165532-33165533 on Build GRCh38
rs766336073 Chr.6: 33165530-33165530 on Build GRCh38
rs200421970 Chr.6: 33165735-33165735 on Build GRCh38
rs759223182 Chr.6: 33165717-33165717 on Build GRCh38
rs377656039 Chr.6: 33165780-33165780 on Build GRCh38
rs371674362 Chr.6: 33165761-33165761 on Build GRCh38
rs779052773 Chr.6: 33165558-33165558 on Build GRCh38
rs28634640 Chr.6: 33165650-33165650 on Build GRCh38
rs375329541 Chr.6: 33165709-33165709 on Build GRCh38
rs953488295 Chr.6: 33165390-33165390 on Build GRCh38
rs966128013 Chr.6: 33165497-33165497 on Build GRCh38
rs780309318 Chr.6: 33165688-33165688 on Build GRCh38
rs752607868 Chr.6: 33165652-33165652 on Build GRCh38
rs759814552 Chr.6: 33165700-33165700 on Build GRCh38
rs201315111 Chr.6: 33165713-33165713 on Build GRCh38
rs749716412 Chr.6: 33165593-33165593 on Build GRCh38
rs930517120 Chr.6: 33165489-33165489 on Build GRCh38
rs903180840 Chr.6: 33165726-33165726 on Build GRCh38
rs145343609 Chr.6: 33165647-33165647 on Build GRCh38
rs775657425 Chr.6: 33165582-33165582 on Build GRCh38
rs761247230 Chr.6: 33165500-33165500 on Build GRCh38
rs199615717 Chr.6: 33165762-33165762 on Build GRCh38
rs770100834 Chr.6: 33165579-33165579 on Build GRCh38
rs780071501 Chr.6: 33165759-33165759 on Build GRCh38
rs143390157 Chr.6: 33165806-33165806 on Build GRCh38
rs370159193 Chr.6: 33165503-33165503 on Build GRCh38
rs566171449 Chr.6: 33165708-33165708 on Build GRCh38
rs370282196 Chr.6: 33165657-33165657 on Build GRCh38
rs576235181 Chr.6: 33165805-33165805 on Build GRCh38
rs34055850 Chr.6: 33165616-33165616 on Build GRCh38
rs760292409 Chr.6: 33165710-33165710 on Build GRCh38
rs756194499 Chr.6: 33165758-33165758 on Build GRCh38
rs368488233 Chr.6: 33165645-33165645 on Build GRCh38
rs748411665 Chr.6: 33165358-33165358 on Build GRCh38
rs748059729 Chr.6: 33165562-33165562 on Build GRCh38
rs770594261 Chr.6: 33165798-33165798 on Build GRCh38
rs565891596 Chr.6: 33165528-33165528 on Build GRCh38
rs727504543 Chr.6: 33165804-33165804 on Build GRCh38
rs764154647 Chr.6: 33165755-33165755 on Build GRCh38
rs781289663 Chr.6: 33165573-33165573 on Build GRCh38
rs748442759 Chr.6: 33165701-33165701 on Build GRCh38
rs772829153 Chr.6: 33165636-33165636 on Build GRCh38
rs367885056 Chr.6: 33165754-33165754 on Build GRCh38
rs919038954 Chr.6: 33165508-33165508 on Build GRCh38
rs749448524 Chr.6: 33165696-33165696 on Build GRCh38
rs763020874 Chr.6: 33165604-33165604 on Build GRCh38
rs529045062 Chr.6: 33165377-33165377 on Build GRCh38
rs1011115366 Chr.6: 33165354-33165354 on Build GRCh38
rs868805420 Chr.6: 33165746-33165746 on Build GRCh38
rs1055629015 Chr.6: 33165491-33165491 on Build GRCh38
rs753728183 Chr.6: 33165552-33165552 on Build GRCh38
rs891320028 Chr.6: 33165819-33165819 on Build GRCh38
rs774980862 Chr.6: 33165699-33165699 on Build GRCh38
rs200374341 Chr.6: 33165479-33165479 on Build GRCh38
rs774845879 Chr.6: 33165623-33165623 on Build GRCh38
rs763735245 Chr.6: 33165818-33165818 on Build GRCh38
rs267600980 Chr.6: 33165750-33165750 on Build GRCh38
rs746100166 Chr.6: 33165574-33165574 on Build GRCh38
rs751549851 Chr.6: 33165825-33165825 on Build GRCh38
rs766962425 Chr.6: 33165634-33165634 on Build GRCh38
rs375518774 Chr.6: 33165637-33165637 on Build GRCh38
rs727504458 Chr.6: 33165594-33165594 on Build GRCh38
rs778359244 Chr.6: 33165789-33165789 on Build GRCh38

View Full Product Details