Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33167172-33167680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A2
Entrez Gene ID
1302
Gene Name
collagen type XI alpha 2
Gene Aliases
DFNA13, DFNB53, FBCG2, HKE5, PARP, STL3
UniGene
Hs.390171
Gene Chromosome Location
Chr.6: 33162692-33193009 on Build GRCh38
Transcripts
NM_080679.2 NM_080680.2 NM_080681.2 XM_011514299.2 XM_011514300.2 XM_011514302.2 XM_017010250.1 XM_017010251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5965765 c.4015-3C>T p.? Chr.6: 33167536-33167536 on Build GRCh38
4899625 c.4087G>A p.G1363S Chr.6: 33167461-33167461 on Build GRCh38
4517723 c.4043_4044GG>AA p.G1348E Chr.6: 33167504-33167505 on Build GRCh38
4893774 c.4126C>T p.Q1376* Chr.6: 33167314-33167314 on Build GRCh38
3873089 c.4055C>T p.P1352L Chr.6: 33167493-33167493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928216125 Chr.6: 33167625-33167625 on Build GRCh38
rs748869320 Chr.6: 33167568-33167569 on Build GRCh38
rs377001136 Chr.6: 33167541-33167541 on Build GRCh38
rs753728330 Chr.6: 33167359-33167359 on Build GRCh38
rs1025659188 Chr.6: 33167536-33167536 on Build GRCh38
rs912957707 Chr.6: 33167656-33167656 on Build GRCh38
rs759017753 Chr.6: 33167521-33167521 on Build GRCh38
rs766110293 Chr.6: 33167343-33167343 on Build GRCh38
rs931440799 Chr.6: 33167270-33167270 on Build GRCh38
rs34438240 Chr.6: 33167614-33167615 on Build GRCh38
rs141341019 Chr.6: 33167302-33167302 on Build GRCh38
rs760358440 Chr.6: 33167337-33167337 on Build GRCh38
rs775196432 Chr.6: 33167556-33167556 on Build GRCh38
rs370381722 Chr.6: 33167392-33167392 on Build GRCh38
rs771417043 Chr.6: 33167331-33167331 on Build GRCh38
rs754994197 Chr.6: 33167227-33167227 on Build GRCh38
rs773033938 Chr.6: 33167335-33167335 on Build GRCh38
rs780346273 Chr.6: 33167301-33167301 on Build GRCh38
rs146288830 Chr.6: 33167619-33167619 on Build GRCh38
rs536381402 Chr.6: 33167371-33167371 on Build GRCh38
rs537455619 Chr.6: 33167468-33167468 on Build GRCh38
rs377598435 Chr.6: 33167217-33167217 on Build GRCh38
rs749721893 Chr.6: 33167397-33167397 on Build GRCh38
rs2855449 Chr.6: 33167615-33167615 on Build GRCh38
rs938323070 Chr.6: 33167647-33167647 on Build GRCh38
rs1052789902 Chr.6: 33167649-33167649 on Build GRCh38
rs1057344653 Chr.6: 33167547-33167547 on Build GRCh38
rs2855450 Chr.6: 33167614-33167614 on Build GRCh38
rs553576969 Chr.6: 33167546-33167546 on Build GRCh38
rs746019545 Chr.6: 33167291-33167291 on Build GRCh38
rs748527563 Chr.6: 33167510-33167510 on Build GRCh38
rs779479651 Chr.6: 33167436-33167436 on Build GRCh38
rs1006789951 Chr.6: 33167499-33167499 on Build GRCh38
rs756180267 Chr.6: 33167501-33167501 on Build GRCh38
rs772910810 Chr.6: 33167448-33167448 on Build GRCh38
rs750541299 Chr.6: 33167494-33167494 on Build GRCh38
rs781209055 Chr.6: 33167382-33167382 on Build GRCh38
rs774218600 Chr.6: 33167576-33167576 on Build GRCh38
rs774709598 Chr.6: 33167306-33167306 on Build GRCh38
rs756376471 Chr.6: 33167388-33167388 on Build GRCh38
rs760749775 Chr.6: 33167580-33167580 on Build GRCh38
rs769169475 Chr.6: 33167492-33167492 on Build GRCh38
rs764870047 Chr.6: 33167482-33167482 on Build GRCh38
rs757267009 Chr.6: 33167240-33167240 on Build GRCh38
rs759385460 Chr.6: 33167481-33167481 on Build GRCh38
rs916800594 Chr.6: 33167333-33167333 on Build GRCh38
rs569997660 Chr.6: 33167435-33167435 on Build GRCh38
rs368543321 Chr.6: 33167441-33167441 on Build GRCh38
rs762703466 Chr.6: 33167564-33167564 on Build GRCh38
rs370994730 Chr.6: 33167581-33167581 on Build GRCh38
rs765273820 Chr.6: 33167362-33167362 on Build GRCh38
rs150784358 Chr.6: 33167314-33167314 on Build GRCh38
rs868061872 Chr.6: 33167320-33167320 on Build GRCh38
rs752515977 Chr.6: 33167377-33167377 on Build GRCh38
rs2855451 Chr.6: 33167401-33167401 on Build GRCh38
rs750665589 Chr.6: 33167383-33167383 on Build GRCh38
rs879600791 Chr.6: 33167614-33167615 on Build GRCh38
rs781600736 Chr.6: 33167551-33167551 on Build GRCh38
rs139283268 Chr.6: 33167507-33167507 on Build GRCh38
rs121912950 Chr.6: 33167305-33167305 on Build GRCh38
rs778686103 Chr.6: 33167229-33167229 on Build GRCh38
rs759244932 Chr.6: 33167360-33167360 on Build GRCh38
rs770175568 Chr.6: 33167292-33167292 on Build GRCh38
rs2855452 Chr.6: 33167381-33167381 on Build GRCh38
rs142890313 Chr.6: 33167508-33167508 on Build GRCh38
rs747449291 Chr.6: 33167458-33167458 on Build GRCh38
rs780270414 Chr.6: 33167504-33167504 on Build GRCh38
rs760233846 Chr.6: 33167459-33167459 on Build GRCh38
rs181895110 Chr.6: 33167304-33167304 on Build GRCh38
rs764083477 Chr.6: 33167572-33167572 on Build GRCh38
rs776406862 Chr.6: 33167474-33167474 on Build GRCh38
rs778423110 Chr.6: 33167522-33167522 on Build GRCh38
rs747177284 Chr.6: 33167234-33167234 on Build GRCh38
rs1030750495 Chr.6: 33167465-33167465 on Build GRCh38
rs1009989757 Chr.6: 33167341-33167341 on Build GRCh38
rs763995767 Chr.6: 33167493-33167493 on Build GRCh38
rs769652523 Chr.6: 33167551-33167551 on Build GRCh38
rs752706766 Chr.6: 33167491-33167491 on Build GRCh38
rs770504397 Chr.6: 33167544-33167544 on Build GRCh38
rs753849434 Chr.6: 33167220-33167220 on Build GRCh38
rs755981662 Chr.6: 33167214-33167214 on Build GRCh38
rs904254101 Chr.6: 33167617-33167617 on Build GRCh38
rs189177654 Chr.6: 33167200-33167200 on Build GRCh38
rs781402333 Chr.6: 33167274-33167274 on Build GRCh38

View Full Product Details